Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 16962776-16963045 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAB4
Entrez Gene ID
128954
Gene Name
GRB2 associated binding protein family member 4
Gene Aliases
-
UniGene
Hs.434113
Gene Chromosome Location
Chr.22: 16961936-17008281 on Build GRCh38
Transcripts
NM_001037814.1 XM_011546114.2 XM_011546115.1 XM_011546117.1 XM_017028575.1 XM_017028576.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1031820 c.1650G>A p.Q550Q Chr.22: 16962808-16962808 on Build GRCh38
4795050 c.1591G>A p.G531S Chr.22: 16962867-16962867 on Build GRCh38
4400913 c.1640A>C p.E547A Chr.22: 16962818-16962818 on Build GRCh38
7156452 c.1644G>A p.K548K Chr.22: 16962814-16962814 on Build GRCh38
3551913 c.1639G>A p.E547K Chr.22: 16962819-16962819 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199986323 Chr.22: 16962835-16962835 on Build GRCh38
rs946467648 Chr.22: 16962901-16962901 on Build GRCh38
rs757826410 Chr.22: 16962822-16962822 on Build GRCh38
rs768612510 Chr.22: 16962863-16962863 on Build GRCh38
rs556225570 Chr.22: 16962982-16962982 on Build GRCh38
rs753647252 Chr.22: 16962802-16962802 on Build GRCh38
rs761883071 Chr.22: 16962872-16962872 on Build GRCh38
rs374940310 Chr.22: 16962926-16962926 on Build GRCh38
rs749690543 Chr.22: 16962907-16962907 on Build GRCh38
rs376254655 Chr.22: 16962861-16962861 on Build GRCh38
rs774246746 Chr.22: 16962912-16962912 on Build GRCh38
rs202160268 Chr.22: 16962898-16962898 on Build GRCh38
rs112963949 Chr.22: 16963019-16963019 on Build GRCh38
rs754860854 Chr.22: 16962809-16962809 on Build GRCh38
rs929180317 Chr.22: 16962978-16962978 on Build GRCh38
rs770267215 Chr.22: 16962906-16962906 on Build GRCh38
rs9606551 Chr.22: 16962919-16962919 on Build GRCh38
rs751098035 Chr.22: 16962900-16962900 on Build GRCh38
rs566406835 Chr.22: 16962973-16962973 on Build GRCh38
rs533867448 Chr.22: 16962975-16962975 on Build GRCh38
rs538048925 Chr.22: 16963016-16963016 on Build GRCh38
rs769217210 Chr.22: 16962909-16962909 on Build GRCh38
rs374466820 Chr.22: 16962815-16962815 on Build GRCh38
rs770624257 Chr.22: 16962849-16962849 on Build GRCh38
rs781357923 Chr.22: 16962852-16962852 on Build GRCh38
rs770318451 Chr.22: 16962857-16962857 on Build GRCh38
rs913541210 Chr.22: 16962856-16962856 on Build GRCh38
rs1005492509 Chr.22: 16963014-16963014 on Build GRCh38
rs777401214 Chr.22: 16962831-16962831 on Build GRCh38
rs371940524 Chr.22: 16962881-16962881 on Build GRCh38
rs758430545 Chr.22: 16962897-16962897 on Build GRCh38
rs371868434 Chr.22: 16962853-16962853 on Build GRCh38
rs745560438 Chr.22: 16962905-16962905 on Build GRCh38
rs186926347 Chr.22: 16962991-16962991 on Build GRCh38
rs556105573 Chr.22: 16962948-16962948 on Build GRCh38
rs145690904 Chr.22: 16963010-16963010 on Build GRCh38
rs780957711 Chr.22: 16962903-16962903 on Build GRCh38
rs774492344 Chr.22: 16962864-16962864 on Build GRCh38
rs539472836 Chr.22: 16962936-16962936 on Build GRCh38
rs767802003 Chr.22: 16962875-16962875 on Build GRCh38
rs751344412 Chr.22: 16962883-16962883 on Build GRCh38
rs1056699127 Chr.22: 16962934-16962934 on Build GRCh38
rs766517716 Chr.22: 16962882-16962882 on Build GRCh38

View Full Product Details