Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 41361124-41361389 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EMILIN3
Entrez Gene ID
90187
Gene Name
elastin microfibril interfacer 3
Gene Aliases
C20orf130, EMILIN5, dJ620E11.4
UniGene
Hs.726525
Gene Chromosome Location
Chr.20: 41359966-41366858 on Build GRCh38
Transcripts
NM_052846.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5718106 c.2297A>T p.N766I Chr.20: 41361272-41361272 on Build GRCh38
4098425 c.2227G>C p.A743P Chr.20: 41361342-41361342 on Build GRCh38
6284957 c.2200A>G p.T734A Chr.20: 41361369-41361369 on Build GRCh38
4885699 c.2190G>T p.Q730H Chr.20: 41361379-41361379 on Build GRCh38
7338204 c.2221G>A p.D741N Chr.20: 41361348-41361348 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756555880 Chr.20: 41361155-41361155 on Build GRCh38
rs201476427 Chr.20: 41361243-41361243 on Build GRCh38
rs753972634 Chr.20: 41361330-41361330 on Build GRCh38
rs779427545 Chr.20: 41361333-41361333 on Build GRCh38
rs756398887 Chr.20: 41361297-41361297 on Build GRCh38
rs769422578 Chr.20: 41361259-41361259 on Build GRCh38
rs746152128 Chr.20: 41361337-41361337 on Build GRCh38
rs772799961 Chr.20: 41361353-41361353 on Build GRCh38
rs756228532 Chr.20: 41361236-41361236 on Build GRCh38
rs747707503 Chr.20: 41361311-41361312 on Build GRCh38
rs766440457 Chr.20: 41361279-41361279 on Build GRCh38
rs2235593 Chr.20: 41361228-41361228 on Build GRCh38
rs752947764 Chr.20: 41361292-41361292 on Build GRCh38
rs777889960 Chr.20: 41361237-41361237 on Build GRCh38
rs767575497 Chr.20: 41361290-41361290 on Build GRCh38
rs141515546 Chr.20: 41361184-41361184 on Build GRCh38
rs147157827 Chr.20: 41361287-41361287 on Build GRCh38
rs565723351 Chr.20: 41361348-41361348 on Build GRCh38
rs369162593 Chr.20: 41361288-41361288 on Build GRCh38
rs780341244 Chr.20: 41361339-41361339 on Build GRCh38
rs996629077 Chr.20: 41361231-41361231 on Build GRCh38
rs768154629 Chr.20: 41361245-41361245 on Build GRCh38
rs200439419 Chr.20: 41361338-41361338 on Build GRCh38
rs749794025 Chr.20: 41361238-41361238 on Build GRCh38
rs373038247 Chr.20: 41361305-41361305 on Build GRCh38
rs764130908 Chr.20: 41361307-41361307 on Build GRCh38
rs755105740 Chr.20: 41361220-41361220 on Build GRCh38
rs943563105 Chr.20: 41361309-41361309 on Build GRCh38
rs199725822 Chr.20: 41361278-41361278 on Build GRCh38
rs767343971 Chr.20: 41361368-41361368 on Build GRCh38
rs754426868 Chr.20: 41361332-41361332 on Build GRCh38
rs917691923 Chr.20: 41361200-41361200 on Build GRCh38
rs57558294 Chr.20: 41361367-41361367 on Build GRCh38
rs781504124 Chr.20: 41361227-41361227 on Build GRCh38
rs991907992 Chr.20: 41361190-41361190 on Build GRCh38
rs776352685 Chr.20: 41361247-41361247 on Build GRCh38
rs779231459 Chr.20: 41361240-41361240 on Build GRCh38
rs199818591 Chr.20: 41361355-41361355 on Build GRCh38
rs999921860 Chr.20: 41361181-41361181 on Build GRCh38
rs769617233 Chr.20: 41361352-41361352 on Build GRCh38
rs959490542 Chr.20: 41361164-41361164 on Build GRCh38
rs760592769 Chr.20: 41361216-41361216 on Build GRCh38
rs757805112 Chr.20: 41361239-41361239 on Build GRCh38
rs141934538 Chr.20: 41361358-41361358 on Build GRCh38
rs748837837 Chr.20: 41361354-41361354 on Build GRCh38

View Full Product Details