Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 5395526-5395795 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPB41L3
Entrez Gene ID
23136
Gene Name
erythrocyte membrane protein band 4.1 like 3
Gene Aliases
4.1B, DAL-1, DAL1
UniGene
Hs.213394
Gene Chromosome Location
Chr.18: 5392381-5630666 on Build GRCh38
Transcripts
NM_001281533.1 NM_001281534.1 NM_001281535.1 NM_012307.3 XM_011525623.2 XM_011525626.2 XM_011525628.2 XM_011525632.2 XM_011525634.2 XM_011525637.2 XM_017025618.1 XM_017025619.1 XM_017025620.1 XM_017025621.1 XM_017025622.1 XM_017025623.1 XM_017025624.1 XM_017025625.1 XM_017025626.1 XM_017025627.1 XM_017025628.1 XM_017025629.1 XM_017025630.1 XM_017025631.1 XM_017025632.1 XM_017025633.1 XM_017025634.1 XM_017025635.1 XM_017025636.1 XM_017025637.1 XM_017025638.1 XM_017025639.1 XM_017025640.1 XM_017025641.1 XM_017025642.1 XM_017025643.1 XM_017025644.1 XM_017025645.1 XM_017025646.1 XM_017025647.1 XM_017025648.1 XM_017025649.1 XM_017025650.1 XM_017025651.1 XM_017025652.1 XM_017025653.1 XM_017025654.1 XM_017025655.1 XM_017025656.1 XM_017025657.1 XM_017025658.1 XM_017025659.1 XM_017025660.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
306970 c.3048C>A p.T1016T Chr.18: 5395633-5395633 on Build GRCh38
989056 c.3004G>A p.V1002M Chr.18: 5395677-5395677 on Build GRCh38
4681177 c.2552C>T p.T851M Chr.18: 5395622-5395622 on Build GRCh38
989055 c.3039C>T p.T1013T Chr.18: 5395642-5395642 on Build GRCh38
5930352 c.2540C>T p.T847I Chr.18: 5395634-5395634 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375353039 Chr.18: 5395621-5395621 on Build GRCh38
rs760444006 Chr.18: 5395628-5395628 on Build GRCh38
rs747293245 Chr.18: 5395591-5395591 on Build GRCh38
rs527461275 Chr.18: 5395622-5395622 on Build GRCh38
rs918389975 Chr.18: 5395636-5395636 on Build GRCh38
rs772448407 Chr.18: 5395752-5395753 on Build GRCh38
rs775649612 Chr.18: 5395766-5395766 on Build GRCh38
rs149824211 Chr.18: 5395678-5395678 on Build GRCh38
rs568894598 Chr.18: 5395723-5395723 on Build GRCh38
rs765087190 Chr.18: 5395659-5395659 on Build GRCh38
rs202000684 Chr.18: 5395618-5395618 on Build GRCh38
rs766104072 Chr.18: 5395638-5395638 on Build GRCh38
rs758321771 Chr.18: 5395677-5395677 on Build GRCh38
rs776277528 Chr.18: 5395728-5395728 on Build GRCh38
rs116148637 Chr.18: 5395705-5395705 on Build GRCh38
rs200995918 Chr.18: 5395592-5395592 on Build GRCh38
rs1039939337 Chr.18: 5395567-5395567 on Build GRCh38
rs752463033 Chr.18: 5395581-5395581 on Build GRCh38
rs970665309 Chr.18: 5395729-5395729 on Build GRCh38
rs777626371 Chr.18: 5395660-5395660 on Build GRCh38
rs746457311 Chr.18: 5395749-5395750 on Build GRCh38
rs772822446 Chr.18: 5395744-5395745 on Build GRCh38
rs144613208 Chr.18: 5395690-5395690 on Build GRCh38
rs368249382 Chr.18: 5395672-5395672 on Build GRCh38
rs1016287264 Chr.18: 5395582-5395582 on Build GRCh38
rs774862899 Chr.18: 5395742-5395742 on Build GRCh38
rs745967198 Chr.18: 5395702-5395702 on Build GRCh38
rs754916797 Chr.18: 5395576-5395576 on Build GRCh38
rs935028147 Chr.18: 5395730-5395730 on Build GRCh38
rs748976507 Chr.18: 5395685-5395685 on Build GRCh38
rs757007794 Chr.18: 5395752-5395752 on Build GRCh38
rs745939800 Chr.18: 5395611-5395611 on Build GRCh38
rs200474691 Chr.18: 5395609-5395609 on Build GRCh38
rs933501842 Chr.18: 5395762-5395762 on Build GRCh38
rs1012123253 Chr.18: 5395692-5395692 on Build GRCh38
rs766053032 Chr.18: 5395568-5395568 on Build GRCh38
rs769447084 Chr.18: 5395744-5395751 on Build GRCh38
rs753306885 Chr.18: 5395651-5395651 on Build GRCh38
rs753356631 Chr.18: 5395569-5395569 on Build GRCh38
rs764862372 Chr.18: 5395738-5395738 on Build GRCh38
rs201833386 Chr.18: 5395561-5395561 on Build GRCh38
rs903290579 Chr.18: 5395639-5395639 on Build GRCh38
rs547679814 Chr.18: 5395657-5395657 on Build GRCh38
rs145834920 Chr.18: 5395704-5395704 on Build GRCh38
rs747760001 Chr.18: 5395743-5395744 on Build GRCh38
rs778858300 Chr.18: 5395580-5395580 on Build GRCh38
rs758125793 Chr.18: 5395588-5395588 on Build GRCh38
rs200212387 Chr.18: 5395751-5395751 on Build GRCh38
rs777987704 Chr.18: 5395590-5395590 on Build GRCh38
rs762922325 Chr.18: 5395748-5395748 on Build GRCh38
rs367734348 Chr.18: 5395722-5395722 on Build GRCh38
rs149003216 Chr.18: 5395731-5395731 on Build GRCh38
rs143833943 Chr.18: 5395756-5395756 on Build GRCh38
rs770737016 Chr.18: 5395725-5395725 on Build GRCh38

View Full Product Details