Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 76022283-76022556 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ART3
Entrez Gene ID
419
Gene Name
ADP-ribosyltransferase 3
Gene Aliases
ARTC3
UniGene
Hs.24976
Gene Chromosome Location
Chr.4: 76011180-76112802 on Build GRCh38
Transcripts
NM_001130017.2 NM_001565.3

Gene Symbol
CXCL10
Entrez Gene ID
3627
Gene Name
C-X-C motif chemokine ligand 10
Gene Aliases
C7, IFI10, INP10, IP-10, SCYB10, crg-2, gIP-10, mob-1
UniGene
Hs.632586
Gene Chromosome Location
Chr.4: 76021116-76023536 on Build GRCh38
Transcripts
NM_001130017.2 NM_001565.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1431049 c.236C>T p.S79L Chr.4: 76022408-76022408 on Build GRCh38
260336 c.275A>T p.E92V Chr.4: 76022369-76022369 on Build GRCh38
6495777 c.249G>C p.K83N Chr.4: 76022395-76022395 on Build GRCh38
4478448 c.223C>T p.L75L Chr.4: 76022421-76022421 on Build GRCh38
1057617 c.249G>T p.K83N Chr.4: 76022395-76022395 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs548356549 Chr.4: 76022521-76022521 on Build GRCh38
rs573246567 Chr.4: 76022319-76022319 on Build GRCh38
rs367836661 Chr.4: 76022442-76022442 on Build GRCh38
rs774712838 Chr.4: 76022496-76022496 on Build GRCh38
rs1054124819 Chr.4: 76022415-76022415 on Build GRCh38
rs751919587 Chr.4: 76022384-76022384 on Build GRCh38
rs530928514 Chr.4: 76022487-76022487 on Build GRCh38
rs532476354 Chr.4: 76022406-76022406 on Build GRCh38
rs4859588 Chr.4: 76022524-76022524 on Build GRCh38
rs1050675273 Chr.4: 76022312-76022312 on Build GRCh38
rs749860619 Chr.4: 76022490-76022490 on Build GRCh38
rs759597737 Chr.4: 76022327-76022327 on Build GRCh38
rs761513968 Chr.4: 76022392-76022392 on Build GRCh38
rs766249571 Chr.4: 76022420-76022420 on Build GRCh38
rs745960921 Chr.4: 76022479-76022479 on Build GRCh38
rs1034650628 Chr.4: 76022517-76022517 on Build GRCh38
rs754930621 Chr.4: 76022439-76022439 on Build GRCh38
rs913918098 Chr.4: 76022511-76022511 on Build GRCh38
rs995105076 Chr.4: 76022433-76022433 on Build GRCh38
rs201393081 Chr.4: 76022323-76022323 on Build GRCh38
rs371479176 Chr.4: 76022352-76022352 on Build GRCh38
rs781438630 Chr.4: 76022471-76022471 on Build GRCh38
rs767185896 Chr.4: 76022503-76022503 on Build GRCh38
rs562734842 Chr.4: 76022448-76022448 on Build GRCh38
rs372262736 Chr.4: 76022465-76022465 on Build GRCh38
rs935617551 Chr.4: 76022414-76022414 on Build GRCh38
rs764482738 Chr.4: 76022374-76022374 on Build GRCh38
rs767097929 Chr.4: 76022400-76022400 on Build GRCh38
rs769968450 Chr.4: 76022482-76022482 on Build GRCh38
rs1010023789 Chr.4: 76022458-76022458 on Build GRCh38
rs757560315 Chr.4: 76022469-76022469 on Build GRCh38
rs755610178 Chr.4: 76022408-76022408 on Build GRCh38
rs771270907 Chr.4: 76022322-76022322 on Build GRCh38
rs139813823 Chr.4: 76022401-76022401 on Build GRCh38
rs762097270 Chr.4: 76022498-76022498 on Build GRCh38
rs188708798 Chr.4: 76022492-76022492 on Build GRCh38
rs368036811 Chr.4: 76022338-76022338 on Build GRCh38
rs763347756 Chr.4: 76022370-76022370 on Build GRCh38

View Full Product Details