Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 3290020-3290291 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CELF5
Entrez Gene ID
60680
Gene Name
CUGBP, Elav-like family member 5
Gene Aliases
BRUNOL-5, BRUNOL5, CELF-5
UniGene
Hs.655747
Gene Chromosome Location
Chr.19: 3224676-3297076 on Build GRCh38
Transcripts
NM_001172673.1 NM_021938.3 NR_033342.1 XM_006722832.1 XM_006722833.1 XM_006722836.2 XM_006722837.3 XM_006722838.3 XM_011528173.1 XM_011528174.2 XM_017027111.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1248043 c.1224C>A p.P408P Chr.19: 3290268-3290268 on Build GRCh38
4832247 c.1228G>T p.E410* Chr.19: 3290272-3290272 on Build GRCh38
994565 c.1231T>G p.F411V Chr.19: 3290275-3290275 on Build GRCh38
4430452 c.1219C>A p.L407I Chr.19: 3290263-3290263 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs376116023 Chr.19: 3290041-3290041 on Build GRCh38
rs947341318 Chr.19: 3290201-3290201 on Build GRCh38
rs751392720 Chr.19: 3290202-3290202 on Build GRCh38
rs916802303 Chr.19: 3290058-3290058 on Build GRCh38
rs900363663 Chr.19: 3290169-3290169 on Build GRCh38
rs939484190 Chr.19: 3290045-3290045 on Build GRCh38
rs769544919 Chr.19: 3290180-3290180 on Build GRCh38
rs976865064 Chr.19: 3290126-3290126 on Build GRCh38
rs994374882 Chr.19: 3290060-3290060 on Build GRCh38
rs756017768 Chr.19: 3290233-3290233 on Build GRCh38
rs952999914 Chr.19: 3290093-3290093 on Build GRCh38
rs1018801495 Chr.19: 3290166-3290166 on Build GRCh38
rs983142784 Chr.19: 3290253-3290256 on Build GRCh38
rs913110279 Chr.19: 3290176-3290176 on Build GRCh38
rs1026276090 Chr.19: 3290080-3290080 on Build GRCh38
rs571819958 Chr.19: 3290174-3290174 on Build GRCh38
rs964107080 Chr.19: 3290125-3290125 on Build GRCh38
rs759499921 Chr.19: 3290222-3290222 on Build GRCh38
rs768079396 Chr.19: 3290175-3290175 on Build GRCh38
rs895520497 Chr.19: 3290055-3290055 on Build GRCh38
rs891217027 Chr.19: 3290156-3290156 on Build GRCh38
rs1044005229 Chr.19: 3290104-3290104 on Build GRCh38
rs1052913371 Chr.19: 3290142-3290142 on Build GRCh38
rs988163975 Chr.19: 3290173-3290173 on Build GRCh38
rs767736193 Chr.19: 3290074-3290074 on Build GRCh38
rs567542456 Chr.19: 3290161-3290161 on Build GRCh38
rs556922940 Chr.19: 3290113-3290113 on Build GRCh38
rs536850487 Chr.19: 3290103-3290103 on Build GRCh38
rs762324500 Chr.19: 3290195-3290195 on Build GRCh38
rs180871810 Chr.19: 3290170-3290170 on Build GRCh38
rs756376770 Chr.19: 3290225-3290225 on Build GRCh38
rs922592713 Chr.19: 3290135-3290135 on Build GRCh38
rs536185194 Chr.19: 3290169-3290169 on Build GRCh38
rs772900280 Chr.19: 3290194-3290194 on Build GRCh38
rs113248923 Chr.19: 3290096-3290096 on Build GRCh38
rs759596948 Chr.19: 3290165-3290165 on Build GRCh38
rs984667338 Chr.19: 3290100-3290100 on Build GRCh38
rs1008709447 Chr.19: 3290159-3290159 on Build GRCh38

View Full Product Details