Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 51028464-51028693 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COBL
Entrez Gene ID
23242
Gene Name
cordon-bleu WH2 repeat protein
Gene Aliases
-
UniGene
Hs.99141
Gene Chromosome Location
Chr.7: 51016212-51316818 on Build GRCh38
Transcripts
NM_001287436.1 NM_015198.3 XM_005271750.1 XM_005271751.1 XM_005271756.1 XM_011515234.1 XM_011515235.1 XM_011515236.1 XM_011515237.1 XM_011515238.1 XM_011515239.1 XM_011515240.1 XM_011515241.1 XM_017011898.1 XM_017011899.1 XM_017011900.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
160104 c.2513C>A p.T838N Chr.7: 51028583-51028583 on Build GRCh38
3881673 c.2495G>A p.G832E Chr.7: 51028601-51028601 on Build GRCh38
5062519 c.2625C>T p.R875R Chr.7: 51028471-51028471 on Build GRCh38
4621716 c.2691G>A p.S897S Chr.7: 51028651-51028651 on Build GRCh38
7180252 c.2678C>T p.P893L Chr.7: 51028664-51028664 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769314593 Chr.7: 51028616-51028616 on Build GRCh38
rs762847170 Chr.7: 51028484-51028484 on Build GRCh38
rs751828135 Chr.7: 51028489-51028489 on Build GRCh38
rs749598549 Chr.7: 51028508-51028508 on Build GRCh38
rs773182408 Chr.7: 51028557-51028557 on Build GRCh38
rs552401674 Chr.7: 51028653-51028653 on Build GRCh38
rs778468088 Chr.7: 51028507-51028507 on Build GRCh38
rs762604321 Chr.7: 51028558-51028558 on Build GRCh38
rs752797513 Chr.7: 51028590-51028590 on Build GRCh38
rs138072195 Chr.7: 51028543-51028543 on Build GRCh38
rs754403191 Chr.7: 51028598-51028598 on Build GRCh38
rs752375985 Chr.7: 51028663-51028663 on Build GRCh38
rs760891581 Chr.7: 51028594-51028594 on Build GRCh38
rs532457394 Chr.7: 51028652-51028652 on Build GRCh38
rs750770329 Chr.7: 51028656-51028656 on Build GRCh38
rs946144443 Chr.7: 51028665-51028665 on Build GRCh38
rs780862999 Chr.7: 51028612-51028612 on Build GRCh38
rs779535304 Chr.7: 51028516-51028516 on Build GRCh38
rs759312340 Chr.7: 51028635-51028635 on Build GRCh38
rs749249733 Chr.7: 51028620-51028620 on Build GRCh38
rs377072681 Chr.7: 51028494-51028494 on Build GRCh38
rs905996299 Chr.7: 51028573-51028573 on Build GRCh38
rs766487026 Chr.7: 51028488-51028488 on Build GRCh38
rs780773513 Chr.7: 51028659-51028659 on Build GRCh38
rs777347687 Chr.7: 51028667-51028667 on Build GRCh38
rs143849688 Chr.7: 51028648-51028648 on Build GRCh38
rs746679834 Chr.7: 51028521-51028521 on Build GRCh38
rs146375254 Chr.7: 51028664-51028664 on Build GRCh38
rs150370389 Chr.7: 51028501-51028501 on Build GRCh38
rs141630570 Chr.7: 51028658-51028658 on Build GRCh38
rs770787924 Chr.7: 51028561-51028561 on Build GRCh38
rs780667032 Chr.7: 51028540-51028540 on Build GRCh38
rs563438174 Chr.7: 51028651-51028651 on Build GRCh38
rs770570854 Chr.7: 51028628-51028628 on Build GRCh38
rs750811837 Chr.7: 51028606-51028606 on Build GRCh38
rs1024494153 Chr.7: 51028570-51028570 on Build GRCh38
rs138592316 Chr.7: 51028649-51028649 on Build GRCh38
rs529941693 Chr.7: 51028531-51028531 on Build GRCh38
rs149461023 Chr.7: 51028614-51028614 on Build GRCh38
rs754621952 Chr.7: 51028611-51028611 on Build GRCh38
rs1027835971 Chr.7: 51028577-51028577 on Build GRCh38
rs779351564 Chr.7: 51028605-51028605 on Build GRCh38
rs759814093 Chr.7: 51028579-51028579 on Build GRCh38
rs765730672 Chr.7: 51028654-51028654 on Build GRCh38
rs370189979 Chr.7: 51028566-51028566 on Build GRCh38
rs756671678 Chr.7: 51028505-51028505 on Build GRCh38
rs370819886 Chr.7: 51028630-51028630 on Build GRCh38
rs764650492 Chr.7: 51028597-51028597 on Build GRCh38
rs757498778 Chr.7: 51028602-51028602 on Build GRCh38
rs376448766 Chr.7: 51028619-51028619 on Build GRCh38
rs373421935 Chr.7: 51028585-51028585 on Build GRCh38
rs757665351 Chr.7: 51028514-51028514 on Build GRCh38

View Full Product Details