Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 92480397-92480630 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GFI1
Entrez Gene ID
2672
Gene Name
growth factor independent 1 transcriptional repressor
Gene Aliases
GFI-1, GFI1A, SCN2, ZNF163
UniGene
Hs.73172
Gene Chromosome Location
Chr.1: 92473043-92486876 on Build GRCh38
Transcripts
NM_001127215.1 NM_001127216.1 NM_005263.3 XM_005270749.3 XM_011541245.2 XM_011541246.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5470958 c.841A>G p.T281A Chr.1: 92480431-92480431 on Build GRCh38
1208114 c.869G>A p.G290D Chr.1: 92480403-92480403 on Build GRCh38
1602792 c.823C>A p.R275S Chr.1: 92480449-92480449 on Build GRCh38
5675046 c.808C>A p.L270I Chr.1: 92480464-92480464 on Build GRCh38
3806114 c.860A>C p.E287A Chr.1: 92480412-92480412 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767464450 Chr.1: 92480418-92480418 on Build GRCh38
rs372974369 Chr.1: 92480453-92480453 on Build GRCh38
rs573402585 Chr.1: 92480519-92480519 on Build GRCh38
rs982883265 Chr.1: 92480560-92480560 on Build GRCh38
rs756137710 Chr.1: 92480589-92480589 on Build GRCh38
rs555127187 Chr.1: 92480497-92480497 on Build GRCh38
rs566318572 Chr.1: 92480479-92480479 on Build GRCh38
rs954470735 Chr.1: 92480521-92480521 on Build GRCh38
rs777176619 Chr.1: 92480525-92480525 on Build GRCh38
rs753590082 Chr.1: 92480512-92480512 on Build GRCh38
rs765190282 Chr.1: 92480561-92480561 on Build GRCh38
rs774671739 Chr.1: 92480472-92480472 on Build GRCh38
rs542012583 Chr.1: 92480530-92480530 on Build GRCh38
rs1025556601 Chr.1: 92480527-92480528 on Build GRCh38
rs750502057 Chr.1: 92480582-92480582 on Build GRCh38
rs936881326 Chr.1: 92480565-92480565 on Build GRCh38
rs772613865 Chr.1: 92480481-92480481 on Build GRCh38
rs914173529 Chr.1: 92480594-92480594 on Build GRCh38
rs557172855 Chr.1: 92480564-92480564 on Build GRCh38
rs961671559 Chr.1: 92480499-92480499 on Build GRCh38
rs766149113 Chr.1: 92480504-92480504 on Build GRCh38
rs958605699 Chr.1: 92480434-92480434 on Build GRCh38
rs370294881 Chr.1: 92480590-92480590 on Build GRCh38
rs971725414 Chr.1: 92480502-92480502 on Build GRCh38
rs376949976 Chr.1: 92480468-92480468 on Build GRCh38
rs758280128 Chr.1: 92480562-92480562 on Build GRCh38
rs759229534 Chr.1: 92480514-92480514 on Build GRCh38
rs114487177 Chr.1: 92480480-92480480 on Build GRCh38

View Full Product Details