Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 45366156-45366429 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GATM
Entrez Gene ID
2628
Gene Name
glycine amidinotransferase
Gene Aliases
AGAT, AT, CCDS3
UniGene
Hs.560354
Gene Chromosome Location
Chr.15: 45361124-45402317 on Build GRCh38
Transcripts
NM_001321015.1 NM_001482.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6669649 c.781G>T p.A261S Chr.15: 45366403-45366403 on Build GRCh38
433966 c.775A>T p.I259F Chr.15: 45366409-45366409 on Build GRCh38
433964 c.856G>T p.A286S Chr.15: 45366168-45366168 on Build GRCh38
962247 c.844C>T p.R282C Chr.15: 45366180-45366180 on Build GRCh38
5764016 c.813+6C>T p.? Chr.15: 45366365-45366365 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs3210026 Chr.15: 45366190-45366190 on Build GRCh38
rs150282769 Chr.15: 45366325-45366325 on Build GRCh38
rs777561689 Chr.15: 45366234-45366234 on Build GRCh38
rs371507418 Chr.15: 45366383-45366383 on Build GRCh38
rs112157082 Chr.15: 45366331-45366331 on Build GRCh38
rs746864923 Chr.15: 45366330-45366330 on Build GRCh38
rs778691153 Chr.15: 45366257-45366257 on Build GRCh38
rs766583032 Chr.15: 45366180-45366180 on Build GRCh38
rs759872969 Chr.15: 45366223-45366223 on Build GRCh38
rs1032883981 Chr.15: 45366358-45366358 on Build GRCh38
rs781058415 Chr.15: 45366359-45366359 on Build GRCh38
rs775933965 Chr.15: 45366406-45366406 on Build GRCh38
rs553676897 Chr.15: 45366363-45366363 on Build GRCh38
rs768644583 Chr.15: 45366355-45366355 on Build GRCh38
rs201984342 Chr.15: 45366243-45366243 on Build GRCh38
rs1044125549 Chr.15: 45366268-45366268 on Build GRCh38
rs749360708 Chr.15: 45366361-45366361 on Build GRCh38
rs1000343408 Chr.15: 45366344-45366344 on Build GRCh38
rs753393267 Chr.15: 45366236-45366236 on Build GRCh38
rs371320329 Chr.15: 45366246-45366246 on Build GRCh38
rs112583776 Chr.15: 45366183-45366183 on Build GRCh38
rs371447931 Chr.15: 45366179-45366179 on Build GRCh38
rs3210027 Chr.15: 45366187-45366187 on Build GRCh38
rs771050735 Chr.15: 45366365-45366365 on Build GRCh38
rs991526064 Chr.15: 45366292-45366292 on Build GRCh38
rs371808484 Chr.15: 45366233-45366233 on Build GRCh38
rs917013788 Chr.15: 45366297-45366297 on Build GRCh38
rs374024706 Chr.15: 45366237-45366237 on Build GRCh38
rs958399901 Chr.15: 45366263-45366263 on Build GRCh38
rs3210025 Chr.15: 45366196-45366196 on Build GRCh38
rs774614737 Chr.15: 45366219-45366219 on Build GRCh38
rs750148049 Chr.15: 45366260-45366260 on Build GRCh38
rs368314780 Chr.15: 45366227-45366227 on Build GRCh38
rs759787223 Chr.15: 45366375-45366375 on Build GRCh38
rs774528761 Chr.15: 45366372-45366372 on Build GRCh38

View Full Product Details