Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 128875855-128876116 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLT1D1
Entrez Gene ID
144423
Gene Name
glycosyltransferase 1 domain containing 1
Gene Aliases
-
UniGene
Hs.655668
Gene Chromosome Location
Chr.12: 128853323-128984964 on Build GRCh38
Transcripts
NM_144669.2 NR_133646.1 XM_005253553.4 XM_011537956.2 XM_011537957.2 XM_017018855.1 XM_017018856.1 XM_017018857.1 XM_017018858.1 XM_017018859.1 XM_017018860.1 XM_017018861.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4776869 c.124G>C p.E42Q Chr.12: 128875969-128875969 on Build GRCh38
6071552 c.124G>T p.E42* Chr.12: 128875969-128875969 on Build GRCh38
7369579 c.175G>T p.A59S Chr.12: 128876020-128876020 on Build GRCh38
4806063 c.99C>T p.C33C Chr.12: 128875944-128875944 on Build GRCh38
692344 c.175G>T p.A59S Chr.12: 128876020-128876020 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs867614295 Chr.12: 128875887-128875887 on Build GRCh38
rs201930455 Chr.12: 128876014-128876014 on Build GRCh38
rs146263464 Chr.12: 128875945-128875945 on Build GRCh38
rs761881814 Chr.12: 128875950-128875950 on Build GRCh38
rs767207876 Chr.12: 128875952-128875952 on Build GRCh38
rs188735757 Chr.12: 128875975-128875975 on Build GRCh38
rs200983869 Chr.12: 128875938-128875938 on Build GRCh38
rs139435767 Chr.12: 128875998-128875998 on Build GRCh38
rs780315506 Chr.12: 128876041-128876041 on Build GRCh38
rs879128062 Chr.12: 128875942-128875942 on Build GRCh38
rs766553443 Chr.12: 128875968-128875968 on Build GRCh38
rs758744624 Chr.12: 128875904-128875904 on Build GRCh38
rs778240811 Chr.12: 128875910-128875910 on Build GRCh38
rs748997146 Chr.12: 128875995-128875995 on Build GRCh38
rs773025838 Chr.12: 128876027-128876027 on Build GRCh38
rs144231014 Chr.12: 128876013-128876013 on Build GRCh38
rs200372553 Chr.12: 128875931-128875931 on Build GRCh38
rs865787701 Chr.12: 128875930-128875930 on Build GRCh38
rs747342544 Chr.12: 128875984-128875984 on Build GRCh38
rs774407847 Chr.12: 128875946-128875946 on Build GRCh38
rs776815903 Chr.12: 128876042-128876042 on Build GRCh38
rs747883586 Chr.12: 128876046-128876046 on Build GRCh38
rs201069032 Chr.12: 128876053-128876053 on Build GRCh38
rs1017794786 Chr.12: 128876029-128876029 on Build GRCh38
rs779706838 Chr.12: 128875989-128875989 on Build GRCh38
rs113674717 Chr.12: 128875939-128875939 on Build GRCh38
rs768249750 Chr.12: 128875996-128875996 on Build GRCh38
rs148754002 Chr.12: 128876051-128876051 on Build GRCh38
rs758971476 Chr.12: 128875980-128875980 on Build GRCh38
rs752833678 Chr.12: 128876071-128876071 on Build GRCh38
rs482401 Chr.12: 128875953-128875953 on Build GRCh38
rs748679664 Chr.12: 128875890-128875890 on Build GRCh38
rs760397695 Chr.12: 128875965-128875965 on Build GRCh38
rs73438369 Chr.12: 128875973-128875973 on Build GRCh38
rs947404760 Chr.12: 128875969-128875969 on Build GRCh38
rs777988195 Chr.12: 128875981-128875981 on Build GRCh38
rs765354223 Chr.12: 128875976-128875976 on Build GRCh38
rs760480575 Chr.12: 128876039-128876039 on Build GRCh38
rs757568901 Chr.12: 128875987-128875987 on Build GRCh38
rs745503932 Chr.12: 128875935-128875935 on Build GRCh38
rs770656717 Chr.12: 128876040-128876040 on Build GRCh38
rs370646672 Chr.12: 128875944-128875944 on Build GRCh38
rs763035076 Chr.12: 128876079-128876079 on Build GRCh38

View Full Product Details