Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 53480621-53480870 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FST
Entrez Gene ID
10468
Gene Name
follistatin
Gene Aliases
FS
UniGene
Hs.9914
Gene Chromosome Location
Chr.5: 53480338-53487134 on Build GRCh38
Transcripts
NM_006350.3 NM_013409.2 XM_005248400.4 XM_005248401.4 XM_005248402.4 XM_005248403.4 XM_017008954.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3429508 c.52C>T p.L18F Chr.5: 53480843-53480843 on Build GRCh38
3429509 c.52C>T p.L18F Chr.5: 53480843-53480843 on Build GRCh38
4439601 c.7C>T p.R3C Chr.5: 53480798-53480798 on Build GRCh38
5490833 c.14G>T p.R5M Chr.5: 53480805-53480805 on Build GRCh38
4439602 c.7C>T p.R3C Chr.5: 53480798-53480798 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200358632 Chr.5: 53480831-53480831 on Build GRCh38
rs768837889 Chr.5: 53480771-53480771 on Build GRCh38
rs994863763 Chr.5: 53480726-53480726 on Build GRCh38
rs377375490 Chr.5: 53480800-53480800 on Build GRCh38
rs891618340 Chr.5: 53480649-53480649 on Build GRCh38
rs749580765 Chr.5: 53480763-53480763 on Build GRCh38
rs550147513 Chr.5: 53480737-53480737 on Build GRCh38
rs901924329 Chr.5: 53480732-53480732 on Build GRCh38
rs780422074 Chr.5: 53480752-53480752 on Build GRCh38
rs150238035 Chr.5: 53480843-53480843 on Build GRCh38
rs1052897490 Chr.5: 53480642-53480642 on Build GRCh38
rs772413605 Chr.5: 53480802-53480802 on Build GRCh38
rs908580952 Chr.5: 53480667-53480668 on Build GRCh38
rs889729550 Chr.5: 53480692-53480692 on Build GRCh38
rs551392710 Chr.5: 53480836-53480836 on Build GRCh38
rs769718104 Chr.5: 53480747-53480747 on Build GRCh38
rs1007935039 Chr.5: 53480713-53480713 on Build GRCh38
rs1017612609 Chr.5: 53480715-53480715 on Build GRCh38
rs542798811 Chr.5: 53480705-53480706 on Build GRCh38
rs950856670 Chr.5: 53480744-53480744 on Build GRCh38
rs867149376 Chr.5: 53480730-53480730 on Build GRCh38
rs568804432 Chr.5: 53480798-53480798 on Build GRCh38
rs1051425771 Chr.5: 53480673-53480673 on Build GRCh38
rs963381807 Chr.5: 53480720-53480720 on Build GRCh38
rs370746064 Chr.5: 53480842-53480842 on Build GRCh38
rs773779294 Chr.5: 53480808-53480808 on Build GRCh38
rs763409214 Chr.5: 53480820-53480820 on Build GRCh38

View Full Product Details