Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 99383599-99383872 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COQ3
Entrez Gene ID
51805
Gene Name
coenzyme Q3, methyltransferase
Gene Aliases
DHHBMT, DHHBMTASE, UG0215E05, bA9819.1
UniGene
Hs.713623
Gene Chromosome Location
Chr.6: 99369401-99394223 on Build GRCh38
Transcripts
NM_017421.3 XM_005267018.4 XM_006715500.3 XM_011535898.2 XM_011535899.1 XM_017010933.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4841313 c.107-1G>A p.? Chr.6: 99383825-99383825 on Build GRCh38
6634141 c.119A>G p.N40S Chr.6: 99383812-99383812 on Build GRCh38
5675815 c.232A>T p.R78W Chr.6: 99383699-99383699 on Build GRCh38
3080022 c.196A>G p.T66A Chr.6: 99383735-99383735 on Build GRCh38
3080021 c.198G>T p.T66T Chr.6: 99383733-99383733 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs539593135 Chr.6: 99383658-99383658 on Build GRCh38
rs760130868 Chr.6: 99383707-99383708 on Build GRCh38
rs925003736 Chr.6: 99383670-99383670 on Build GRCh38
rs776961483 Chr.6: 99383704-99383704 on Build GRCh38
rs149717575 Chr.6: 99383734-99383734 on Build GRCh38
rs779971687 Chr.6: 99383672-99383672 on Build GRCh38
rs777491812 Chr.6: 99383819-99383819 on Build GRCh38
rs753082700 Chr.6: 99383764-99383764 on Build GRCh38
rs767290578 Chr.6: 99383828-99383828 on Build GRCh38
rs200092962 Chr.6: 99383752-99383752 on Build GRCh38
rs770594278 Chr.6: 99383835-99383835 on Build GRCh38
rs755406663 Chr.6: 99383662-99383662 on Build GRCh38
rs757344308 Chr.6: 99383778-99383778 on Build GRCh38
rs772753636 Chr.6: 99383651-99383651 on Build GRCh38
rs776055716 Chr.6: 99383729-99383730 on Build GRCh38
rs760080328 Chr.6: 99383754-99383754 on Build GRCh38
rs562096103 Chr.6: 99383709-99383709 on Build GRCh38
rs751530962 Chr.6: 99383801-99383801 on Build GRCh38
rs758397950 Chr.6: 99383668-99383668 on Build GRCh38
rs763609902 Chr.6: 99383655-99383655 on Build GRCh38
rs1024648718 Chr.6: 99383716-99383716 on Build GRCh38
rs970040638 Chr.6: 99383722-99383722 on Build GRCh38
rs780903321 Chr.6: 99383810-99383810 on Build GRCh38
rs770504283 Chr.6: 99383748-99383748 on Build GRCh38
rs747798585 Chr.6: 99383733-99383733 on Build GRCh38
rs1001913144 Chr.6: 99383797-99383797 on Build GRCh38
rs769405773 Chr.6: 99383814-99383814 on Build GRCh38
rs765819302 Chr.6: 99383723-99383729 on Build GRCh38
rs201283634 Chr.6: 99383827-99383828 on Build GRCh38
rs369360291 Chr.6: 99383770-99383770 on Build GRCh38
rs371786381 Chr.6: 99383667-99383667 on Build GRCh38
rs547653484 Chr.6: 99383821-99383821 on Build GRCh38
rs773968193 Chr.6: 99383836-99383836 on Build GRCh38
rs201434886 Chr.6: 99383838-99383838 on Build GRCh38
rs531347620 Chr.6: 99383664-99383664 on Build GRCh38
rs760864787 Chr.6: 99383767-99383767 on Build GRCh38
rs139543322 Chr.6: 99383713-99383713 on Build GRCh38
rs754883278 Chr.6: 99383807-99383807 on Build GRCh38
rs533241969 Chr.6: 99383737-99383737 on Build GRCh38
rs200652928 Chr.6: 99383659-99383659 on Build GRCh38
rs905138900 Chr.6: 99383790-99383790 on Build GRCh38
rs753994014 Chr.6: 99383773-99383773 on Build GRCh38
rs767810694 Chr.6: 99383755-99383755 on Build GRCh38
rs762552282 Chr.6: 99383746-99383746 on Build GRCh38
rs752892027 Chr.6: 99383665-99383665 on Build GRCh38
rs778918272 Chr.6: 99383781-99383781 on Build GRCh38
rs35446098 Chr.6: 99383666-99383666 on Build GRCh38
rs781003608 Chr.6: 99383721-99383721 on Build GRCh38
rs1052178319 Chr.6: 99383711-99383711 on Build GRCh38
rs772662627 Chr.6: 99383744-99383744 on Build GRCh38

View Full Product Details