Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 48670566-48670839 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTIF
Entrez Gene ID
9811
Gene Name
cap binding complex dependent translation initiation factor
Gene Aliases
Gm672, KIAA0427
UniGene
Hs.145230
Gene Chromosome Location
Chr.18: 48539056-48863217 on Build GRCh38
Transcripts
NM_001142397.1 NM_014772.2 XM_005258392.4 XM_006722583.3 XM_006722586.3 XM_006722587.3 XM_006722588.3 XM_006722589.3 XM_011526278.2 XM_011526279.2 XM_017026100.1 XM_017026101.1 XM_017026102.1 NR_039897.2

Gene Symbol
MIR4743
Entrez Gene ID
100616366
Gene Name
microRNA 4743
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.18: 48670600-48670668 on Build GRCh38
Transcripts
NM_001142397.1 NM_014772.2 XM_005258392.4 XM_006722583.3 XM_006722586.3 XM_006722587.3 XM_006722588.3 XM_006722589.3 XM_011526278.2 XM_011526279.2 XM_017026100.1 XM_017026101.1 XM_017026102.1 NR_039897.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6637461 c.434G>A p.C145Y Chr.18: 48670671-48670671 on Build GRCh38
6637462 c.290G>A p.C97Y Chr.18: 48670671-48670671 on Build GRCh38
399781 c.334G>C p.D112H Chr.18: 48670715-48670715 on Build GRCh38
359213 c.288-1G>T p.? Chr.18: 48670668-48670668 on Build GRCh38
5542799 c.497C>T p.P166L Chr.18: 48670734-48670734 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766164347 Chr.18: 48670690-48670690 on Build GRCh38
rs998472760 Chr.18: 48670794-48670794 on Build GRCh38
rs760450593 Chr.18: 48670680-48670680 on Build GRCh38
rs765094831 Chr.18: 48670699-48670699 on Build GRCh38
rs35389018 Chr.18: 48670598-48670599 on Build GRCh38
rs749628840 Chr.18: 48670740-48670740 on Build GRCh38
rs747715114 Chr.18: 48670760-48670760 on Build GRCh38
rs7235219 Chr.18: 48670627-48670627 on Build GRCh38
rs766282351 Chr.18: 48670598-48670598 on Build GRCh38
rs78396863 Chr.18: 48670600-48670600 on Build GRCh38
rs780490742 Chr.18: 48670727-48670727 on Build GRCh38
rs951675883 Chr.18: 48670809-48670809 on Build GRCh38
rs774027223 Chr.18: 48670646-48670646 on Build GRCh38
rs770453940 Chr.18: 48670783-48670783 on Build GRCh38
rs780467818 Chr.18: 48670631-48670631 on Build GRCh38
rs376137526 Chr.18: 48670619-48670619 on Build GRCh38
rs201846069 Chr.18: 48670780-48670780 on Build GRCh38
rs769236249 Chr.18: 48670644-48670644 on Build GRCh38
rs757319746 Chr.18: 48670714-48670714 on Build GRCh38
rs934522725 Chr.18: 48670696-48670696 on Build GRCh38
rs756486451 Chr.18: 48670721-48670721 on Build GRCh38
rs368580312 Chr.18: 48670623-48670623 on Build GRCh38
rs781337267 Chr.18: 48670718-48670718 on Build GRCh38
rs376167919 Chr.18: 48670757-48670757 on Build GRCh38
rs747349933 Chr.18: 48670782-48670783 on Build GRCh38
rs1019047741 Chr.18: 48670806-48670806 on Build GRCh38
rs750187970 Chr.18: 48670591-48670591 on Build GRCh38
rs771863567 Chr.18: 48670651-48670651 on Build GRCh38
rs895731698 Chr.18: 48670648-48670648 on Build GRCh38
rs776491838 Chr.18: 48670784-48670784 on Build GRCh38
rs987150428 Chr.18: 48670801-48670801 on Build GRCh38
rs1018308655 Chr.18: 48670759-48670759 on Build GRCh38
rs772903719 Chr.18: 48670652-48670652 on Build GRCh38
rs751601028 Chr.18: 48670615-48670615 on Build GRCh38
rs1045621995 Chr.18: 48670672-48670672 on Build GRCh38
rs994673887 Chr.18: 48670758-48670758 on Build GRCh38
rs200054717 Chr.18: 48670697-48670697 on Build GRCh38
rs771603294 Chr.18: 48670768-48670768 on Build GRCh38
rs769289443 Chr.18: 48670748-48670748 on Build GRCh38
rs767239251 Chr.18: 48670587-48670587 on Build GRCh38
rs375891314 Chr.18: 48670620-48670620 on Build GRCh38
rs535781992 Chr.18: 48670781-48670781 on Build GRCh38
rs375588934 Chr.18: 48670643-48670643 on Build GRCh38
rs147083066 Chr.18: 48670720-48670720 on Build GRCh38
rs903051268 Chr.18: 48670673-48670673 on Build GRCh38
rs368684997 Chr.18: 48670710-48670710 on Build GRCh38
rs75980206 Chr.18: 48670796-48670796 on Build GRCh38
rs768333677 Chr.18: 48670711-48670711 on Build GRCh38
rs777856459 Chr.18: 48670607-48670607 on Build GRCh38
rs755876988 Chr.18: 48670597-48670597 on Build GRCh38
rs141766192 Chr.18: 48670606-48670606 on Build GRCh38
rs555379847 Chr.18: 48670785-48670785 on Build GRCh38

View Full Product Details