Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 45713248-45713521 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO46
Entrez Gene ID
23403
Gene Name
F-box protein 46
Gene Aliases
20D7-FC4, FBXO34L, Fbx46
UniGene
Hs.128702
Gene Chromosome Location
Chr.19: 45710629-45730893 on Build GRCh38
Transcripts
NM_001080469.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3680972 c.112C>T p.P38S Chr.19: 45713384-45713384 on Build GRCh38
5689165 c.81G>T p.P27P Chr.19: 45713415-45713415 on Build GRCh38
6368580 c.133G>A p.E45K Chr.19: 45713363-45713363 on Build GRCh38
712316 c.7C>T p.R3C Chr.19: 45713489-45713489 on Build GRCh38
998275 c.62C>A p.S21* Chr.19: 45713434-45713434 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760276284 Chr.19: 45713289-45713289 on Build GRCh38
rs745769278 Chr.19: 45713428-45713428 on Build GRCh38
rs760199564 Chr.19: 45713379-45713379 on Build GRCh38
rs1000652101 Chr.19: 45713319-45713319 on Build GRCh38
rs374630294 Chr.19: 45713415-45713415 on Build GRCh38
rs775242932 Chr.19: 45713371-45713371 on Build GRCh38
rs370501759 Chr.19: 45713369-45713369 on Build GRCh38
rs781072997 Chr.19: 45713363-45713363 on Build GRCh38
rs367697053 Chr.19: 45713424-45713424 on Build GRCh38
rs758865512 Chr.19: 45713413-45713413 on Build GRCh38
rs761458955 Chr.19: 45713489-45713489 on Build GRCh38
rs763476298 Chr.19: 45713299-45713299 on Build GRCh38
rs775092183 Chr.19: 45713452-45713452 on Build GRCh38
rs779263838 Chr.19: 45713282-45713290 on Build GRCh38
rs201503195 Chr.19: 45713283-45713283 on Build GRCh38
rs776410945 Chr.19: 45713467-45713467 on Build GRCh38
rs753334672 Chr.19: 45713385-45713385 on Build GRCh38
rs768802197 Chr.19: 45713361-45713361 on Build GRCh38
rs865844887 Chr.19: 45713362-45713362 on Build GRCh38
rs759318995 Chr.19: 45713277-45713277 on Build GRCh38
rs761758759 Chr.19: 45713277-45713279 on Build GRCh38
rs372194078 Chr.19: 45713407-45713407 on Build GRCh38
rs549110315 Chr.19: 45713423-45713423 on Build GRCh38
rs200615699 Chr.19: 45713331-45713331 on Build GRCh38
rs759372079 Chr.19: 45713370-45713370 on Build GRCh38
rs751819786 Chr.19: 45713416-45713416 on Build GRCh38
rs868700968 Chr.19: 45713356-45713356 on Build GRCh38
rs376603564 Chr.19: 45713345-45713345 on Build GRCh38
rs11537711 Chr.19: 45713384-45713384 on Build GRCh38
rs755087299 Chr.19: 45713419-45713419 on Build GRCh38
rs771926520 Chr.19: 45713444-45713444 on Build GRCh38
rs758822939 Chr.19: 45713326-45713326 on Build GRCh38
rs559016267 Chr.19: 45713482-45713482 on Build GRCh38
rs368353647 Chr.19: 45713406-45713406 on Build GRCh38
rs760382297 Chr.19: 45713457-45713457 on Build GRCh38
rs779427638 Chr.19: 45713321-45713321 on Build GRCh38
rs768261197 Chr.19: 45713463-45713463 on Build GRCh38
rs544663802 Chr.19: 45713322-45713322 on Build GRCh38
rs374651411 Chr.19: 45713310-45713310 on Build GRCh38
rs576590274 Chr.19: 45713408-45713408 on Build GRCh38
rs749226167 Chr.19: 45713364-45713364 on Build GRCh38
rs965176155 Chr.19: 45713499-45713499 on Build GRCh38
rs748282680 Chr.19: 45713420-45713420 on Build GRCh38

View Full Product Details