Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 164699338-164699607 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COBLL1
Entrez Gene ID
22837
Gene Name
cordon-bleu WH2 repeat protein like 1
Gene Aliases
COBLR1
UniGene
Hs.470457
Gene Chromosome Location
Chr.2: 164680185-164842168 on Build GRCh38
Transcripts
NM_001278458.1 NM_001278460.1 NM_001278461.1 NM_014900.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
222714 c.1600G>A p.D534N Chr.2: 164699477-164699477 on Build GRCh38
6632147 c.1582_1590delCCACACAGT p.P528_S530delPHS Chr.2: 164699487-164699495 on Build GRCh38
6314600 c.1587C>A p.H529Q Chr.2: 164699490-164699490 on Build GRCh38
6232174 c.1585C>A p.H529N Chr.2: 164699492-164699492 on Build GRCh38
1248702 c.1637G>C p.R546T Chr.2: 164699440-164699440 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772871866 Chr.2: 164699568-164699569 on Build GRCh38
rs113688595 Chr.2: 164699564-164699564 on Build GRCh38
rs372370676 Chr.2: 164699567-164699568 on Build GRCh38
rs767713516 Chr.2: 164699400-164699400 on Build GRCh38
rs751878467 Chr.2: 164699382-164699382 on Build GRCh38
rs759634167 Chr.2: 164699398-164699398 on Build GRCh38
rs779623431 Chr.2: 164699364-164699364 on Build GRCh38
rs532465805 Chr.2: 164699459-164699459 on Build GRCh38
rs755282988 Chr.2: 164699509-164699511 on Build GRCh38
rs781420690 Chr.2: 164699518-164699522 on Build GRCh38
rs917274651 Chr.2: 164699566-164699566 on Build GRCh38
rs993208961 Chr.2: 164699552-164699552 on Build GRCh38
rs757755954 Chr.2: 164699428-164699428 on Build GRCh38
rs746589050 Chr.2: 164699468-164699468 on Build GRCh38
rs754705134 Chr.2: 164699486-164699486 on Build GRCh38
rs768293481 Chr.2: 164699370-164699370 on Build GRCh38
rs775536186 Chr.2: 164699406-164699406 on Build GRCh38
rs1015406092 Chr.2: 164699492-164699492 on Build GRCh38
rs760981303 Chr.2: 164699421-164699421 on Build GRCh38
rs985335770 Chr.2: 164699566-164699567 on Build GRCh38
rs760778696 Chr.2: 164699550-164699550 on Build GRCh38
rs201899325 Chr.2: 164699508-164699508 on Build GRCh38
rs769726735 Chr.2: 164699513-164699513 on Build GRCh38
rs776313992 Chr.2: 164699426-164699426 on Build GRCh38
rs773219795 Chr.2: 164699525-164699525 on Build GRCh38
rs911083427 Chr.2: 164699570-164699570 on Build GRCh38
rs377692408 Chr.2: 164699524-164699524 on Build GRCh38
rs1044995181 Chr.2: 164699574-164699574 on Build GRCh38
rs148013206 Chr.2: 164699444-164699444 on Build GRCh38
rs776503778 Chr.2: 164699379-164699379 on Build GRCh38
rs748407404 Chr.2: 164699522-164699523 on Build GRCh38
rs761629947 Chr.2: 164699383-164699383 on Build GRCh38
rs756304905 Chr.2: 164699543-164699544 on Build GRCh38
rs145983847 Chr.2: 164699510-164699510 on Build GRCh38
rs111634619 Chr.2: 164699386-164699386 on Build GRCh38
rs376370995 Chr.2: 164699366-164699366 on Build GRCh38
rs139984226 Chr.2: 164699553-164699554 on Build GRCh38
rs765790478 Chr.2: 164699431-164699431 on Build GRCh38
rs770645205 Chr.2: 164699387-164699387 on Build GRCh38
rs763045374 Chr.2: 164699538-164699538 on Build GRCh38
rs764343353 Chr.2: 164699423-164699423 on Build GRCh38
rs920561945 Chr.2: 164699548-164699548 on Build GRCh38
rs139867643 Chr.2: 164699511-164699511 on Build GRCh38
rs550952018 Chr.2: 164699572-164699572 on Build GRCh38
rs371327024 Chr.2: 164699427-164699427 on Build GRCh38
rs568702283 Chr.2: 164699539-164699539 on Build GRCh38
rs188506595 Chr.2: 164699537-164699537 on Build GRCh38
rs750939544 Chr.2: 164699443-164699443 on Build GRCh38
rs766908844 Chr.2: 164699375-164699387 on Build GRCh38

View Full Product Details