Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67831717-67831984 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CENPT
Entrez Gene ID
80152
Gene Name
centromere protein T
Gene Aliases
C16orf56, CENP-T
UniGene
Hs.288382
Gene Chromosome Location
Chr.16: 67828157-67847458 on Build GRCh38
Transcripts
NM_025082.3 XM_006721277.2 XM_011523344.1 XM_017023712.1 XM_017023713.1 XM_017023714.1 XM_017023715.1 XM_017023716.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3795020 c.124C>G p.P42A Chr.16: 67831844-67831844 on Build GRCh38
3511121 c.411C>T p.L137L Chr.16: 67831866-67831866 on Build GRCh38
3511119 c.469C>T p.L157L Chr.16: 67831808-67831808 on Build GRCh38
1679191 c.523G>T p.E175* Chr.16: 67831754-67831754 on Build GRCh38
6328204 c.103G>A p.E35K Chr.16: 67831865-67831865 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs556225780 Chr.16: 67831786-67831786 on Build GRCh38
rs764975014 Chr.16: 67831885-67831885 on Build GRCh38
rs746006421 Chr.16: 67831858-67831858 on Build GRCh38
rs750201233 Chr.16: 67831778-67831778 on Build GRCh38
rs747717633 Chr.16: 67831825-67831825 on Build GRCh38
rs963435382 Chr.16: 67831948-67831950 on Build GRCh38
rs764309153 Chr.16: 67831862-67831862 on Build GRCh38
rs201102788 Chr.16: 67831924-67831924 on Build GRCh38
rs764993047 Chr.16: 67831964-67831964 on Build GRCh38
rs760752826 Chr.16: 67831866-67831866 on Build GRCh38
rs11558535 Chr.16: 67831859-67831859 on Build GRCh38
rs367603713 Chr.16: 67831920-67831920 on Build GRCh38
rs780043405 Chr.16: 67831787-67831787 on Build GRCh38
rs200384622 Chr.16: 67831770-67831770 on Build GRCh38
rs775380845 Chr.16: 67831861-67831861 on Build GRCh38
rs757884730 Chr.16: 67831889-67831889 on Build GRCh38
rs191010035 Chr.16: 67831927-67831927 on Build GRCh38
rs758793161 Chr.16: 67831850-67831850 on Build GRCh38
rs754540431 Chr.16: 67831874-67831874 on Build GRCh38
rs778229845 Chr.16: 67831814-67831814 on Build GRCh38
rs535231412 Chr.16: 67831820-67831820 on Build GRCh38
rs766442339 Chr.16: 67831939-67831939 on Build GRCh38
rs749520828 Chr.16: 67831915-67831915 on Build GRCh38
rs746263011 Chr.16: 67831902-67831902 on Build GRCh38
rs572614065 Chr.16: 67831908-67831908 on Build GRCh38
rs752335935 Chr.16: 67831887-67831887 on Build GRCh38
rs756547928 Chr.16: 67831764-67831770 on Build GRCh38
rs542018475 Chr.16: 67831838-67831838 on Build GRCh38
rs776251728 Chr.16: 67831962-67831962 on Build GRCh38
rs566246287 Chr.16: 67831767-67831767 on Build GRCh38
rs562051979 Chr.16: 67831857-67831857 on Build GRCh38
rs778225207 Chr.16: 67831851-67831851 on Build GRCh38
rs766211190 Chr.16: 67831870-67831870 on Build GRCh38
rs753681583 Chr.16: 67831872-67831872 on Build GRCh38
rs773260273 Chr.16: 67831865-67831865 on Build GRCh38
rs372047486 Chr.16: 67831890-67831890 on Build GRCh38
rs969575353 Chr.16: 67831863-67831863 on Build GRCh38
rs934338833 Chr.16: 67831753-67831753 on Build GRCh38
rs372795270 Chr.16: 67831963-67831963 on Build GRCh38
rs760632199 Chr.16: 67831935-67831935 on Build GRCh38
rs973911359 Chr.16: 67831950-67831950 on Build GRCh38
rs771362580 Chr.16: 67831929-67831929 on Build GRCh38
rs778386288 Chr.16: 67831796-67831797 on Build GRCh38
rs768592676 Chr.16: 67831948-67831948 on Build GRCh38
rs554491310 Chr.16: 67831893-67831893 on Build GRCh38
rs765224013 Chr.16: 67831745-67831745 on Build GRCh38
rs747514427 Chr.16: 67831891-67831891 on Build GRCh38
rs200046488 Chr.16: 67831806-67831806 on Build GRCh38
rs749694330 Chr.16: 67831856-67831857 on Build GRCh38
rs375024438 Chr.16: 67831900-67831900 on Build GRCh38
rs11558536 Chr.16: 67831845-67831845 on Build GRCh38
rs371678424 Chr.16: 67831856-67831856 on Build GRCh38
rs753419395 Chr.16: 67831803-67831803 on Build GRCh38
rs759863422 Chr.16: 67831740-67831740 on Build GRCh38

View Full Product Details