Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 47937277-47937522 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNGA1
Entrez Gene ID
1259
Gene Name
cyclic nucleotide gated channel alpha 1
Gene Aliases
CNCG, CNCG1, CNG-1, CNG1, RCNC1, RCNCa, RCNCalpha, RP49
UniGene
Hs.1323
Gene Chromosome Location
Chr.4: 47935015-48016718 on Build GRCh38
Transcripts
NM_000087.3 NM_001142564.1 XM_005248049.4 XM_011513623.2 XM_017007712.1 NR_125879.1

Gene Symbol
LOC101927157
Entrez Gene ID
101927157
Gene Name
uncharacterized LOC101927157
Gene Aliases
-
UniGene
Hs.134190
Gene Chromosome Location
Chr.4: 47914227-47990737 on Build GRCh38
Transcripts
NM_000087.3 NM_001142564.1 XM_005248049.4 XM_011513623.2 XM_017007712.1 NR_125879.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3683287 c.1123T>G p.Y375D Chr.4: 47937371-47937371 on Build GRCh38
5653481 c.1090G>A p.G364S Chr.4: 47937404-47937404 on Build GRCh38
301959 c.1086C>A p.T362T Chr.4: 47937408-47937408 on Build GRCh38
3683288 c.1121A>G p.E374G Chr.4: 47937373-47937373 on Build GRCh38
6631112 c.1047C>T p.Y349Y Chr.4: 47937447-47937447 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759079269 Chr.4: 47937438-47937438 on Build GRCh38
rs760217797 Chr.4: 47937429-47937429 on Build GRCh38
rs150374036 Chr.4: 47937463-47937463 on Build GRCh38
rs748034014 Chr.4: 47937467-47937467 on Build GRCh38
rs749990018 Chr.4: 47937340-47937340 on Build GRCh38
rs370018275 Chr.4: 47937390-47937390 on Build GRCh38
rs140419673 Chr.4: 47937451-47937451 on Build GRCh38
rs562266812 Chr.4: 47937308-47937308 on Build GRCh38
rs764845127 Chr.4: 47937440-47937440 on Build GRCh38
rs756125453 Chr.4: 47937464-47937464 on Build GRCh38
rs377508142 Chr.4: 47937484-47937484 on Build GRCh38
rs766184739 Chr.4: 47937433-47937433 on Build GRCh38
rs780241363 Chr.4: 47937466-47937466 on Build GRCh38
rs529523430 Chr.4: 47937357-47937357 on Build GRCh38
rs776445769 Chr.4: 47937437-47937437 on Build GRCh38
rs774292169 Chr.4: 47937319-47937319 on Build GRCh38
rs757509793 Chr.4: 47937388-47937388 on Build GRCh38
rs748126956 Chr.4: 47937421-47937421 on Build GRCh38
rs370261914 Chr.4: 47937447-47937447 on Build GRCh38
rs761470459 Chr.4: 47937320-47937320 on Build GRCh38
rs1007613180 Chr.4: 47937351-47937351 on Build GRCh38
rs569092237 Chr.4: 47937387-47937387 on Build GRCh38
rs910102517 Chr.4: 47937475-47937479 on Build GRCh38
rs749874187 Chr.4: 47937406-47937406 on Build GRCh38
rs774204499 Chr.4: 47937402-47937402 on Build GRCh38
rs550831196 Chr.4: 47937318-47937318 on Build GRCh38
rs999544084 Chr.4: 47937395-47937395 on Build GRCh38
rs965734036 Chr.4: 47937403-47937403 on Build GRCh38
rs753278950 Chr.4: 47937372-47937372 on Build GRCh38
rs781347335 Chr.4: 47937460-47937460 on Build GRCh38
rs759687707 Chr.4: 47937476-47937476 on Build GRCh38
rs767346086 Chr.4: 47937334-47937334 on Build GRCh38
rs772864864 Chr.4: 47937480-47937480 on Build GRCh38
rs376177601 Chr.4: 47937446-47937446 on Build GRCh38
rs758625358 Chr.4: 47937386-47937386 on Build GRCh38
rs536327629 Chr.4: 47937404-47937404 on Build GRCh38
rs762502746 Chr.4: 47937327-47937328 on Build GRCh38
rs772091709 Chr.4: 47937471-47937471 on Build GRCh38
rs771754004 Chr.4: 47937422-47937422 on Build GRCh38
rs765792753 Chr.4: 47937361-47937361 on Build GRCh38
rs760437638 Chr.4: 47937348-47937348 on Build GRCh38
rs755520422 Chr.4: 47937360-47937360 on Build GRCh38
rs752003888 Chr.4: 47937454-47937454 on Build GRCh38
rs183197573 Chr.4: 47937424-47937424 on Build GRCh38
rs769889406 Chr.4: 47937396-47937396 on Build GRCh38
rs757637308 Chr.4: 47937458-47937458 on Build GRCh38
rs867483436 Chr.4: 47937389-47937389 on Build GRCh38
rs770901877 Chr.4: 47937304-47937304 on Build GRCh38

View Full Product Details