Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 25826229-25826498 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MTFR1L
Entrez Gene ID
56181
Gene Name
mitochondrial fission regulator 1 like
Gene Aliases
FAM54B, HYST1888, MST116, MSTP116
UniGene
Hs.149305
Gene Chromosome Location
Chr.1: 25819906-25832942 on Build GRCh38
Transcripts
NM_001099625.1 NM_001099626.1 NM_001099627.1 NM_019557.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2234674 c.214G>A p.E72K Chr.1: 25826386-25826386 on Build GRCh38
6716320 c.141C>T p.N47N Chr.1: 25826313-25826313 on Build GRCh38
6716319 c.182C>T p.T61I Chr.1: 25826354-25826354 on Build GRCh38
464338 c.239+1G>T p.? Chr.1: 25826412-25826412 on Build GRCh38
4030899 c.227A>G p.Y76C Chr.1: 25826399-25826399 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752491963 Chr.1: 25826381-25826381 on Build GRCh38
rs767246551 Chr.1: 25826425-25826425 on Build GRCh38
rs35448678 Chr.1: 25826344-25826344 on Build GRCh38
rs925237158 Chr.1: 25826418-25826418 on Build GRCh38
rs765409679 Chr.1: 25826368-25826368 on Build GRCh38
rs945151129 Chr.1: 25826464-25826464 on Build GRCh38
rs761199847 Chr.1: 25826277-25826277 on Build GRCh38
rs34991366 Chr.1: 25826382-25826382 on Build GRCh38
rs761094415 Chr.1: 25826397-25826397 on Build GRCh38
rs750671319 Chr.1: 25826307-25826307 on Build GRCh38
rs770502588 Chr.1: 25826266-25826266 on Build GRCh38
rs368834313 Chr.1: 25826367-25826367 on Build GRCh38
rs767916807 Chr.1: 25826272-25826272 on Build GRCh38
rs765378162 Chr.1: 25826408-25826408 on Build GRCh38
rs974939754 Chr.1: 25826281-25826281 on Build GRCh38
rs760522594 Chr.1: 25826415-25826415 on Build GRCh38
rs748058436 Chr.1: 25826332-25826332 on Build GRCh38
rs369504638 Chr.1: 25826325-25826325 on Build GRCh38
rs756796954 Chr.1: 25826459-25826459 on Build GRCh38
rs909734437 Chr.1: 25826327-25826327 on Build GRCh38
rs762053080 Chr.1: 25826405-25826405 on Build GRCh38
rs753929619 Chr.1: 25826358-25826358 on Build GRCh38
rs769236139 Chr.1: 25826258-25826258 on Build GRCh38
rs6598924 Chr.1: 25826267-25826267 on Build GRCh38
rs760712862 Chr.1: 25826309-25826309 on Build GRCh38
rs201685445 Chr.1: 25826299-25826299 on Build GRCh38
rs758596616 Chr.1: 25826416-25826416 on Build GRCh38
rs370001280 Chr.1: 25826261-25826261 on Build GRCh38
rs753180305 Chr.1: 25826274-25826274 on Build GRCh38
rs376576989 Chr.1: 25826271-25826271 on Build GRCh38
rs752467723 Chr.1: 25826430-25826430 on Build GRCh38
rs34144426 Chr.1: 25826364-25826364 on Build GRCh38
rs374546355 Chr.1: 25826437-25826437 on Build GRCh38
rs765615031 Chr.1: 25826303-25826303 on Build GRCh38
rs772659963 Chr.1: 25826259-25826259 on Build GRCh38
rs764640732 Chr.1: 25826398-25826398 on Build GRCh38
rs764114233 Chr.1: 25826282-25826282 on Build GRCh38
rs1016539001 Chr.1: 25826268-25826268 on Build GRCh38
rs749216197 Chr.1: 25826444-25826444 on Build GRCh38
rs748088147 Chr.1: 25826257-25826257 on Build GRCh38
rs377011692 Chr.1: 25826433-25826433 on Build GRCh38
rs756037870 Chr.1: 25826342-25826342 on Build GRCh38
rs754771390 Chr.1: 25826324-25826324 on Build GRCh38

View Full Product Details