Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 36591971-36592220 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHRS11
Entrez Gene ID
79154
Gene Name
dehydrogenase/reductase 11
Gene Aliases
ARPG836, SDR24C1, spDHRS11
UniGene
Hs.462859
Gene Chromosome Location
Chr.17: 36591791-36600804 on Build GRCh38
Transcripts
NM_024308.3 XM_005257658.2 XM_011525233.2

SNP Detail

SNP ID Chromosome Location
rs960348595 Chr.17: 36592104-36592104 on Build GRCh38
rs1051518217 Chr.17: 36592170-36592170 on Build GRCh38
rs879260339 Chr.17: 36592086-36592086 on Build GRCh38
rs969834307 Chr.17: 36591994-36591994 on Build GRCh38
rs892743423 Chr.17: 36592014-36592014 on Build GRCh38
rs372656712 Chr.17: 36592181-36592181 on Build GRCh38
rs986159887 Chr.17: 36592160-36592160 on Build GRCh38
rs759756723 Chr.17: 36592175-36592175 on Build GRCh38
rs1001974175 Chr.17: 36592080-36592080 on Build GRCh38
rs776892540 Chr.17: 36592168-36592168 on Build GRCh38
rs1015508707 Chr.17: 36592114-36592114 on Build GRCh38
rs747568347 Chr.17: 36592108-36592108 on Build GRCh38
rs550268452 Chr.17: 36592111-36592111 on Build GRCh38
rs944489833 Chr.17: 36592199-36592199 on Build GRCh38
rs754606247 Chr.17: 36592051-36592051 on Build GRCh38
rs763018770 Chr.17: 36592183-36592183 on Build GRCh38
rs747664624 Chr.17: 36592094-36592094 on Build GRCh38
rs571831892 Chr.17: 36592040-36592040 on Build GRCh38
rs766145709 Chr.17: 36592203-36592203 on Build GRCh38
rs912005228 Chr.17: 36592161-36592161 on Build GRCh38
rs921109050 Chr.17: 36592145-36592145 on Build GRCh38
rs1047764246 Chr.17: 36592060-36592069 on Build GRCh38
rs962950560 Chr.17: 36592128-36592128 on Build GRCh38
rs932044287 Chr.17: 36592155-36592155 on Build GRCh38
rs535889094 Chr.17: 36592187-36592187 on Build GRCh38
rs973899759 Chr.17: 36592137-36592137 on Build GRCh38
rs796668065 Chr.17: 36592071-36592071 on Build GRCh38
rs778411015 Chr.17: 36592100-36592100 on Build GRCh38
rs750341845 Chr.17: 36592190-36592190 on Build GRCh38
rs1043879552 Chr.17: 36592202-36592202 on Build GRCh38
rs963143057 Chr.17: 36592060-36592060 on Build GRCh38
rs760557042 Chr.17: 36592200-36592200 on Build GRCh38
rs778505157 Chr.17: 36592073-36592073 on Build GRCh38
rs1035389052 Chr.17: 36592023-36592023 on Build GRCh38
rs771773444 Chr.17: 36592097-36592097 on Build GRCh38
rs769923759 Chr.17: 36592177-36592177 on Build GRCh38
rs1053337606 Chr.17: 36592189-36592189 on Build GRCh38
rs753485461 Chr.17: 36592043-36592043 on Build GRCh38
rs1011070389 Chr.17: 36592021-36592021 on Build GRCh38
rs537212851 Chr.17: 36592064-36592064 on Build GRCh38

View Full Product Details