Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 40051907-40052158 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DSCAM
Entrez Gene ID
1826
Gene Name
DS cell adhesion molecule
Gene Aliases
CHD2, CHD2-42, CHD2-52
UniGene
Hs.397800
Gene Chromosome Location
Chr.21: 40010999-40847113 on Build GRCh38
Transcripts
NM_001271534.1 NM_001389.3 NR_073202.1 XM_017028281.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3693791 c.5177C>T p.P1726L Chr.21: 40051966-40051966 on Build GRCh38
3550793 c.5103G>A p.T1701T Chr.21: 40052040-40052040 on Build GRCh38
3550794 c.5062G>A p.D1688N Chr.21: 40052081-40052081 on Build GRCh38
4413019 c.5093A>C p.K1698T Chr.21: 40052050-40052050 on Build GRCh38
1566060 c.5069_5082del14 p.D1690fs*2 Chr.21: 40052061-40052074 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769552483 Chr.21: 40052101-40052101 on Build GRCh38
rs778990483 Chr.21: 40052047-40052047 on Build GRCh38
rs749836667 Chr.21: 40051946-40051946 on Build GRCh38
rs994959581 Chr.21: 40051966-40051966 on Build GRCh38
rs778015131 Chr.21: 40051968-40051968 on Build GRCh38
rs772835544 Chr.21: 40052029-40052029 on Build GRCh38
rs371677891 Chr.21: 40052041-40052041 on Build GRCh38
rs761507454 Chr.21: 40051932-40051932 on Build GRCh38
rs12483104 Chr.21: 40051985-40051985 on Build GRCh38
rs746027341 Chr.21: 40052062-40052062 on Build GRCh38
rs775284853 Chr.21: 40051997-40051997 on Build GRCh38
rs80135610 Chr.21: 40051981-40051981 on Build GRCh38
rs761177151 Chr.21: 40052127-40052127 on Build GRCh38
rs913310992 Chr.21: 40052076-40052076 on Build GRCh38
rs201457318 Chr.21: 40052082-40052082 on Build GRCh38
rs1045762627 Chr.21: 40052113-40052113 on Build GRCh38
rs530348291 Chr.21: 40052094-40052094 on Build GRCh38
rs142338430 Chr.21: 40051943-40051943 on Build GRCh38
rs747942534 Chr.21: 40052095-40052095 on Build GRCh38
rs149328996 Chr.21: 40052046-40052046 on Build GRCh38
rs764531692 Chr.21: 40052128-40052128 on Build GRCh38
rs371247303 Chr.21: 40052040-40052040 on Build GRCh38
rs753177407 Chr.21: 40052033-40052033 on Build GRCh38
rs1005332532 Chr.21: 40051939-40051939 on Build GRCh38
rs779058480 Chr.21: 40051974-40051974 on Build GRCh38
rs776525485 Chr.21: 40052014-40052014 on Build GRCh38
rs761560557 Chr.21: 40052022-40052022 on Build GRCh38
rs545172220 Chr.21: 40051959-40051959 on Build GRCh38
rs372192336 Chr.21: 40052093-40052093 on Build GRCh38
rs772705523 Chr.21: 40052102-40052102 on Build GRCh38
rs767893246 Chr.21: 40051960-40051960 on Build GRCh38
rs756560778 Chr.21: 40052035-40052035 on Build GRCh38
rs367766104 Chr.21: 40052003-40052003 on Build GRCh38
rs758379371 Chr.21: 40052080-40052080 on Build GRCh38
rs374542622 Chr.21: 40052070-40052070 on Build GRCh38
rs770366380 Chr.21: 40052119-40052119 on Build GRCh38
rs772106207 Chr.21: 40051986-40051986 on Build GRCh38
rs753123898 Chr.21: 40051963-40051963 on Build GRCh38
rs201772634 Chr.21: 40052028-40052028 on Build GRCh38
rs754263501 Chr.21: 40052130-40052130 on Build GRCh38
rs900479365 Chr.21: 40052026-40052026 on Build GRCh38
rs756437746 Chr.21: 40051965-40051965 on Build GRCh38
rs757432560 Chr.21: 40051973-40051973 on Build GRCh38
rs1009133746 Chr.21: 40052010-40052010 on Build GRCh38
rs768105140 Chr.21: 40052032-40052032 on Build GRCh38
rs773814290 Chr.21: 40052120-40052120 on Build GRCh38
rs527926618 Chr.21: 40052013-40052013 on Build GRCh38
rs375235951 Chr.21: 40052134-40052134 on Build GRCh38
rs199574826 Chr.21: 40051969-40051969 on Build GRCh38

View Full Product Details