Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 22582070-22582313 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXA2
Entrez Gene ID
3170
Gene Name
forkhead box A2
Gene Aliases
HNF3B, TCF3B
UniGene
Hs.155651
Gene Chromosome Location
Chr.20: 22581004-22585463 on Build GRCh38
Transcripts
NM_021784.4 NM_153675.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6436181 c.991A>G p.S331G Chr.20: 22582233-22582233 on Build GRCh38
1025149 c.1023_1024delGC p.Q342fs*19 Chr.20: 22582200-22582201 on Build GRCh38
1025150 c.997C>T p.P333S Chr.20: 22582227-22582227 on Build GRCh38
306991 c.1143C>A p.L381L Chr.20: 22582081-22582081 on Build GRCh38
1025148 c.1104_1105insAC p.H369fs*41 Chr.20: 22582119-22582120 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764677040 Chr.20: 22582272-22582272 on Build GRCh38
rs140377203 Chr.20: 22582123-22582123 on Build GRCh38
rs781573020 Chr.20: 22582178-22582178 on Build GRCh38
rs562230926 Chr.20: 22582189-22582189 on Build GRCh38
rs749146776 Chr.20: 22582207-22582207 on Build GRCh38
rs199796119 Chr.20: 22582241-22582241 on Build GRCh38
rs558508968 Chr.20: 22582234-22582234 on Build GRCh38
rs979439753 Chr.20: 22582181-22582181 on Build GRCh38
rs770661422 Chr.20: 22582192-22582194 on Build GRCh38
rs200818750 Chr.20: 22582239-22582239 on Build GRCh38
rs761326049 Chr.20: 22582231-22582231 on Build GRCh38
rs773719307 Chr.20: 22582228-22582228 on Build GRCh38
rs751531721 Chr.20: 22582274-22582274 on Build GRCh38
rs565350443 Chr.20: 22582169-22582169 on Build GRCh38
rs918206114 Chr.20: 22582249-22582249 on Build GRCh38
rs781511903 Chr.20: 22582288-22582288 on Build GRCh38
rs775402453 Chr.20: 22582206-22582206 on Build GRCh38
rs563473069 Chr.20: 22582205-22582205 on Build GRCh38
rs753174581 Chr.20: 22582271-22582271 on Build GRCh38
rs542253305 Chr.20: 22582142-22582142 on Build GRCh38
rs758986166 Chr.20: 22582148-22582148 on Build GRCh38
rs984887418 Chr.20: 22582219-22582219 on Build GRCh38
rs781041744 Chr.20: 22582225-22582226 on Build GRCh38
rs757447343 Chr.20: 22582283-22582283 on Build GRCh38
rs965871393 Chr.20: 22582094-22582094 on Build GRCh38
rs968567663 Chr.20: 22582139-22582139 on Build GRCh38
rs200305574 Chr.20: 22582185-22582185 on Build GRCh38
rs964403017 Chr.20: 22582247-22582247 on Build GRCh38
rs751804752 Chr.20: 22582285-22582293 on Build GRCh38
rs988419655 Chr.20: 22582101-22582101 on Build GRCh38
rs765489023 Chr.20: 22582251-22582251 on Build GRCh38
rs777713858 Chr.20: 22582156-22582156 on Build GRCh38
rs1210131 Chr.20: 22582184-22582184 on Build GRCh38
rs754143212 Chr.20: 22582126-22582126 on Build GRCh38
rs755340142 Chr.20: 22582127-22582127 on Build GRCh38
rs767212576 Chr.20: 22582124-22582124 on Build GRCh38
rs199678088 Chr.20: 22582187-22582187 on Build GRCh38
rs768704026 Chr.20: 22582217-22582217 on Build GRCh38
rs747174796 Chr.20: 22582157-22582157 on Build GRCh38
rs576889997 Chr.20: 22582295-22582295 on Build GRCh38
rs773948952 Chr.20: 22582115-22582115 on Build GRCh38
rs765684208 Chr.20: 22582134-22582134 on Build GRCh38

View Full Product Details