Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 30944765-30945034 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GALNT14
Entrez Gene ID
79623
Gene Name
polypeptide N-acetylgalactosaminyltransferase 14
Gene Aliases
GALNT15, GalNac-T10, GalNac-T14
UniGene
Hs.468058
Gene Chromosome Location
Chr.2: 30888518-31138726 on Build GRCh38
Transcripts
NM_001253826.1 NM_001253827.1 NM_024572.3 XM_011533105.1 XM_017004906.1 XM_017004907.1 XM_017004908.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6669025 c.817G>A p.E273K Chr.2: 30944868-30944868 on Build GRCh38
5396896 c.784C>T p.P262S Chr.2: 30944901-30944901 on Build GRCh38
377567 c.785C>G p.P262R Chr.2: 30944900-30944900 on Build GRCh38
5396897 c.783C>T p.S261S Chr.2: 30944902-30944902 on Build GRCh38
7369165 c.781T>C p.S261P Chr.2: 30944904-30944904 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs925990094 Chr.2: 30944965-30944965 on Build GRCh38
rs199792871 Chr.2: 30944889-30944889 on Build GRCh38
rs780108826 Chr.2: 30944831-30944831 on Build GRCh38
rs776978137 Chr.2: 30944884-30944884 on Build GRCh38
rs745575848 Chr.2: 30944973-30944973 on Build GRCh38
rs766434664 Chr.2: 30944900-30944900 on Build GRCh38
rs993760072 Chr.2: 30944860-30944860 on Build GRCh38
rs139053216 Chr.2: 30944869-30944869 on Build GRCh38
rs762711851 Chr.2: 30944896-30944896 on Build GRCh38
rs556866271 Chr.2: 30944935-30944935 on Build GRCh38
rs755502476 Chr.2: 30945003-30945003 on Build GRCh38
rs770652926 Chr.2: 30944881-30944881 on Build GRCh38
rs767281270 Chr.2: 30944931-30944931 on Build GRCh38
rs1023455216 Chr.2: 30944946-30944946 on Build GRCh38
rs533593653 Chr.2: 30944822-30944822 on Build GRCh38
rs748784694 Chr.2: 30944880-30944880 on Build GRCh38
rs568015783 Chr.2: 30944890-30944890 on Build GRCh38
rs748485333 Chr.2: 30944993-30944993 on Build GRCh38
rs143143842 Chr.2: 30944883-30944883 on Build GRCh38
rs770256628 Chr.2: 30944828-30944828 on Build GRCh38
rs754753068 Chr.2: 30944839-30944839 on Build GRCh38
rs777482134 Chr.2: 30944875-30944875 on Build GRCh38
rs369801106 Chr.2: 30944820-30944820 on Build GRCh38
rs536636313 Chr.2: 30944893-30944893 on Build GRCh38
rs201925273 Chr.2: 30944955-30944955 on Build GRCh38
rs769848980 Chr.2: 30944870-30944870 on Build GRCh38
rs761702989 Chr.2: 30944842-30944843 on Build GRCh38
rs1016678988 Chr.2: 30944805-30944805 on Build GRCh38
rs758672960 Chr.2: 30944826-30944826 on Build GRCh38
rs771744679 Chr.2: 30944975-30944975 on Build GRCh38
rs988801949 Chr.2: 30944984-30944984 on Build GRCh38
rs148827477 Chr.2: 30944979-30944979 on Build GRCh38
rs755159802 Chr.2: 30944928-30944928 on Build GRCh38
rs760051562 Chr.2: 30944797-30944797 on Build GRCh38
rs980181037 Chr.2: 30944962-30944962 on Build GRCh38
rs751766658 Chr.2: 30944838-30944838 on Build GRCh38
rs778426622 Chr.2: 30944966-30944966 on Build GRCh38
rs201375957 Chr.2: 30944882-30944882 on Build GRCh38
rs751745088 Chr.2: 30944910-30944910 on Build GRCh38
rs898250725 Chr.2: 30944833-30944833 on Build GRCh38
rs762369063 Chr.2: 30944885-30944885 on Build GRCh38
rs749193997 Chr.2: 30944951-30944951 on Build GRCh38
rs777817334 Chr.2: 30944947-30944947 on Build GRCh38
rs780985155 Chr.2: 30944863-30944863 on Build GRCh38
rs756168790 Chr.2: 30944940-30944940 on Build GRCh38
rs138204092 Chr.2: 30944886-30944886 on Build GRCh38

View Full Product Details