Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 45816416-45816673 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRHR1
Entrez Gene ID
1394
Gene Name
corticotropin releasing hormone receptor 1
Gene Aliases
CRF-R, CRF-R-1, CRF-R1, CRF1, CRFR-1, CRFR1, CRH-R-1, CRH-R1, CRHR, CRHR1L
UniGene
Hs.417628
Gene Chromosome Location
Chr.17: 45784280-45835828 on Build GRCh38
Transcripts
NM_001145146.1 NM_001145147.1 NM_001145148.1 NM_001303018.1 NM_001303020.1 NM_004382.4 NM_001256299.2 NM_001303016.1

Gene Symbol
MGC57346-CRHR1
Entrez Gene ID
104909134
Gene Name
MGC57346-CRHR1 readthrough
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.17: 45620329-45835828 on Build GRCh38
Transcripts
NM_001145146.1 NM_001145147.1 NM_001145148.1 NM_001303018.1 NM_001303020.1 NM_004382.4 NM_001256299.2 NM_001303016.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5833314 c.218delA p.Y73fs*25 Chr.17: 45816559-45816559 on Build GRCh38
1383819 c.169C>T p.R57C Chr.17: 45816510-45816510 on Build GRCh38
1383818 c.145G>A p.D49N Chr.17: 45816486-45816486 on Build GRCh38
706086 c.184C>A p.Q62K Chr.17: 45816525-45816525 on Build GRCh38
4579770 c.210C>G p.A70A Chr.17: 45816551-45816551 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs16940655 Chr.17: 45816520-45816520 on Build GRCh38
rs538744624 Chr.17: 45816467-45816467 on Build GRCh38
rs929229913 Chr.17: 45816511-45816511 on Build GRCh38
rs1035277621 Chr.17: 45816482-45816482 on Build GRCh38
rs976852629 Chr.17: 45816578-45816578 on Build GRCh38
rs763057553 Chr.17: 45816448-45816448 on Build GRCh38
rs371408858 Chr.17: 45816555-45816555 on Build GRCh38
rs558442160 Chr.17: 45816493-45816493 on Build GRCh38
rs373364791 Chr.17: 45816500-45816500 on Build GRCh38
rs752343666 Chr.17: 45816631-45816632 on Build GRCh38
rs760589181 Chr.17: 45816568-45816568 on Build GRCh38
rs989104073 Chr.17: 45816490-45816490 on Build GRCh38
rs768850949 Chr.17: 45816545-45816545 on Build GRCh38
rs1002654749 Chr.17: 45816538-45816538 on Build GRCh38
rs911300898 Chr.17: 45816525-45816525 on Build GRCh38
rs750228881 Chr.17: 45816455-45816456 on Build GRCh38
rs746206222 Chr.17: 45816633-45816633 on Build GRCh38
rs574439783 Chr.17: 45816649-45816649 on Build GRCh38
rs747554865 Chr.17: 45816514-45816514 on Build GRCh38
rs771142847 Chr.17: 45816516-45816516 on Build GRCh38
rs767901943 Chr.17: 45816628-45816628 on Build GRCh38
rs149144271 Chr.17: 45816449-45816449 on Build GRCh38
rs560345108 Chr.17: 45816622-45816622 on Build GRCh38
rs888600879 Chr.17: 45816436-45816436 on Build GRCh38
rs751958030 Chr.17: 45816620-45816620 on Build GRCh38
rs778498473 Chr.17: 45816614-45816614 on Build GRCh38
rs8077279 Chr.17: 45816518-45816518 on Build GRCh38
rs374275151 Chr.17: 45816593-45816593 on Build GRCh38
rs755852981 Chr.17: 45816477-45816477 on Build GRCh38
rs922569051 Chr.17: 45816582-45816582 on Build GRCh38
rs751574843 Chr.17: 45816456-45816456 on Build GRCh38
rs368605173 Chr.17: 45816537-45816537 on Build GRCh38
rs767352596 Chr.17: 45816470-45816470 on Build GRCh38
rs942722884 Chr.17: 45816574-45816574 on Build GRCh38
rs776998696 Chr.17: 45816517-45816517 on Build GRCh38
rs757810291 Chr.17: 45816621-45816621 on Build GRCh38
rs774093576 Chr.17: 45816549-45816549 on Build GRCh38
rs748497078 Chr.17: 45816476-45816476 on Build GRCh38
rs775735379 Chr.17: 45816521-45816521 on Build GRCh38
rs779673418 Chr.17: 45816483-45816483 on Build GRCh38
rs763010512 Chr.17: 45816535-45816535 on Build GRCh38
rs778136105 Chr.17: 45816510-45816510 on Build GRCh38
rs761827423 Chr.17: 45816551-45816551 on Build GRCh38
rs116593005 Chr.17: 45816444-45816444 on Build GRCh38
rs367597306 Chr.17: 45816603-45816603 on Build GRCh38

View Full Product Details