Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 15537402-15537675 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP4F22
Entrez Gene ID
126410
Gene Name
cytochrome P450 family 4 subfamily F member 22
Gene Aliases
ARCI5, INLNE, LI3
UniGene
Hs.156452
Gene Chromosome Location
Chr.19: 15508487-15552317 on Build GRCh38
Transcripts
NM_173483.3 XM_011527692.2 XM_011527693.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3283844 c.534C>T p.S178S Chr.19: 15537647-15537647 on Build GRCh38
3283845 c.538G>A p.D180N Chr.19: 15537651-15537651 on Build GRCh38
3283839 c.466C>T p.R156C Chr.19: 15537579-15537579 on Build GRCh38
709661 c.467G>T p.R156L Chr.19: 15537580-15537580 on Build GRCh38
3283840 c.467G>A p.R156H Chr.19: 15537580-15537580 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753883685 Chr.19: 15537621-15537621 on Build GRCh38
rs1003060629 Chr.19: 15537578-15537578 on Build GRCh38
rs749840377 Chr.19: 15537566-15537566 on Build GRCh38
rs750743002 Chr.19: 15537606-15537606 on Build GRCh38
rs751335732 Chr.19: 15537559-15537559 on Build GRCh38
rs368139333 Chr.19: 15537490-15537490 on Build GRCh38
rs752980761 Chr.19: 15537491-15537492 on Build GRCh38
rs16980531 Chr.19: 15537645-15537645 on Build GRCh38
rs762648974 Chr.19: 15537530-15537530 on Build GRCh38
rs775553816 Chr.19: 15537516-15537516 on Build GRCh38
rs758816247 Chr.19: 15537648-15537648 on Build GRCh38
rs942677357 Chr.19: 15537476-15537476 on Build GRCh38
rs780816103 Chr.19: 15537649-15537649 on Build GRCh38
rs759047343 Chr.19: 15537511-15537511 on Build GRCh38
rs765096948 Chr.19: 15537512-15537512 on Build GRCh38
rs199641250 Chr.19: 15537583-15537583 on Build GRCh38
rs755471652 Chr.19: 15537634-15537634 on Build GRCh38
rs868030805 Chr.19: 15537629-15537629 on Build GRCh38
rs137942239 Chr.19: 15537537-15537537 on Build GRCh38
rs187004457 Chr.19: 15537598-15537598 on Build GRCh38
rs374464817 Chr.19: 15537596-15537596 on Build GRCh38
rs879290617 Chr.19: 15537425-15537425 on Build GRCh38
rs750472966 Chr.19: 15537561-15537561 on Build GRCh38
rs368813073 Chr.19: 15537590-15537590 on Build GRCh38
rs769133197 Chr.19: 15537466-15537466 on Build GRCh38
rs114980833 Chr.19: 15537560-15537560 on Build GRCh38
rs377542346 Chr.19: 15537547-15537547 on Build GRCh38
rs772707707 Chr.19: 15537486-15537486 on Build GRCh38
rs779288178 Chr.19: 15537574-15537574 on Build GRCh38
rs535257155 Chr.19: 15537432-15537437 on Build GRCh38
rs756321971 Chr.19: 15537562-15537562 on Build GRCh38
rs770592663 Chr.19: 15537497-15537497 on Build GRCh38
rs776078737 Chr.19: 15537498-15537498 on Build GRCh38
rs973011727 Chr.19: 15537558-15537558 on Build GRCh38
rs377230723 Chr.19: 15537474-15537474 on Build GRCh38
rs776275777 Chr.19: 15537580-15537580 on Build GRCh38
rs745979345 Chr.19: 15537424-15537424 on Build GRCh38
rs761196649 Chr.19: 15537582-15537582 on Build GRCh38
rs764002212 Chr.19: 15537533-15537533 on Build GRCh38
rs372117862 Chr.19: 15537502-15537502 on Build GRCh38
rs141755148 Chr.19: 15537605-15537605 on Build GRCh38
rs749604934 Chr.19: 15537462-15537462 on Build GRCh38
rs770359214 Chr.19: 15537432-15537432 on Build GRCh38
rs780704118 Chr.19: 15537563-15537563 on Build GRCh38
rs780720676 Chr.19: 15537449-15537449 on Build GRCh38
rs766528561 Chr.19: 15537614-15537614 on Build GRCh38
rs1010835839 Chr.19: 15537627-15537627 on Build GRCh38
rs373223970 Chr.19: 15537613-15537613 on Build GRCh38
rs146036982 Chr.19: 15537571-15537571 on Build GRCh38
rs770500550 Chr.19: 15537579-15537579 on Build GRCh38
rs755273482 Chr.19: 15537573-15537573 on Build GRCh38
rs141382984 Chr.19: 15537576-15537576 on Build GRCh38
rs200108978 Chr.19: 15537422-15537422 on Build GRCh38
rs569166154 Chr.19: 15537597-15537597 on Build GRCh38
rs775270460 Chr.19: 15537586-15537586 on Build GRCh38
rs377373101 Chr.19: 15537647-15537647 on Build GRCh38

View Full Product Details