Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7225778-7226045 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DVL2
Entrez Gene ID
1856
Gene Name
dishevelled segment polarity protein 2
Gene Aliases
-
UniGene
Hs.118640
Gene Chromosome Location
Chr.17: 7225342-7234548 on Build GRCh38
Transcripts
NM_004422.2 XM_005256502.1 XM_017024298.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4069208 c.2174T>C p.M725T Chr.17: 7225902-7225902 on Build GRCh38
5596634 c.2187C>T p.S729S Chr.17: 7225889-7225889 on Build GRCh38
7415064 c.2211G>T p.*737Y Chr.17: 7225865-7225865 on Build GRCh38
6523199 c.2152C>T p.R718C Chr.17: 7225924-7225924 on Build GRCh38
384759 c.2143A>C p.T715P Chr.17: 7225933-7225933 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770399260 Chr.17: 7226015-7226015 on Build GRCh38
rs751934976 Chr.17: 7226003-7226003 on Build GRCh38
rs373299689 Chr.17: 7225889-7225889 on Build GRCh38
rs769201038 Chr.17: 7225948-7225948 on Build GRCh38
rs751418609 Chr.17: 7225892-7225892 on Build GRCh38
rs370334570 Chr.17: 7226014-7226014 on Build GRCh38
rs201947862 Chr.17: 7225958-7225958 on Build GRCh38
rs922618549 Chr.17: 7225983-7225983 on Build GRCh38
rs772583318 Chr.17: 7225837-7225837 on Build GRCh38
rs761271808 Chr.17: 7225847-7225847 on Build GRCh38
rs777821724 Chr.17: 7225821-7225821 on Build GRCh38
rs147861581 Chr.17: 7226008-7226008 on Build GRCh38
rs754891875 Chr.17: 7225900-7225900 on Build GRCh38
rs748841149 Chr.17: 7226026-7226026 on Build GRCh38
rs750574732 Chr.17: 7225990-7225990 on Build GRCh38
rs190342528 Chr.17: 7225824-7225824 on Build GRCh38
rs774108089 Chr.17: 7225976-7225976 on Build GRCh38
rs769765897 Chr.17: 7225822-7225823 on Build GRCh38
rs780465165 Chr.17: 7225997-7225997 on Build GRCh38
rs767490193 Chr.17: 7225902-7225902 on Build GRCh38
rs201114631 Chr.17: 7225873-7225873 on Build GRCh38
rs757476635 Chr.17: 7225828-7225828 on Build GRCh38
rs746147990 Chr.17: 7225904-7225904 on Build GRCh38
rs777896852 Chr.17: 7226002-7226003 on Build GRCh38
rs990046300 Chr.17: 7225890-7225890 on Build GRCh38
rs144529320 Chr.17: 7225985-7225985 on Build GRCh38
rs996082042 Chr.17: 7225807-7225807 on Build GRCh38
rs758673316 Chr.17: 7225995-7225995 on Build GRCh38
rs1015385824 Chr.17: 7225798-7225798 on Build GRCh38
rs769344849 Chr.17: 7225845-7225845 on Build GRCh38
rs562872082 Chr.17: 7225806-7225806 on Build GRCh38
rs747427559 Chr.17: 7225925-7225925 on Build GRCh38
rs757298992 Chr.17: 7225981-7225981 on Build GRCh38
rs769256723 Chr.17: 7225933-7225933 on Build GRCh38
rs752593583 Chr.17: 7225980-7225980 on Build GRCh38
rs766166785 Chr.17: 7225891-7225891 on Build GRCh38
rs141433972 Chr.17: 7226013-7226013 on Build GRCh38
rs186227085 Chr.17: 7225822-7225822 on Build GRCh38
rs776260732 Chr.17: 7225846-7225846 on Build GRCh38
rs58009529 Chr.17: 7225853-7225853 on Build GRCh38
rs747644543 Chr.17: 7225946-7225946 on Build GRCh38
rs183704016 Chr.17: 7225803-7225803 on Build GRCh38
rs201451263 Chr.17: 7225836-7225836 on Build GRCh38
rs202054703 Chr.17: 7225923-7225923 on Build GRCh38
rs764902925 Chr.17: 7225856-7225856 on Build GRCh38
rs777033867 Chr.17: 7225941-7225941 on Build GRCh38
rs779037587 Chr.17: 7225832-7225832 on Build GRCh38
rs780552994 Chr.17: 7225924-7225924 on Build GRCh38
rs753921561 Chr.17: 7225903-7225903 on Build GRCh38
rs375466737 Chr.17: 7226016-7226016 on Build GRCh38
rs139229864 Chr.17: 7225919-7225919 on Build GRCh38
rs201425014 Chr.17: 7225979-7225979 on Build GRCh38
rs772854013 Chr.17: 7225952-7225952 on Build GRCh38
rs867994732 Chr.17: 7225989-7225989 on Build GRCh38
rs747515228 Chr.17: 7225841-7225841 on Build GRCh38
rs776076357 Chr.17: 7225840-7225840 on Build GRCh38
rs765232184 Chr.17: 7225988-7225988 on Build GRCh38

View Full Product Details