Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 14751471-14751698 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADGRE2
Entrez Gene ID
30817
Gene Name
adhesion G protein-coupled receptor E2
Gene Aliases
CD312, EMR2, VBU
UniGene
Hs.531619
Gene Chromosome Location
Chr.19: 14732697-14778541 on Build GRCh38
Transcripts
NM_001271052.1 NM_013447.3 XM_011527948.2 XM_011527949.2 XM_011527951.2 XM_011527952.2 XM_011527953.2 XM_011527954.2 XM_011527955.2 XM_017026726.1 XM_017026727.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7126761 c.1842G>C p.L614F Chr.19: 14751618-14751618 on Build GRCh38
4653042 c.1921A>G p.R641G Chr.19: 14751539-14751539 on Build GRCh38
4074868 c.1907A>G p.Y636C Chr.19: 14751553-14751553 on Build GRCh38
6557556 c.1876C>A p.L626I Chr.19: 14751584-14751584 on Build GRCh38
5680587 c.1902C>A p.V634V Chr.19: 14751558-14751558 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760441774 Chr.19: 14751550-14751550 on Build GRCh38
rs760081009 Chr.19: 14751653-14751653 on Build GRCh38
rs767441479 Chr.19: 14751587-14751587 on Build GRCh38
rs368691146 Chr.19: 14751677-14751677 on Build GRCh38
rs778069519 Chr.19: 14751590-14751590 on Build GRCh38
rs374478211 Chr.19: 14751676-14751676 on Build GRCh38
rs113941589 Chr.19: 14751541-14751541 on Build GRCh38
rs923591343 Chr.19: 14751582-14751582 on Build GRCh38
rs45563436 Chr.19: 14751565-14751565 on Build GRCh38
rs756780184 Chr.19: 14751501-14751501 on Build GRCh38
rs765184165 Chr.19: 14751662-14751662 on Build GRCh38
rs779848758 Chr.19: 14751654-14751654 on Build GRCh38
rs374518362 Chr.19: 14751520-14751520 on Build GRCh38
rs759208927 Chr.19: 14751661-14751661 on Build GRCh38
rs781288814 Chr.19: 14751611-14751611 on Build GRCh38
rs143530914 Chr.19: 14751494-14751494 on Build GRCh38
rs202154380 Chr.19: 14751637-14751637 on Build GRCh38
rs751566429 Chr.19: 14751591-14751591 on Build GRCh38
rs145425662 Chr.19: 14751584-14751584 on Build GRCh38
rs780336044 Chr.19: 14751640-14751640 on Build GRCh38
rs997616385 Chr.19: 14751670-14751670 on Build GRCh38
rs752539813 Chr.19: 14751666-14751666 on Build GRCh38
rs763381476 Chr.19: 14751518-14751518 on Build GRCh38
rs763027863 Chr.19: 14751667-14751667 on Build GRCh38
rs765059458 Chr.19: 14751570-14751570 on Build GRCh38
rs143659792 Chr.19: 14751650-14751650 on Build GRCh38
rs4410209 Chr.19: 14751646-14751646 on Build GRCh38
rs994249363 Chr.19: 14751659-14751659 on Build GRCh38
rs368370128 Chr.19: 14751531-14751531 on Build GRCh38
rs111634121 Chr.19: 14751574-14751574 on Build GRCh38
rs772817258 Chr.19: 14751651-14751651 on Build GRCh38
rs370157037 Chr.19: 14751503-14751503 on Build GRCh38
rs2524383 Chr.19: 14751618-14751618 on Build GRCh38
rs757552971 Chr.19: 14751606-14751606 on Build GRCh38
rs753481314 Chr.19: 14751564-14751564 on Build GRCh38
rs765916098 Chr.19: 14751558-14751558 on Build GRCh38
rs776005214 Chr.19: 14751504-14751504 on Build GRCh38
rs534741485 Chr.19: 14751655-14751655 on Build GRCh38
rs766380701 Chr.19: 14751535-14751535 on Build GRCh38

View Full Product Details