Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 89314848-89315121 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABRR2
Entrez Gene ID
2570
Gene Name
gamma-aminobutyric acid type A receptor rho2 subunit
Gene Aliases
-
UniGene
Hs.379330
Gene Chromosome Location
Chr.6: 89254464-89315299 on Build GRCh38
Transcripts
NM_002043.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4644755 c.154C>T p.R52* Chr.6: 89315087-89315087 on Build GRCh38
6439218 c.97G>A p.E33K Chr.6: 89315069-89315069 on Build GRCh38
4357407 c.127G>A p.V43M Chr.6: 89315114-89315114 on Build GRCh38
5357821 c.173A>G p.E58G Chr.6: 89315068-89315068 on Build GRCh38
4812526 c.121G>T p.V41F Chr.6: 89315120-89315120 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs535400421 Chr.6: 89314955-89314955 on Build GRCh38
rs534841130 Chr.6: 89314907-89314907 on Build GRCh38
rs747471702 Chr.6: 89315015-89315015 on Build GRCh38
rs951769919 Chr.6: 89314894-89314894 on Build GRCh38
rs756116338 Chr.6: 89315033-89315033 on Build GRCh38
rs748499069 Chr.6: 89315050-89315050 on Build GRCh38
rs368041500 Chr.6: 89315084-89315084 on Build GRCh38
rs373983940 Chr.6: 89315087-89315087 on Build GRCh38
rs9344923 Chr.6: 89315042-89315042 on Build GRCh38
rs942207100 Chr.6: 89314927-89314927 on Build GRCh38
rs145522581 Chr.6: 89314971-89314971 on Build GRCh38
rs1023332332 Chr.6: 89314870-89314870 on Build GRCh38
rs114487708 Chr.6: 89314950-89314950 on Build GRCh38
rs908064651 Chr.6: 89314922-89314922 on Build GRCh38
rs544086673 Chr.6: 89314993-89314993 on Build GRCh38
rs750476427 Chr.6: 89315018-89315025 on Build GRCh38
rs757870089 Chr.6: 89315004-89315004 on Build GRCh38
rs41273313 Chr.6: 89314962-89314962 on Build GRCh38
rs770316103 Chr.6: 89315063-89315063 on Build GRCh38
rs182930849 Chr.6: 89314913-89314913 on Build GRCh38
rs148209347 Chr.6: 89315086-89315086 on Build GRCh38
rs768590491 Chr.6: 89314900-89314900 on Build GRCh38
rs779584644 Chr.6: 89315006-89315006 on Build GRCh38
rs755450220 Chr.6: 89315032-89315032 on Build GRCh38

View Full Product Details