Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 117734898-117735125 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTTNBP2
Entrez Gene ID
83992
Gene Name
cortactin binding protein 2
Gene Aliases
C7orf8, CORTBP2, Orf4
UniGene
Hs.592285
Gene Chromosome Location
Chr.7: 117710648-117873507 on Build GRCh38
Transcripts
NM_033427.2 XM_005250634.4 XM_005250635.3 XM_011516615.2 XM_011516616.2 XM_017012706.1 XM_017012708.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3831711 c.3712T>C p.Y1238H Chr.7: 117735077-117735077 on Build GRCh38
3257371 c.3770C>T p.S1257F Chr.7: 117735019-117735019 on Build GRCh38
5898921 c.3857G>A p.R1286Q Chr.7: 117734932-117734932 on Build GRCh38
1622249 c.3702C>T p.S1234S Chr.7: 117735087-117735087 on Build GRCh38
1568667 c.3701C>T p.S1234F Chr.7: 117735088-117735088 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs944856056 Chr.7: 117735080-117735080 on Build GRCh38
rs773516534 Chr.7: 117735053-117735053 on Build GRCh38
rs767012577 Chr.7: 117734968-117734968 on Build GRCh38
rs756503736 Chr.7: 117734967-117734967 on Build GRCh38
rs761917492 Chr.7: 117735027-117735027 on Build GRCh38
rs200782612 Chr.7: 117734992-117734992 on Build GRCh38
rs1047966732 Chr.7: 117735017-117735017 on Build GRCh38
rs776346934 Chr.7: 117734931-117734931 on Build GRCh38
rs775369236 Chr.7: 117734934-117734934 on Build GRCh38
rs769531182 Chr.7: 117734999-117734999 on Build GRCh38
rs779981810 Chr.7: 117735000-117735000 on Build GRCh38
rs775826231 Chr.7: 117735021-117735045 on Build GRCh38
rs774639892 Chr.7: 117734949-117734949 on Build GRCh38
rs780731601 Chr.7: 117735087-117735087 on Build GRCh38
rs375469429 Chr.7: 117735001-117735001 on Build GRCh38
rs372382974 Chr.7: 117734993-117734993 on Build GRCh38
rs35288952 Chr.7: 117734994-117734995 on Build GRCh38
rs750789291 Chr.7: 117734966-117734966 on Build GRCh38
rs770531339 Chr.7: 117735066-117735066 on Build GRCh38
rs758953932 Chr.7: 117734981-117735001 on Build GRCh38
rs769392706 Chr.7: 117735016-117735017 on Build GRCh38
rs754336610 Chr.7: 117734972-117734972 on Build GRCh38
rs992110761 Chr.7: 117735097-117735097 on Build GRCh38
rs543041784 Chr.7: 117734932-117734932 on Build GRCh38
rs762012037 Chr.7: 117734956-117734956 on Build GRCh38
rs761131670 Chr.7: 117735054-117735054 on Build GRCh38
rs561305383 Chr.7: 117734933-117734933 on Build GRCh38
rs749663962 Chr.7: 117735021-117735021 on Build GRCh38
rs751071829 Chr.7: 117735076-117735076 on Build GRCh38
rs772211877 Chr.7: 117735041-117735041 on Build GRCh38
rs755910571 Chr.7: 117735092-117735092 on Build GRCh38
rs763044974 Chr.7: 117734941-117734941 on Build GRCh38
rs149557894 Chr.7: 117734978-117734978 on Build GRCh38
rs766782453 Chr.7: 117735063-117735063 on Build GRCh38
rs748672952 Chr.7: 117734928-117734928 on Build GRCh38
rs751065536 Chr.7: 117734987-117734987 on Build GRCh38
rs144248062 Chr.7: 117735093-117735093 on Build GRCh38
rs1049021862 Chr.7: 117734937-117734937 on Build GRCh38
rs573012431 Chr.7: 117734959-117734959 on Build GRCh38
rs561099662 Chr.7: 117735032-117735032 on Build GRCh38
rs199634048 Chr.7: 117735077-117735077 on Build GRCh38
rs768638495 Chr.7: 117735018-117735018 on Build GRCh38
rs368897036 Chr.7: 117734977-117734977 on Build GRCh38
rs146038841 Chr.7: 117735071-117735071 on Build GRCh38
rs764106006 Chr.7: 117734943-117734943 on Build GRCh38
rs765752389 Chr.7: 117735074-117735074 on Build GRCh38

View Full Product Details