Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 78631129-78631360 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRNB4
Entrez Gene ID
1143
Gene Name
cholinergic receptor nicotinic beta 4 subunit
Gene Aliases
-
UniGene
Hs.624178
Gene Chromosome Location
Chr.15: 78623282-78655586 on Build GRCh38
Transcripts
NM_000750.3 NM_001256567.1 XM_011521186.2 XM_011521187.2 XM_011521190.2 XM_011521191.2 XM_011521192.2 XM_017021885.1 XM_017021886.1 XM_017021887.1 XM_017021888.1 XM_017021889.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5440596 c.265C>T p.R89C Chr.15: 78631170-78631170 on Build GRCh38
3504339 c.255G>A p.W85* Chr.15: 78631180-78631180 on Build GRCh38
2011234 c.295G>A p.G99S Chr.15: 78631140-78631140 on Build GRCh38
6312733 c.272C>A p.T91N Chr.15: 78631163-78631163 on Build GRCh38
5567005 c.266G>A p.R89H Chr.15: 78631169-78631169 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1007333155 Chr.15: 78631173-78631173 on Build GRCh38
rs762780433 Chr.15: 78631338-78631338 on Build GRCh38
rs866726298 Chr.15: 78631266-78631266 on Build GRCh38
rs775450590 Chr.15: 78631337-78631337 on Build GRCh38
rs776484763 Chr.15: 78631286-78631286 on Build GRCh38
rs756474226 Chr.15: 78631237-78631237 on Build GRCh38
rs368701860 Chr.15: 78631223-78631223 on Build GRCh38
rs770463086 Chr.15: 78631276-78631276 on Build GRCh38
rs772788774 Chr.15: 78631272-78631272 on Build GRCh38
rs372231167 Chr.15: 78631303-78631303 on Build GRCh38
rs888855338 Chr.15: 78631256-78631256 on Build GRCh38
rs757408203 Chr.15: 78631209-78631209 on Build GRCh38
rs879114631 Chr.15: 78631153-78631153 on Build GRCh38
rs749744080 Chr.15: 78631243-78631243 on Build GRCh38
rs749603795 Chr.15: 78631262-78631262 on Build GRCh38
rs1027117404 Chr.15: 78631314-78631314 on Build GRCh38
rs746592611 Chr.15: 78631275-78631275 on Build GRCh38
rs767411189 Chr.15: 78631265-78631274 on Build GRCh38
rs764362711 Chr.15: 78631183-78631183 on Build GRCh38
rs868731332 Chr.15: 78631309-78631310 on Build GRCh38
rs376605650 Chr.15: 78631210-78631210 on Build GRCh38
rs374441948 Chr.15: 78631241-78631241 on Build GRCh38
rs752904319 Chr.15: 78631170-78631170 on Build GRCh38
rs377516897 Chr.15: 78631224-78631224 on Build GRCh38
rs765046135 Chr.15: 78631326-78631326 on Build GRCh38
rs746362829 Chr.15: 78631227-78631227 on Build GRCh38
rs759310176 Chr.15: 78631169-78631169 on Build GRCh38
rs774295532 Chr.15: 78631189-78631192 on Build GRCh38
rs758726852 Chr.15: 78631178-78631178 on Build GRCh38
rs373299350 Chr.15: 78631149-78631149 on Build GRCh38
rs12914008 Chr.15: 78631163-78631163 on Build GRCh38
rs759350284 Chr.15: 78631249-78631249 on Build GRCh38
rs751704323 Chr.15: 78631195-78631195 on Build GRCh38
rs759487570 Chr.15: 78631290-78631290 on Build GRCh38

View Full Product Details