Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 113115649-113115876 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
F7
Entrez Gene ID
2155
Gene Name
coagulation factor VII
Gene Aliases
SPCA
UniGene
Hs.36989
Gene Chromosome Location
Chr.13: 113105773-113120681 on Build GRCh38
Transcripts
NM_000131.4 NM_001267554.1 NM_019616.3 NR_051961.1 XM_006719963.3 XM_011537474.2 XM_011537475.2 XM_011537476.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7366901 c.334C>A p.L112M Chr.13: 113115770-113115770 on Build GRCh38
6657794 c.327_328delCT p.L111fs*12 Chr.13: 113115763-113115764 on Build GRCh38
4821584 c.475G>C p.E159Q Chr.13: 113115704-113115704 on Build GRCh38
7422783 c.259G>A p.G87S Chr.13: 113115695-113115695 on Build GRCh38
6657793 c.534_535delCT p.L180fs*12 Chr.13: 113115763-113115764 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758351183 Chr.13: 113115683-113115683 on Build GRCh38
rs755048394 Chr.13: 113115740-113115740 on Build GRCh38
rs771465632 Chr.13: 113115726-113115726 on Build GRCh38
rs563972504 Chr.13: 113115695-113115695 on Build GRCh38
rs768973596 Chr.13: 113115690-113115690 on Build GRCh38
rs778174347 Chr.13: 113115747-113115747 on Build GRCh38
rs931452769 Chr.13: 113115756-113115756 on Build GRCh38
rs373795186 Chr.13: 113115832-113115832 on Build GRCh38
rs149461706 Chr.13: 113115733-113115733 on Build GRCh38
rs765137012 Chr.13: 113115728-113115728 on Build GRCh38
rs6040 Chr.13: 113115778-113115778 on Build GRCh38
rs139304588 Chr.13: 113115688-113115688 on Build GRCh38
rs764971156 Chr.13: 113115786-113115786 on Build GRCh38
rs774685435 Chr.13: 113115755-113115755 on Build GRCh38
rs766158245 Chr.13: 113115818-113115818 on Build GRCh38
rs372307787 Chr.13: 113115696-113115696 on Build GRCh38
rs763458490 Chr.13: 113115698-113115698 on Build GRCh38
rs977631643 Chr.13: 113115732-113115732 on Build GRCh38
rs1041302431 Chr.13: 113115841-113115841 on Build GRCh38
rs761560680 Chr.13: 113115780-113115780 on Build GRCh38
rs761507342 Chr.13: 113115727-113115727 on Build GRCh38
rs751478466 Chr.13: 113115819-113115819 on Build GRCh38
rs1050691539 Chr.13: 113115830-113115830 on Build GRCh38
rs148226963 Chr.13: 113115746-113115746 on Build GRCh38
rs182854845 Chr.13: 113115835-113115835 on Build GRCh38
rs146795869 Chr.13: 113115689-113115689 on Build GRCh38
rs200016360 Chr.13: 113115708-113115708 on Build GRCh38
rs544992484 Chr.13: 113115694-113115694 on Build GRCh38
rs771335282 Chr.13: 113115699-113115699 on Build GRCh38
rs750240162 Chr.13: 113115729-113115729 on Build GRCh38
rs928615588 Chr.13: 113115742-113115742 on Build GRCh38
rs762624669 Chr.13: 113115798-113115798 on Build GRCh38
rs375134790 Chr.13: 113115738-113115738 on Build GRCh38
rs6042 Chr.13: 113115754-113115754 on Build GRCh38
rs746665783 Chr.13: 113115843-113115843 on Build GRCh38
rs752772591 Chr.13: 113115844-113115844 on Build GRCh38
rs143855920 Chr.13: 113115741-113115741 on Build GRCh38
rs760076907 Chr.13: 113115716-113115716 on Build GRCh38
rs779834113 Chr.13: 113115737-113115737 on Build GRCh38
rs773192876 Chr.13: 113115795-113115795 on Build GRCh38
rs372005651 Chr.13: 113115779-113115779 on Build GRCh38
rs781240802 Chr.13: 113115691-113115691 on Build GRCh38
rs140716599 Chr.13: 113115767-113115767 on Build GRCh38

View Full Product Details