Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 6975131-6975404 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EMG1
Entrez Gene ID
10436
Gene Name
EMG1, N1-specific pseudouridine methyltransferase
Gene Aliases
C2F, Grcc2f, NEP1
UniGene
Hs.558447
Gene Chromosome Location
Chr.12: 6970781-6997428 on Build GRCh38
Transcripts
NM_001320049.1 NM_006331.7 NR_135131.1

SNP Detail

SNP ID Chromosome Location
rs782334197 Chr.12: 6975269-6975269 on Build GRCh38
rs782141588 Chr.12: 6975324-6975324 on Build GRCh38
rs782281203 Chr.12: 6975383-6975383 on Build GRCh38
rs782643000 Chr.12: 6975241-6975241 on Build GRCh38
rs781834410 Chr.12: 6975347-6975347 on Build GRCh38
rs187355231 Chr.12: 6975363-6975363 on Build GRCh38
rs782050058 Chr.12: 6975186-6975186 on Build GRCh38
rs201757338 Chr.12: 6975313-6975313 on Build GRCh38
rs782019701 Chr.12: 6975293-6975293 on Build GRCh38
rs368451177 Chr.12: 6975157-6975157 on Build GRCh38
rs781987441 Chr.12: 6975164-6975164 on Build GRCh38
rs900422616 Chr.12: 6975300-6975300 on Build GRCh38
rs782391946 Chr.12: 6975278-6975278 on Build GRCh38
rs781882780 Chr.12: 6975242-6975242 on Build GRCh38
rs781937222 Chr.12: 6975308-6975308 on Build GRCh38
rs782598532 Chr.12: 6975225-6975225 on Build GRCh38
rs782605254 Chr.12: 6975343-6975343 on Build GRCh38
rs782554231 Chr.12: 6975248-6975248 on Build GRCh38
rs782245976 Chr.12: 6975273-6975273 on Build GRCh38
rs183026884 Chr.12: 6975256-6975256 on Build GRCh38
rs139761201 Chr.12: 6975270-6975270 on Build GRCh38
rs782080669 Chr.12: 6975153-6975153 on Build GRCh38
rs971421303 Chr.12: 6975362-6975362 on Build GRCh38
rs781905678 Chr.12: 6975179-6975179 on Build GRCh38
rs1008578641 Chr.12: 6975291-6975291 on Build GRCh38
rs372110507 Chr.12: 6975244-6975244 on Build GRCh38
rs782021249 Chr.12: 6975317-6975317 on Build GRCh38
rs375741608 Chr.12: 6975345-6975345 on Build GRCh38
rs782741904 Chr.12: 6975316-6975316 on Build GRCh38
rs782168206 Chr.12: 6975299-6975299 on Build GRCh38
rs782795372 Chr.12: 6975173-6975173 on Build GRCh38
rs782511052 Chr.12: 6975361-6975361 on Build GRCh38
rs34784123 Chr.12: 6975333-6975333 on Build GRCh38
rs782148409 Chr.12: 6975181-6975182 on Build GRCh38
rs782307546 Chr.12: 6975302-6975302 on Build GRCh38
rs782732321 Chr.12: 6975187-6975187 on Build GRCh38
rs782416705 Chr.12: 6975325-6975325 on Build GRCh38
rs782493737 Chr.12: 6975233-6975233 on Build GRCh38
rs781782863 Chr.12: 6975330-6975330 on Build GRCh38
rs771790052 Chr.12: 6975223-6975223 on Build GRCh38

View Full Product Details