Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67675855-67676082 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GFOD2
Entrez Gene ID
81577
Gene Name
glucose-fructose oxidoreductase domain containing 2
Gene Aliases
-
UniGene
Hs.307084
Gene Chromosome Location
Chr.16: 67674533-67719370 on Build GRCh38
Transcripts
NM_030819.3 NR_027398.1 XM_006721288.3 XM_017023738.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4485983 c.299C>T p.S100L Chr.16: 67676014-67676014 on Build GRCh38
161249 c.277G>A p.V93I Chr.16: 67676036-67676036 on Build GRCh38
3511083 c.312C>T p.F104F Chr.16: 67676001-67676001 on Build GRCh38
972559 c.313C>T p.R105W Chr.16: 67676000-67676000 on Build GRCh38
6670545 c.373C>T p.R125C Chr.16: 67675940-67675940 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs140195756 Chr.16: 67676031-67676031 on Build GRCh38
rs772317050 Chr.16: 67675897-67675897 on Build GRCh38
rs759973834 Chr.16: 67676030-67676030 on Build GRCh38
rs201540487 Chr.16: 67675971-67675971 on Build GRCh38
rs761195735 Chr.16: 67676005-67676005 on Build GRCh38
rs780291977 Chr.16: 67675882-67675882 on Build GRCh38
rs748214647 Chr.16: 67675893-67675893 on Build GRCh38
rs773117453 Chr.16: 67675962-67675962 on Build GRCh38
rs766726703 Chr.16: 67675938-67675938 on Build GRCh38
rs548450884 Chr.16: 67675910-67675910 on Build GRCh38
rs780092089 Chr.16: 67675983-67675983 on Build GRCh38
rs146852241 Chr.16: 67676053-67676053 on Build GRCh38
rs377405476 Chr.16: 67675916-67675916 on Build GRCh38
rs752901884 Chr.16: 67676042-67676042 on Build GRCh38
rs142530623 Chr.16: 67675923-67675923 on Build GRCh38
rs1019379030 Chr.16: 67675985-67675985 on Build GRCh38
rs769645322 Chr.16: 67675982-67675982 on Build GRCh38
rs149276567 Chr.16: 67675899-67675899 on Build GRCh38
rs779244772 Chr.16: 67675959-67675959 on Build GRCh38
rs766991070 Chr.16: 67675922-67675922 on Build GRCh38
rs199807726 Chr.16: 67676017-67676017 on Build GRCh38
rs761464857 Chr.16: 67675997-67675997 on Build GRCh38
rs749055390 Chr.16: 67675987-67675987 on Build GRCh38
rs144593745 Chr.16: 67675977-67675977 on Build GRCh38
rs747900792 Chr.16: 67675946-67675946 on Build GRCh38
rs773977871 Chr.16: 67675993-67675993 on Build GRCh38
rs146152087 Chr.16: 67675947-67675947 on Build GRCh38
rs768507870 Chr.16: 67675990-67675990 on Build GRCh38
rs775184036 Chr.16: 67675909-67675909 on Build GRCh38
rs147858714 Chr.16: 67676013-67676013 on Build GRCh38
rs755282055 Chr.16: 67675890-67675890 on Build GRCh38
rs777720225 Chr.16: 67675972-67675972 on Build GRCh38
rs753028515 Chr.16: 67675963-67675963 on Build GRCh38
rs758580613 Chr.16: 67676049-67676049 on Build GRCh38
rs144416857 Chr.16: 67675918-67675918 on Build GRCh38
rs552516505 Chr.16: 67676000-67676000 on Build GRCh38
rs148404598 Chr.16: 67675919-67675919 on Build GRCh38
rs762547133 Chr.16: 67675913-67675913 on Build GRCh38
rs771540117 Chr.16: 67675999-67675999 on Build GRCh38
rs373292631 Chr.16: 67675887-67675887 on Build GRCh38
rs896869278 Chr.16: 67676016-67676016 on Build GRCh38
rs769566468 Chr.16: 67675900-67675900 on Build GRCh38
rs779085608 Chr.16: 67675891-67675891 on Build GRCh38
rs954479013 Chr.16: 67675924-67675924 on Build GRCh38
rs140363708 Chr.16: 67676052-67676052 on Build GRCh38
rs1008420816 Chr.16: 67675954-67675954 on Build GRCh38
rs762120591 Chr.16: 67675890-67675891 on Build GRCh38
rs756484111 Chr.16: 67675881-67675881 on Build GRCh38

View Full Product Details