Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 59124338-59124605 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM111B
Entrez Gene ID
374393
Gene Name
family with sequence similarity 111 member B
Gene Aliases
CANP, POIKTMP
UniGene
Hs.186579
Gene Chromosome Location
Chr.11: 59107185-59127415 on Build GRCh38
Transcripts
NM_001142703.1 NM_001142704.1 NM_198947.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7332177 c.277G>A p.D93N Chr.11: 59124374-59124374 on Build GRCh38
6658102 c.280C>T p.R94C Chr.11: 59124377-59124377 on Build GRCh38
327999 c.392delT p.Y132fs*6 Chr.11: 59124489-59124489 on Build GRCh38
396858 c.366G>C p.K122N Chr.11: 59124463-59124463 on Build GRCh38
7400974 c.255G>A p.L85L Chr.11: 59124352-59124352 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763358585 Chr.11: 59124381-59124381 on Build GRCh38
rs374363569 Chr.11: 59124515-59124515 on Build GRCh38
rs756153449 Chr.11: 59124415-59124415 on Build GRCh38
rs370956741 Chr.11: 59124418-59124418 on Build GRCh38
rs199851144 Chr.11: 59124489-59124489 on Build GRCh38
rs766826697 Chr.11: 59124397-59124397 on Build GRCh38
rs745734356 Chr.11: 59124456-59124456 on Build GRCh38
rs774896090 Chr.11: 59124398-59124398 on Build GRCh38
rs749780165 Chr.11: 59124507-59124507 on Build GRCh38
rs865803055 Chr.11: 59124464-59124465 on Build GRCh38
rs767223238 Chr.11: 59124465-59124466 on Build GRCh38
rs779573992 Chr.11: 59124491-59124491 on Build GRCh38
rs760761564 Chr.11: 59124537-59124537 on Build GRCh38
rs963698138 Chr.11: 59124387-59124387 on Build GRCh38
rs768070950 Chr.11: 59124497-59124497 on Build GRCh38
rs776328175 Chr.11: 59124502-59124502 on Build GRCh38
rs775607632 Chr.11: 59124533-59124533 on Build GRCh38
rs970857703 Chr.11: 59124457-59124457 on Build GRCh38
rs533200319 Chr.11: 59124522-59124522 on Build GRCh38
rs374250882 Chr.11: 59124410-59124410 on Build GRCh38
rs759833100 Chr.11: 59124404-59124404 on Build GRCh38
rs758006739 Chr.11: 59124472-59124472 on Build GRCh38
rs202196116 Chr.11: 59124560-59124560 on Build GRCh38
rs765221469 Chr.11: 59124578-59124578 on Build GRCh38
rs150852006 Chr.11: 59124409-59124409 on Build GRCh38
rs143973452 Chr.11: 59124511-59124511 on Build GRCh38
rs764055213 Chr.11: 59124417-59124417 on Build GRCh38
rs773621212 Chr.11: 59124378-59124378 on Build GRCh38
rs778650838 Chr.11: 59124453-59124453 on Build GRCh38
rs757236776 Chr.11: 59124439-59124439 on Build GRCh38
rs753962616 Chr.11: 59124541-59124541 on Build GRCh38
rs879028584 Chr.11: 59124573-59124573 on Build GRCh38
rs1025257710 Chr.11: 59124504-59124504 on Build GRCh38
rs770921473 Chr.11: 59124547-59124548 on Build GRCh38
rs139291529 Chr.11: 59124423-59124423 on Build GRCh38
rs759992548 Chr.11: 59124516-59124516 on Build GRCh38
rs746511108 Chr.11: 59124377-59124377 on Build GRCh38
rs746485552 Chr.11: 59124494-59124494 on Build GRCh38
rs764062328 Chr.11: 59124538-59124538 on Build GRCh38

View Full Product Details