Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 38404845-38405118 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM98C
Entrez Gene ID
147965
Gene Name
family with sequence similarity 98 member C
Gene Aliases
-
UniGene
Hs.355162
Gene Chromosome Location
Chr.19: 38403124-38409088 on Build GRCh38
Transcripts
NM_174905.3 XM_017026354.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1481005 c.483C>G p.L161L Chr.19: 38405041-38405041 on Build GRCh38
5855645 c.437G>A p.G146E Chr.19: 38404995-38404995 on Build GRCh38
3721747 c.453C>T p.A151A Chr.19: 38405011-38405011 on Build GRCh38
1612179 c.522C>T p.P174P Chr.19: 38405080-38405080 on Build GRCh38
7427743 c.510A>T p.A170A Chr.19: 38405068-38405068 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773735559 Chr.19: 38404948-38404948 on Build GRCh38
rs764298594 Chr.19: 38405084-38405084 on Build GRCh38
rs769754971 Chr.19: 38404925-38404925 on Build GRCh38
rs768245449 Chr.19: 38404946-38404946 on Build GRCh38
rs199758385 Chr.19: 38405080-38405080 on Build GRCh38
rs780029465 Chr.19: 38405015-38405015 on Build GRCh38
rs997473198 Chr.19: 38404959-38404959 on Build GRCh38
rs747604691 Chr.19: 38405041-38405041 on Build GRCh38
rs751327938 Chr.19: 38405007-38405008 on Build GRCh38
rs763825029 Chr.19: 38405005-38405006 on Build GRCh38
rs201084520 Chr.19: 38404968-38404968 on Build GRCh38
rs200101425 Chr.19: 38404953-38404953 on Build GRCh38
rs373599906 Chr.19: 38405077-38405077 on Build GRCh38
rs369912781 Chr.19: 38404952-38404952 on Build GRCh38
rs756800178 Chr.19: 38405008-38405009 on Build GRCh38
rs758451045 Chr.19: 38404883-38404883 on Build GRCh38
rs571892119 Chr.19: 38405059-38405059 on Build GRCh38
rs755408596 Chr.19: 38404993-38404994 on Build GRCh38
rs748878883 Chr.19: 38405024-38405024 on Build GRCh38
rs780962258 Chr.19: 38405011-38405011 on Build GRCh38
rs771652219 Chr.19: 38405067-38405067 on Build GRCh38
rs376393197 Chr.19: 38405012-38405012 on Build GRCh38
rs745647258 Chr.19: 38404909-38404909 on Build GRCh38
rs764103906 Chr.19: 38404975-38404975 on Build GRCh38
rs777852334 Chr.19: 38404888-38404888 on Build GRCh38
rs758650390 Chr.19: 38404973-38404973 on Build GRCh38
rs746898599 Chr.19: 38404890-38404890 on Build GRCh38
rs757427527 Chr.19: 38404996-38404996 on Build GRCh38
rs768431843 Chr.19: 38405032-38405032 on Build GRCh38
rs535087927 Chr.19: 38404906-38404906 on Build GRCh38
rs373413614 Chr.19: 38404969-38404969 on Build GRCh38
rs74987063 Chr.19: 38405037-38405037 on Build GRCh38
rs762721238 Chr.19: 38404938-38404938 on Build GRCh38
rs751848762 Chr.19: 38405088-38405088 on Build GRCh38
rs938305387 Chr.19: 38405061-38405061 on Build GRCh38
rs752954578 Chr.19: 38404872-38404872 on Build GRCh38
rs759787841 Chr.19: 38404961-38404961 on Build GRCh38
rs775262999 Chr.19: 38404937-38404937 on Build GRCh38
rs763367017 Chr.19: 38405082-38405082 on Build GRCh38
rs751579130 Chr.19: 38404978-38404978 on Build GRCh38

View Full Product Details