Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 150968178-150968437 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CERS2
Entrez Gene ID
29956
Gene Name
ceramide synthase 2
Gene Aliases
L3, LASS2, SP260, TMSG1
UniGene
Hs.744017
Gene Chromosome Location
Chr.1: 150965173-150975003 on Build GRCh38
Transcripts
NM_022075.4 NM_181746.3 XM_011509451.2 XM_011509452.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5473141 c.314G>A p.R105Q Chr.1: 150968179-150968179 on Build GRCh38
5936852 c.313C>T p.R105W Chr.1: 150968180-150968180 on Build GRCh38
5937933 c.284C>T p.P95L Chr.1: 150968402-150968402 on Build GRCh38
895652 c.315G>A p.R105R Chr.1: 150968178-150968178 on Build GRCh38
4671794 c.255delA p.E85fs*5 Chr.1: 150968431-150968431 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1001893967 Chr.1: 150968261-150968261 on Build GRCh38
rs906185383 Chr.1: 150968258-150968258 on Build GRCh38
rs370891037 Chr.1: 150968391-150968391 on Build GRCh38
rs774868784 Chr.1: 150968353-150968353 on Build GRCh38
rs982297389 Chr.1: 150968392-150968392 on Build GRCh38
rs989223954 Chr.1: 150968338-150968338 on Build GRCh38
rs587602258 Chr.1: 150968241-150968241 on Build GRCh38
rs753499314 Chr.1: 150968378-150968378 on Build GRCh38
rs780191276 Chr.1: 150968387-150968387 on Build GRCh38
rs759864180 Chr.1: 150968360-150968360 on Build GRCh38
rs750442438 Chr.1: 150968217-150968217 on Build GRCh38
rs587738500 Chr.1: 150968240-150968240 on Build GRCh38
rs773793016 Chr.1: 150968207-150968208 on Build GRCh38
rs587664975 Chr.1: 150968231-150968231 on Build GRCh38
rs762957467 Chr.1: 150968347-150968347 on Build GRCh38
rs748400739 Chr.1: 150968344-150968344 on Build GRCh38
rs756755798 Chr.1: 150968200-150968200 on Build GRCh38
rs957981496 Chr.1: 150968269-150968269 on Build GRCh38
rs936161388 Chr.1: 150968207-150968207 on Build GRCh38
rs1054873434 Chr.1: 150968295-150968295 on Build GRCh38
rs761512748 Chr.1: 150968379-150968379 on Build GRCh38
rs950308187 Chr.1: 150968225-150968225 on Build GRCh38
rs146940612 Chr.1: 150968400-150968400 on Build GRCh38
rs769922329 Chr.1: 150968345-150968345 on Build GRCh38
rs773296912 Chr.1: 150968346-150968346 on Build GRCh38
rs757934047 Chr.1: 150968386-150968386 on Build GRCh38
rs376191508 Chr.1: 150968381-150968381 on Build GRCh38
rs757930538 Chr.1: 150968305-150968305 on Build GRCh38
rs754799939 Chr.1: 150968398-150968398 on Build GRCh38
rs1019466237 Chr.1: 150968230-150968230 on Build GRCh38
rs377147425 Chr.1: 150968222-150968222 on Build GRCh38
rs771455150 Chr.1: 150968349-150968349 on Build GRCh38
rs969519210 Chr.1: 150968382-150968382 on Build GRCh38
rs749984571 Chr.1: 150968385-150968385 on Build GRCh38
rs587677688 Chr.1: 150968267-150968267 on Build GRCh38
rs201277521 Chr.1: 150968215-150968215 on Build GRCh38
rs755047867 Chr.1: 150968339-150968340 on Build GRCh38
rs758423224 Chr.1: 150968224-150968224 on Build GRCh38
rs201840860 Chr.1: 150968372-150968372 on Build GRCh38
rs747843520 Chr.1: 150968401-150968401 on Build GRCh38

View Full Product Details