Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 94775019-94775262 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP2C19
Entrez Gene ID
1557
Gene Name
cytochrome P450 family 2 subfamily C member 19
Gene Aliases
CPCJ, CYP2C, CYPIIC17, CYPIIC19, P450C2C, P450IIC19
UniGene
Hs.282409
Gene Chromosome Location
Chr.10: 94762681-94853205 on Build GRCh38
Transcripts
NM_000769.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1349996 c.193T>C p.F65L Chr.10: 94775082-94775082 on Build GRCh38
3675928 c.331+1G>C p.? Chr.10: 94775221-94775221 on Build GRCh38
6639197 c.268C>G p.L90V Chr.10: 94775157-94775157 on Build GRCh38
3675929 c.331+1G>T p.? Chr.10: 94775221-94775221 on Build GRCh38
228528 c.214G>A p.E72K Chr.10: 94775103-94775103 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs554542134 Chr.10: 94775099-94775099 on Build GRCh38
rs868548693 Chr.10: 94775202-94775202 on Build GRCh38
rs145328984 Chr.10: 94775106-94775106 on Build GRCh38
rs202036394 Chr.10: 94775082-94775082 on Build GRCh38
rs758114852 Chr.10: 94775149-94775149 on Build GRCh38
rs771508729 Chr.10: 94775058-94775058 on Build GRCh38
rs201306972 Chr.10: 94775107-94775107 on Build GRCh38
rs761894050 Chr.10: 94775216-94775216 on Build GRCh38
rs199549982 Chr.10: 94775236-94775236 on Build GRCh38
rs868063722 Chr.10: 94775077-94775077 on Build GRCh38
rs200347843 Chr.10: 94775215-94775215 on Build GRCh38
rs759673891 Chr.10: 94775193-94775193 on Build GRCh38
rs757145258 Chr.10: 94775135-94775135 on Build GRCh38
rs778204929 Chr.10: 94775235-94775235 on Build GRCh38
rs143075956 Chr.10: 94775154-94775154 on Build GRCh38
rs753021273 Chr.10: 94775225-94775225 on Build GRCh38
rs749613282 Chr.10: 94775182-94775182 on Build GRCh38
rs778656274 Chr.10: 94775138-94775138 on Build GRCh38
rs763322365 Chr.10: 94775086-94775086 on Build GRCh38
rs759558872 Chr.10: 94775183-94775184 on Build GRCh38
rs774817817 Chr.10: 94775189-94775189 on Build GRCh38
rs777165098 Chr.10: 94775146-94775146 on Build GRCh38
rs921062799 Chr.10: 94775224-94775224 on Build GRCh38
rs760514392 Chr.10: 94775112-94775112 on Build GRCh38
rs17878459 Chr.10: 94775165-94775165 on Build GRCh38
rs28399505 Chr.10: 94775110-94775110 on Build GRCh38
rs1009854414 Chr.10: 94775047-94775047 on Build GRCh38
rs771308153 Chr.10: 94775183-94775183 on Build GRCh38
rs754514180 Chr.10: 94775232-94775232 on Build GRCh38
rs770229037 Chr.10: 94775169-94775169 on Build GRCh38
rs112197069 Chr.10: 94775218-94775218 on Build GRCh38
rs764078534 Chr.10: 94775211-94775211 on Build GRCh38
rs150045105 Chr.10: 94775079-94775079 on Build GRCh38
rs375159521 Chr.10: 94775181-94775181 on Build GRCh38
rs765198346 Chr.10: 94775185-94775186 on Build GRCh38
rs748575466 Chr.10: 94775069-94775069 on Build GRCh38
rs750025111 Chr.10: 94775148-94775148 on Build GRCh38
rs770401619 Chr.10: 94775074-94775074 on Build GRCh38
rs372985779 Chr.10: 94775194-94775194 on Build GRCh38
rs375151331 Chr.10: 94775055-94775055 on Build GRCh38
rs775372710 Chr.10: 94775195-94775195 on Build GRCh38
rs765389862 Chr.10: 94775220-94775220 on Build GRCh38
rs957285543 Chr.10: 94775209-94775209 on Build GRCh38
rs545642100 Chr.10: 94775206-94775206 on Build GRCh38
rs750969702 Chr.10: 94775228-94775228 on Build GRCh38
rs776518980 Chr.10: 94775111-94775111 on Build GRCh38
rs753618673 Chr.10: 94775134-94775134 on Build GRCh38
rs757977424 Chr.10: 94775222-94775222 on Build GRCh38
rs779668039 Chr.10: 94775060-94775060 on Build GRCh38
rs118203756 Chr.10: 94775160-94775160 on Build GRCh38
rs149072229 Chr.10: 94775130-94775130 on Build GRCh38

View Full Product Details