Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 37591275-37591548 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXA1
Entrez Gene ID
3169
Gene Name
forkhead box A1
Gene Aliases
HNF3A, TCF3A
UniGene
Hs.163484
Gene Chromosome Location
Chr.14: 37589552-37595120 on Build GRCh38
Transcripts
NM_004496.3 XM_017021246.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6666041 c.1293T>C p.S431S Chr.14: 37591491-37591491 on Build GRCh38
4685294 c.1302C>T p.P434P Chr.14: 37591482-37591482 on Build GRCh38
6983377 c.1415C>T p.S472F Chr.14: 37591369-37591369 on Build GRCh38
6666038 c.1363C>A p.L455M Chr.14: 37591421-37591421 on Build GRCh38
1734398 c.1268C>T p.A423V Chr.14: 37591516-37591516 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774156543 Chr.14: 37591511-37591511 on Build GRCh38
rs780579934 Chr.14: 37591481-37591481 on Build GRCh38
rs752035534 Chr.14: 37591464-37591464 on Build GRCh38
rs920525981 Chr.14: 37591478-37591478 on Build GRCh38
rs780538350 Chr.14: 37591329-37591329 on Build GRCh38
rs760849516 Chr.14: 37591417-37591417 on Build GRCh38
rs765380828 Chr.14: 37591449-37591449 on Build GRCh38
rs766907954 Chr.14: 37591463-37591463 on Build GRCh38
rs764112817 Chr.14: 37591433-37591433 on Build GRCh38
rs990956184 Chr.14: 37591383-37591383 on Build GRCh38
rs755462256 Chr.14: 37591474-37591474 on Build GRCh38
rs867072933 Chr.14: 37591443-37591443 on Build GRCh38
rs749443757 Chr.14: 37591520-37591520 on Build GRCh38
rs1022566815 Chr.14: 37591324-37591324 on Build GRCh38
rs141004703 Chr.14: 37591412-37591412 on Build GRCh38
rs897795973 Chr.14: 37591437-37591437 on Build GRCh38
rs909497251 Chr.14: 37591402-37591402 on Build GRCh38
rs373020885 Chr.14: 37591413-37591413 on Build GRCh38
rs750731146 Chr.14: 37591322-37591322 on Build GRCh38
rs771991531 Chr.14: 37591525-37591525 on Build GRCh38
rs746573702 Chr.14: 37591339-37591339 on Build GRCh38
rs367849941 Chr.14: 37591351-37591351 on Build GRCh38
rs776725888 Chr.14: 37591436-37591436 on Build GRCh38
rs768015890 Chr.14: 37591348-37591348 on Build GRCh38
rs181561875 Chr.14: 37591328-37591328 on Build GRCh38
rs33984772 Chr.14: 37591441-37591441 on Build GRCh38
rs150277709 Chr.14: 37591452-37591452 on Build GRCh38
rs773144107 Chr.14: 37591397-37591397 on Build GRCh38
rs780709606 Chr.14: 37591359-37591359 on Build GRCh38
rs916651137 Chr.14: 37591440-37591440 on Build GRCh38
rs776976444 Chr.14: 37591347-37591348 on Build GRCh38
rs375938424 Chr.14: 37591488-37591488 on Build GRCh38
rs770819038 Chr.14: 37591502-37591502 on Build GRCh38
rs749145227 Chr.14: 37591496-37591496 on Build GRCh38
rs758736377 Chr.14: 37591461-37591461 on Build GRCh38
rs530973207 Chr.14: 37591515-37591515 on Build GRCh38
rs561930261 Chr.14: 37591483-37591483 on Build GRCh38
rs749090160 Chr.14: 37591411-37591411 on Build GRCh38
rs923005164 Chr.14: 37591307-37591308 on Build GRCh38
rs747775950 Chr.14: 37591380-37591380 on Build GRCh38
rs2698 Chr.14: 37591307-37591307 on Build GRCh38
rs779274463 Chr.14: 37591316-37591316 on Build GRCh38
rs769510593 Chr.14: 37591384-37591384 on Build GRCh38

View Full Product Details