Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 89191292-89191545 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GBP4
Entrez Gene ID
115361
Gene Name
guanylate binding protein 4
Gene Aliases
Mpa2
UniGene
Hs.409925
Gene Chromosome Location
Chr.1: 89181148-89198950 on Build GRCh38
Transcripts
NM_052941.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4511010 c.854C>T p.S285F Chr.1: 89191323-89191323 on Build GRCh38
7369288 c.737G>T p.R246L Chr.1: 89191440-89191440 on Build GRCh38
7369290 c.713G>A p.C238Y Chr.1: 89191464-89191464 on Build GRCh38
2258927 c.704C>T p.P235L Chr.1: 89191473-89191473 on Build GRCh38
6669802 c.758G>A p.R253Q Chr.1: 89191419-89191419 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370597600 Chr.1: 89191419-89191419 on Build GRCh38
rs141089744 Chr.1: 89191441-89191441 on Build GRCh38
rs552803965 Chr.1: 89191421-89191421 on Build GRCh38
rs769820508 Chr.1: 89191340-89191340 on Build GRCh38
rs535437588 Chr.1: 89191409-89191409 on Build GRCh38
rs148433415 Chr.1: 89191326-89191326 on Build GRCh38
rs776679511 Chr.1: 89191464-89191464 on Build GRCh38
rs772221139 Chr.1: 89191364-89191364 on Build GRCh38
rs754754322 Chr.1: 89191447-89191447 on Build GRCh38
rs770497292 Chr.1: 89191319-89191319 on Build GRCh38
rs138876066 Chr.1: 89191446-89191446 on Build GRCh38
rs780772734 Chr.1: 89191515-89191515 on Build GRCh38
rs746618664 Chr.1: 89191315-89191315 on Build GRCh38
rs760715601 Chr.1: 89191456-89191456 on Build GRCh38
rs147368240 Chr.1: 89191502-89191502 on Build GRCh38
rs143553693 Chr.1: 89191518-89191518 on Build GRCh38
rs539360281 Chr.1: 89191382-89191382 on Build GRCh38
rs765622517 Chr.1: 89191399-89191399 on Build GRCh38
rs199790202 Chr.1: 89191433-89191433 on Build GRCh38
rs1002752639 Chr.1: 89191312-89191312 on Build GRCh38
rs753154560 Chr.1: 89191407-89191407 on Build GRCh38
rs765528610 Chr.1: 89191476-89191476 on Build GRCh38
rs777286949 Chr.1: 89191313-89191313 on Build GRCh38
rs140840134 Chr.1: 89191337-89191337 on Build GRCh38
rs138029071 Chr.1: 89191517-89191517 on Build GRCh38
rs756680047 Chr.1: 89191511-89191511 on Build GRCh38
rs745543939 Chr.1: 89191434-89191434 on Build GRCh38
rs147051288 Chr.1: 89191506-89191506 on Build GRCh38
rs766566761 Chr.1: 89191381-89191381 on Build GRCh38
rs769660126 Chr.1: 89191440-89191440 on Build GRCh38
rs753068951 Chr.1: 89191501-89191501 on Build GRCh38
rs760945523 Chr.1: 89191372-89191372 on Build GRCh38
rs773327494 Chr.1: 89191449-89191449 on Build GRCh38
rs146643676 Chr.1: 89191420-89191420 on Build GRCh38
rs374464600 Chr.1: 89191357-89191357 on Build GRCh38
rs974530190 Chr.1: 89191406-89191406 on Build GRCh38
rs375364072 Chr.1: 89191439-89191439 on Build GRCh38
rs759777047 Chr.1: 89191468-89191468 on Build GRCh38
rs763003067 Chr.1: 89191362-89191362 on Build GRCh38
rs771155723 Chr.1: 89191459-89191459 on Build GRCh38
rs372589831 Chr.1: 89191332-89191332 on Build GRCh38
rs751061154 Chr.1: 89191508-89191508 on Build GRCh38
rs201411513 Chr.1: 89191383-89191383 on Build GRCh38

View Full Product Details