Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 74567188-74567461 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNDP1
Entrez Gene ID
84735
Gene Name
carnosine dipeptidase 1
Gene Aliases
CN1, CPGL2, HsT2308
UniGene
Hs.400613
Gene Chromosome Location
Chr.18: 74534457-74585025 on Build GRCh38
Transcripts
NM_032649.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6448373 c.642G>A p.K214K Chr.18: 74567319-74567319 on Build GRCh38
4553767 c.600G>A p.E200E Chr.18: 74567277-74567277 on Build GRCh38
7361682 c.713C>T p.A238V Chr.18: 74567390-74567390 on Build GRCh38
7418802 c.614C>A p.A205D Chr.18: 74567291-74567291 on Build GRCh38
1303847 c.613G>A p.A205T Chr.18: 74567290-74567290 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775829747 Chr.18: 74567369-74567369 on Build GRCh38
rs779192235 Chr.18: 74567425-74567425 on Build GRCh38
rs369159038 Chr.18: 74567417-74567417 on Build GRCh38
rs750904832 Chr.18: 74567408-74567408 on Build GRCh38
rs773967009 Chr.18: 74567374-74567374 on Build GRCh38
rs762861558 Chr.18: 74567261-74567261 on Build GRCh38
rs144622039 Chr.18: 74567407-74567407 on Build GRCh38
rs761847077 Chr.18: 74567379-74567379 on Build GRCh38
rs753411332 Chr.18: 74567323-74567323 on Build GRCh38
rs202228025 Chr.18: 74567410-74567410 on Build GRCh38
rs777961866 Chr.18: 74567347-74567347 on Build GRCh38
rs778396198 Chr.18: 74567232-74567232 on Build GRCh38
rs773324396 Chr.18: 74567433-74567433 on Build GRCh38
rs376040268 Chr.18: 74567391-74567391 on Build GRCh38
rs909957008 Chr.18: 74567430-74567430 on Build GRCh38
rs779954861 Chr.18: 74567411-74567411 on Build GRCh38
rs138746215 Chr.18: 74567430-74567430 on Build GRCh38
rs749437461 Chr.18: 74567371-74567371 on Build GRCh38
rs747577303 Chr.18: 74567348-74567348 on Build GRCh38
rs747445325 Chr.18: 74567235-74567235 on Build GRCh38
rs373344061 Chr.18: 74567427-74567427 on Build GRCh38
rs140836083 Chr.18: 74567390-74567390 on Build GRCh38
rs1036811068 Chr.18: 74567338-74567338 on Build GRCh38
rs760311524 Chr.18: 74567388-74567388 on Build GRCh38
rs769932083 Chr.18: 74567256-74567256 on Build GRCh38
rs757706839 Chr.18: 74567350-74567350 on Build GRCh38
rs767157567 Chr.18: 74567283-74567283 on Build GRCh38
rs768702340 Chr.18: 74567421-74567421 on Build GRCh38
rs369390962 Chr.18: 74567221-74567221 on Build GRCh38
rs773271265 Chr.18: 74567384-74567384 on Build GRCh38
rs779426008 Chr.18: 74567211-74567211 on Build GRCh38
rs775568626 Chr.18: 74567257-74567257 on Build GRCh38
rs755680847 Chr.18: 74567292-74567292 on Build GRCh38
rs767437203 Chr.18: 74567217-74567217 on Build GRCh38
rs763773745 Chr.18: 74567268-74567268 on Build GRCh38
rs1004120545 Chr.18: 74567322-74567322 on Build GRCh38
rs771034260 Chr.18: 74567236-74567236 on Build GRCh38
rs749886572 Chr.18: 74567290-74567290 on Build GRCh38
rs200332997 Chr.18: 74567324-74567324 on Build GRCh38
rs373184921 Chr.18: 74567278-74567278 on Build GRCh38
rs746099553 Chr.18: 74567250-74567250 on Build GRCh38
rs758678609 Chr.18: 74567227-74567227 on Build GRCh38
rs144874700 Chr.18: 74567402-74567402 on Build GRCh38
rs1050930409 Chr.18: 74567228-74567228 on Build GRCh38
rs12960862 Chr.18: 74567400-74567400 on Build GRCh38
rs764536721 Chr.18: 74567395-74567395 on Build GRCh38
rs568823832 Chr.18: 74567301-74567301 on Build GRCh38
rs776646526 Chr.18: 74567246-74567246 on Build GRCh38
rs151304098 Chr.18: 74567364-74567364 on Build GRCh38
rs781249995 Chr.18: 74567361-74567361 on Build GRCh38

View Full Product Details