Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 65320801-65321026 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC42EP2
Entrez Gene ID
10435
Gene Name
CDC42 effector protein 2
Gene Aliases
BORG1, CEP2
UniGene
Hs.343380
Gene Chromosome Location
Chr.11: 65314729-65322429 on Build GRCh38
Transcripts
NM_006779.3 XM_017017094.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1355923 c.98C>T p.P33L Chr.11: 65320996-65320996 on Build GRCh38
5341673 c.31C>T p.R11C Chr.11: 65320929-65320929 on Build GRCh38
6624424 c.80C>T p.S27L Chr.11: 65320978-65320978 on Build GRCh38
1746464 c.127A>G p.I43V Chr.11: 65321025-65321025 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs565029628 Chr.11: 65320939-65320939 on Build GRCh38
rs573773203 Chr.11: 65320821-65320821 on Build GRCh38
rs769508038 Chr.11: 65320871-65320871 on Build GRCh38
rs143861143 Chr.11: 65320930-65320930 on Build GRCh38
rs367722501 Chr.11: 65320859-65320859 on Build GRCh38
rs756430782 Chr.11: 65320916-65320916 on Build GRCh38
rs372684048 Chr.11: 65320963-65320963 on Build GRCh38
rs761268858 Chr.11: 65320983-65320983 on Build GRCh38
rs1021807508 Chr.11: 65320996-65320996 on Build GRCh38
rs775435247 Chr.11: 65320880-65320880 on Build GRCh38
rs760301829 Chr.11: 65320997-65320997 on Build GRCh38
rs150488159 Chr.11: 65320948-65320948 on Build GRCh38
rs767015957 Chr.11: 65320991-65320991 on Build GRCh38
rs370024868 Chr.11: 65320921-65320921 on Build GRCh38
rs748132612 Chr.11: 65320929-65320929 on Build GRCh38
rs772465319 Chr.11: 65320864-65320864 on Build GRCh38
rs141215100 Chr.11: 65320979-65320979 on Build GRCh38
rs767105914 Chr.11: 65320909-65320909 on Build GRCh38
rs763649496 Chr.11: 65320890-65320890 on Build GRCh38
rs961142370 Chr.11: 65320840-65320840 on Build GRCh38
rs372621101 Chr.11: 65320884-65320884 on Build GRCh38
rs766759614 Chr.11: 65320999-65320999 on Build GRCh38
rs148229585 Chr.11: 65320962-65320962 on Build GRCh38
rs542733338 Chr.11: 65320826-65320826 on Build GRCh38
rs1047015948 Chr.11: 65320938-65320938 on Build GRCh38
rs768625934 Chr.11: 65320978-65320978 on Build GRCh38
rs375958650 Chr.11: 65320887-65320887 on Build GRCh38
rs754356149 Chr.11: 65320928-65320928 on Build GRCh38
rs201674051 Chr.11: 65320995-65320995 on Build GRCh38
rs773920762 Chr.11: 65320895-65320895 on Build GRCh38
rs531413970 Chr.11: 65320865-65320865 on Build GRCh38
rs759327919 Chr.11: 65320867-65320867 on Build GRCh38
rs545241921 Chr.11: 65320915-65320915 on Build GRCh38
rs377166460 Chr.11: 65320966-65320966 on Build GRCh38
rs374784454 Chr.11: 65320858-65320858 on Build GRCh38
rs761429865 Chr.11: 65320903-65320903 on Build GRCh38
rs749247580 Chr.11: 65320970-65320970 on Build GRCh38
rs562667731 Chr.11: 65320831-65320831 on Build GRCh38
rs779030033 Chr.11: 65320863-65320863 on Build GRCh38
rs778102151 Chr.11: 65320957-65320957 on Build GRCh38
rs977351301 Chr.11: 65320842-65320842 on Build GRCh38

View Full Product Details