Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 71712622-71712895 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C10orf105
Entrez Gene ID
414152
Gene Name
chromosome 10 open reading frame 105
Gene Aliases
-
UniGene
Hs.568788
Gene Chromosome Location
Chr.10: 71711701-71737824 on Build GRCh38
Transcripts
NM_001164375.2 NM_001168390.1 XM_011539808.2 XM_011539809.2 NM_001171930.1 NM_022124.5 XM_006717940.3 XM_006717942.3 XM_011540039.2 XM_011540042.2 XM_011540043.2 XM_011540044.2 XM_011540045.2 XM_011540046.2 XM_011540047.2 XM_011540048.2 XM_011540049.2 XM_011540051.2 XM_017016499.1 XM_017016500.1 XM_017016501.1 XM_017016502.1

Gene Symbol
CDH23
Entrez Gene ID
64072
Gene Name
cadherin-related 23
Gene Aliases
CDHR23, USH1D
UniGene
Hs.656032
Gene Chromosome Location
Chr.10: 71396934-71815947 on Build GRCh38
Transcripts
NM_001164375.2 NM_001168390.1 XM_011539808.2 XM_011539809.2 NM_001171930.1 NM_022124.5 XM_006717940.3 XM_006717942.3 XM_011540039.2 XM_011540042.2 XM_011540043.2 XM_011540044.2 XM_011540045.2 XM_011540046.2 XM_011540047.2 XM_011540048.2 XM_011540049.2 XM_011540051.2 XM_017016499.1 XM_017016500.1 XM_017016501.1 XM_017016502.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1128007 c.3291C>A p.V1097V Chr.10: 71712720-71712720 on Build GRCh38
6313829 c.1920G>T p.Q640H Chr.10: 71712813-71712813 on Build GRCh38
7092379 c.3378C>A p.I1126I Chr.10: 71712807-71712807 on Build GRCh38
4180338 c.1846C>T p.R616W Chr.10: 71712739-71712739 on Build GRCh38
5561406 c.3349C>A p.P1117T Chr.10: 71712778-71712778 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs34236496 Chr.10: 71712692-71712693 on Build GRCh38
rs950530239 Chr.10: 71712872-71712872 on Build GRCh38
rs777616794 Chr.10: 71712870-71712870 on Build GRCh38
rs767680481 Chr.10: 71712669-71712669 on Build GRCh38
rs727504705 Chr.10: 71712770-71712770 on Build GRCh38
rs760800475 Chr.10: 71712739-71712739 on Build GRCh38
rs773479197 Chr.10: 71712721-71712721 on Build GRCh38
rs377126049 Chr.10: 71712711-71712711 on Build GRCh38
rs879069808 Chr.10: 71712850-71712850 on Build GRCh38
rs200542052 Chr.10: 71712805-71712805 on Build GRCh38
rs377676159 Chr.10: 71712742-71712742 on Build GRCh38
rs202101019 Chr.10: 71712674-71712674 on Build GRCh38
rs397517321 Chr.10: 71712775-71712775 on Build GRCh38
rs539034178 Chr.10: 71712824-71712824 on Build GRCh38
rs769524849 Chr.10: 71712814-71712814 on Build GRCh38
rs752839402 Chr.10: 71712783-71712783 on Build GRCh38
rs765385038 Chr.10: 71712866-71712866 on Build GRCh38
rs776650486 Chr.10: 71712863-71712863 on Build GRCh38
rs569390669 Chr.10: 71712860-71712860 on Build GRCh38
rs371962929 Chr.10: 71712774-71712774 on Build GRCh38
rs79805606 Chr.10: 71712693-71712693 on Build GRCh38
rs766261756 Chr.10: 71712675-71712675 on Build GRCh38
rs397517322 Chr.10: 71712781-71712781 on Build GRCh38
rs866459900 Chr.10: 71712849-71712849 on Build GRCh38
rs754122678 Chr.10: 71712695-71712695 on Build GRCh38
rs756165682 Chr.10: 71712692-71712692 on Build GRCh38
rs761607917 Chr.10: 71712718-71712718 on Build GRCh38
rs368487578 Chr.10: 71712712-71712712 on Build GRCh38
rs372378807 Chr.10: 71712710-71712710 on Build GRCh38
rs779296930 Chr.10: 71712705-71712705 on Build GRCh38
rs757508152 Chr.10: 71712772-71712772 on Build GRCh38
rs949198537 Chr.10: 71712843-71712843 on Build GRCh38
rs941805977 Chr.10: 71712661-71712661 on Build GRCh38
rs41281310 Chr.10: 71712737-71712737 on Build GRCh38
rs762681991 Chr.10: 71712729-71712729 on Build GRCh38
rs113714164 Chr.10: 71712751-71712751 on Build GRCh38
rs876657753 Chr.10: 71712743-71712743 on Build GRCh38
rs1046140667 Chr.10: 71712846-71712846 on Build GRCh38
rs934025237 Chr.10: 71712730-71712730 on Build GRCh38
rs759437235 Chr.10: 71712859-71712859 on Build GRCh38
rs866435331 Chr.10: 71712685-71712685 on Build GRCh38
rs77821631 Chr.10: 71712808-71712808 on Build GRCh38
rs199510686 Chr.10: 71712745-71712745 on Build GRCh38
rs200632520 Chr.10: 71712706-71712706 on Build GRCh38
rs776427467 Chr.10: 71712658-71712658 on Build GRCh38
rs773015477 Chr.10: 71712821-71712821 on Build GRCh38
rs187975106 Chr.10: 71712825-71712825 on Build GRCh38
rs752776373 Chr.10: 71712686-71712686 on Build GRCh38
rs766285590 Chr.10: 71712731-71712731 on Build GRCh38
rs773105137 Chr.10: 71712727-71712727 on Build GRCh38
rs774329375 Chr.10: 71712836-71712836 on Build GRCh38
rs74147036 Chr.10: 71712871-71712871 on Build GRCh38
rs35865328 Chr.10: 71712669-71712670 on Build GRCh38
rs750364937 Chr.10: 71712672-71712672 on Build GRCh38
rs769682743 Chr.10: 71712720-71712720 on Build GRCh38
rs562859768 Chr.10: 71712676-71712676 on Build GRCh38
rs34242194 Chr.10: 71712739-71712740 on Build GRCh38
rs369002480 Chr.10: 71712749-71712749 on Build GRCh38
rs368147248 Chr.10: 71712698-71712698 on Build GRCh38
rs529191251 Chr.10: 71712797-71712797 on Build GRCh38
rs768305516 Chr.10: 71712641-71712641 on Build GRCh38
rs562052236 Chr.10: 71712796-71712796 on Build GRCh38

View Full Product Details