Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 137111662-137111897 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPP7
Entrez Gene ID
29952
Gene Name
dipeptidyl peptidase 7
Gene Aliases
DPP2, DPPII, QPP
UniGene
Hs.37916
Gene Chromosome Location
Chr.9: 137110540-137117980 on Build GRCh38
Transcripts
NM_013379.2 XM_005266075.3 XM_006717083.3 XM_011518599.2 XM_011518600.1 XM_017014651.1 XM_017014652.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7413778 c.1194C>A p.S398R Chr.9: 137111886-137111886 on Build GRCh38
292638 c.1205_1206insG p.D403fs*1 Chr.9: 137111874-137111875 on Build GRCh38
7417193 c.1200G>T p.W400C Chr.9: 137111880-137111880 on Build GRCh38
5156044 c.1212_1213delCA p.R405fs*55 Chr.9: 137111749-137111750 on Build GRCh38
3763812 c.1212C>T p.L404L Chr.9: 137111750-137111750 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749013361 Chr.9: 137111842-137111842 on Build GRCh38
rs754291597 Chr.9: 137111726-137111726 on Build GRCh38
rs759971920 Chr.9: 137111728-137111728 on Build GRCh38
rs763568074 Chr.9: 137111799-137111799 on Build GRCh38
rs759768087 Chr.9: 137111788-137111788 on Build GRCh38
rs765916104 Chr.9: 137111874-137111874 on Build GRCh38
rs13631 Chr.9: 137111750-137111750 on Build GRCh38
rs571126074 Chr.9: 137111850-137111850 on Build GRCh38
rs769791444 Chr.9: 137111695-137111695 on Build GRCh38
rs777369315 Chr.9: 137111734-137111734 on Build GRCh38
rs760636809 Chr.9: 137111777-137111777 on Build GRCh38
rs767042968 Chr.9: 137111868-137111868 on Build GRCh38
rs557735369 Chr.9: 137111846-137111846 on Build GRCh38
rs775395900 Chr.9: 137111697-137111697 on Build GRCh38
rs755483176 Chr.9: 137111856-137111856 on Build GRCh38
rs199630935 Chr.9: 137111852-137111852 on Build GRCh38
rs745632439 Chr.9: 137111690-137111690 on Build GRCh38
rs371237783 Chr.9: 137111830-137111830 on Build GRCh38
rs753254520 Chr.9: 137111731-137111731 on Build GRCh38
rs989918118 Chr.9: 137111797-137111797 on Build GRCh38
rs753858105 Chr.9: 137111694-137111694 on Build GRCh38
rs562251063 Chr.9: 137111855-137111856 on Build GRCh38
rs753728877 Chr.9: 137111879-137111879 on Build GRCh38
rs111299368 Chr.9: 137111687-137111687 on Build GRCh38
rs764477908 Chr.9: 137111806-137111806 on Build GRCh38
rs1006268057 Chr.9: 137111693-137111693 on Build GRCh38
rs765731409 Chr.9: 137111794-137111794 on Build GRCh38
rs749278220 Chr.9: 137111773-137111773 on Build GRCh38
rs948591912 Chr.9: 137111815-137111815 on Build GRCh38
rs111502395 Chr.9: 137111757-137111757 on Build GRCh38
rs202238625 Chr.9: 137111721-137111721 on Build GRCh38
rs775967056 Chr.9: 137111857-137111857 on Build GRCh38
rs773386646 Chr.9: 137111774-137111774 on Build GRCh38
rs372692652 Chr.9: 137111780-137111780 on Build GRCh38
rs758794974 Chr.9: 137111732-137111732 on Build GRCh38
rs143691643 Chr.9: 137111701-137111701 on Build GRCh38
rs763223840 Chr.9: 137111858-137111858 on Build GRCh38
rs556621023 Chr.9: 137111859-137111859 on Build GRCh38
rs369202485 Chr.9: 137111685-137111685 on Build GRCh38
rs769591813 Chr.9: 137111761-137111761 on Build GRCh38
rs779921211 Chr.9: 137111839-137111839 on Build GRCh38
rs746710335 Chr.9: 137111680-137111680 on Build GRCh38
rs779755124 Chr.9: 137111770-137111770 on Build GRCh38
rs749885398 Chr.9: 137111872-137111872 on Build GRCh38
rs111838018 Chr.9: 137111829-137111829 on Build GRCh38
rs373618825 Chr.9: 137111847-137111847 on Build GRCh38
rs934276274 Chr.9: 137111754-137111754 on Build GRCh38
rs371372857 Chr.9: 137111682-137111682 on Build GRCh38
rs756944196 Chr.9: 137111743-137111743 on Build GRCh38
rs914298658 Chr.9: 137111808-137111808 on Build GRCh38
rs780818415 Chr.9: 137111744-137111744 on Build GRCh38
rs755722017 Chr.9: 137111836-137111836 on Build GRCh38
rs201900304 Chr.9: 137111705-137111705 on Build GRCh38
rs748560644 Chr.9: 137111874-137111875 on Build GRCh38
rs762242625 Chr.9: 137111861-137111861 on Build GRCh38
rs750078178 Chr.9: 137111832-137111832 on Build GRCh38
rs752959422 Chr.9: 137111796-137111796 on Build GRCh38
rs774736108 Chr.9: 137111860-137111860 on Build GRCh38
rs377463433 Chr.9: 137111782-137111782 on Build GRCh38
rs111751757 Chr.9: 137111756-137111756 on Build GRCh38
rs767638501 Chr.9: 137111714-137111714 on Build GRCh38
rs755812109 Chr.9: 137111873-137111873 on Build GRCh38
rs759863578 Chr.9: 137111856-137111856 on Build GRCh38
rs776977163 Chr.9: 137111781-137111781 on Build GRCh38
rs773124388 Chr.9: 137111719-137111719 on Build GRCh38
rs200432704 Chr.9: 137111720-137111720 on Build GRCh38

View Full Product Details