Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 44963638-44963903 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM179B
Entrez Gene ID
23116
Gene Name
family with sequence similarity 179 member B
Gene Aliases
KIAA0423
UniGene
Hs.371078
Gene Chromosome Location
Chr.14: 44962190-45074431 on Build GRCh38
Transcripts
NM_001308120.1 NM_015091.3 NR_131765.1 XM_011536571.1 XM_017021098.1 XM_017021099.1 XM_017021100.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6659054 c.1367T>C p.M456T Chr.14: 44963788-44963788 on Build GRCh38
3706282 c.1436T>A p.L479H Chr.14: 44963857-44963857 on Build GRCh38
4524742 c.1290G>A p.Q430Q Chr.14: 44963711-44963711 on Build GRCh38
6521476 c.1380C>A p.L460L Chr.14: 44963801-44963801 on Build GRCh38
1206212 c.1238G>T p.G413V Chr.14: 44963659-44963659 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs972018580 Chr.14: 44963820-44963820 on Build GRCh38
rs532764935 Chr.14: 44963835-44963835 on Build GRCh38
rs764845901 Chr.14: 44963815-44963815 on Build GRCh38
rs188299008 Chr.14: 44963796-44963796 on Build GRCh38
rs771100518 Chr.14: 44963787-44963787 on Build GRCh38
rs371633827 Chr.14: 44963675-44963675 on Build GRCh38
rs779869465 Chr.14: 44963752-44963752 on Build GRCh38
rs774719524 Chr.14: 44963685-44963685 on Build GRCh38
rs748317794 Chr.14: 44963756-44963756 on Build GRCh38
rs760337851 Chr.14: 44963804-44963804 on Build GRCh38
rs969619145 Chr.14: 44963801-44963801 on Build GRCh38
rs149557242 Chr.14: 44963722-44963722 on Build GRCh38
rs779072932 Chr.14: 44963863-44963863 on Build GRCh38
rs759804447 Chr.14: 44963687-44963687 on Build GRCh38
rs968178220 Chr.14: 44963708-44963708 on Build GRCh38
rs758352445 Chr.14: 44963822-44963822 on Build GRCh38
rs958989238 Chr.14: 44963772-44963772 on Build GRCh38
rs200300639 Chr.14: 44963774-44963774 on Build GRCh38
rs776031134 Chr.14: 44963800-44963800 on Build GRCh38
rs757164099 Chr.14: 44963713-44963713 on Build GRCh38
rs746338277 Chr.14: 44963878-44963878 on Build GRCh38
rs542685317 Chr.14: 44963769-44963769 on Build GRCh38
rs766800061 Chr.14: 44963684-44963684 on Build GRCh38
rs752376719 Chr.14: 44963692-44963692 on Build GRCh38
rs749340329 Chr.14: 44963779-44963779 on Build GRCh38
rs139341319 Chr.14: 44963859-44963859 on Build GRCh38
rs769840369 Chr.14: 44963673-44963673 on Build GRCh38
rs1020732592 Chr.14: 44963696-44963696 on Build GRCh38
rs148321239 Chr.14: 44963788-44963788 on Build GRCh38
rs763855732 Chr.14: 44963806-44963806 on Build GRCh38
rs763473895 Chr.14: 44963681-44963681 on Build GRCh38
rs1000380654 Chr.14: 44963768-44963768 on Build GRCh38
rs749589830 Chr.14: 44963856-44963856 on Build GRCh38
rs751356950 Chr.14: 44963847-44963847 on Build GRCh38
rs962418201 Chr.14: 44963714-44963714 on Build GRCh38
rs370658518 Chr.14: 44963689-44963689 on Build GRCh38
rs765929204 Chr.14: 44963706-44963706 on Build GRCh38
rs758160687 Chr.14: 44963746-44963746 on Build GRCh38
rs760128686 Chr.14: 44963795-44963795 on Build GRCh38
rs565435000 Chr.14: 44963816-44963816 on Build GRCh38
rs3825629 Chr.14: 44963667-44963667 on Build GRCh38
rs756270301 Chr.14: 44963854-44963854 on Build GRCh38
rs575568809 Chr.14: 44963704-44963704 on Build GRCh38
rs755855059 Chr.14: 44963697-44963697 on Build GRCh38
rs367642428 Chr.14: 44963730-44963730 on Build GRCh38
rs778706373 Chr.14: 44963726-44963726 on Build GRCh38
rs981006167 Chr.14: 44963848-44963848 on Build GRCh38
rs761227117 Chr.14: 44963810-44963810 on Build GRCh38

View Full Product Details