Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 109553804-109554033 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EBAG9
Entrez Gene ID
9166
Gene Name
estrogen receptor binding site associated, antigen, 9
Gene Aliases
EB9, PDAF
UniGene
Hs.409368
Gene Chromosome Location
Chr.8: 109539700-109565996 on Build GRCh38
Transcripts
NM_001278938.1 NM_004215.4 NM_198120.2 XM_017013960.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6222328 c.97C>T p.R33W Chr.8: 109553878-109553878 on Build GRCh38
5367743 c.162+5_162+6insT p.? Chr.8: 109553948-109553949 on Build GRCh38
6499211 c.139G>C p.D47H Chr.8: 109553920-109553920 on Build GRCh38
6651116 c.162+6delT p.? Chr.8: 109553949-109553949 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs756966123 Chr.8: 109553838-109553838 on Build GRCh38
rs188777792 Chr.8: 109553993-109553993 on Build GRCh38
rs199898599 Chr.8: 109553969-109553969 on Build GRCh38
rs767199434 Chr.8: 109553968-109553968 on Build GRCh38
rs931430890 Chr.8: 109553839-109553839 on Build GRCh38
rs139788553 Chr.8: 109553932-109553932 on Build GRCh38
rs773684977 Chr.8: 109553914-109553914 on Build GRCh38
rs376074688 Chr.8: 109553844-109553844 on Build GRCh38
rs761545199 Chr.8: 109553957-109553957 on Build GRCh38
rs747078563 Chr.8: 109553965-109553969 on Build GRCh38
rs755168596 Chr.8: 109553879-109553879 on Build GRCh38
rs750273931 Chr.8: 109553864-109553864 on Build GRCh38
rs373590104 Chr.8: 109553964-109553965 on Build GRCh38
rs199580138 Chr.8: 109553878-109553878 on Build GRCh38
rs143877969 Chr.8: 109553895-109553895 on Build GRCh38
rs753687874 Chr.8: 109553948-109553949 on Build GRCh38
rs1004016994 Chr.8: 109554000-109554000 on Build GRCh38
rs756130426 Chr.8: 109553874-109553874 on Build GRCh38
rs779670710 Chr.8: 109553948-109553956 on Build GRCh38
rs371503766 Chr.8: 109553964-109553964 on Build GRCh38
rs2703377 Chr.8: 109553955-109553955 on Build GRCh38
rs373171389 Chr.8: 109553928-109553928 on Build GRCh38
rs748399467 Chr.8: 109553899-109553899 on Build GRCh38
rs763890372 Chr.8: 109553831-109553831 on Build GRCh38
rs536381456 Chr.8: 109553949-109553949 on Build GRCh38
rs772258835 Chr.8: 109553911-109553911 on Build GRCh38
rs1047773753 Chr.8: 109553974-109553974 on Build GRCh38
rs780099814 Chr.8: 109553876-109553876 on Build GRCh38
rs73700652 Chr.8: 109553948-109553948 on Build GRCh38
rs755970929 Chr.8: 109553975-109553975 on Build GRCh38
rs771509590 Chr.8: 109553931-109553931 on Build GRCh38
rs758106672 Chr.8: 109553900-109553900 on Build GRCh38
rs536248169 Chr.8: 109553947-109553947 on Build GRCh38
rs752323333 Chr.8: 109553830-109553830 on Build GRCh38
rs914794078 Chr.8: 109553934-109553934 on Build GRCh38
rs1044667077 Chr.8: 109553949-109553949 on Build GRCh38

View Full Product Details