Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 71283744-71284017 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CMTR2
Entrez Gene ID
55783
Gene Name
cap methyltransferase 2
Gene Aliases
AFT, FTSJD1, HMTr2, MTr2
UniGene
Hs.72782
Gene Chromosome Location
Chr.16: 71282300-71290178 on Build GRCh38
Transcripts
NM_001099642.1 NM_001324374.1 NM_001324377.1 NM_001324378.1 NM_001324379.1 NM_018348.5 XM_011523228.1 XM_011523229.1 XM_017023444.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4062516 c.2099G>A p.R700Q Chr.16: 71283822-71283822 on Build GRCh38
4569820 c.2070T>C p.G690G Chr.16: 71283851-71283851 on Build GRCh38
6667665 c.1977C>A p.I659I Chr.16: 71283944-71283944 on Build GRCh38
704255 c.2050G>A p.A684T Chr.16: 71283871-71283871 on Build GRCh38
3275787 c.2100A>G p.R700R Chr.16: 71283821-71283821 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753886091 Chr.16: 71283946-71283947 on Build GRCh38
rs776452784 Chr.16: 71283812-71283812 on Build GRCh38
rs770555786 Chr.16: 71283811-71283811 on Build GRCh38
rs761026424 Chr.16: 71283838-71283838 on Build GRCh38
rs777295271 Chr.16: 71283797-71283797 on Build GRCh38
rs779872886 Chr.16: 71283918-71283918 on Build GRCh38
rs769746395 Chr.16: 71283820-71283820 on Build GRCh38
rs143985943 Chr.16: 71283860-71283860 on Build GRCh38
rs757885960 Chr.16: 71283775-71283775 on Build GRCh38
rs756954551 Chr.16: 71283902-71283902 on Build GRCh38
rs555474743 Chr.16: 71283905-71283905 on Build GRCh38
rs147595116 Chr.16: 71283952-71283952 on Build GRCh38
rs761807488 Chr.16: 71283822-71283822 on Build GRCh38
rs202046539 Chr.16: 71283865-71283865 on Build GRCh38
rs868416312 Chr.16: 71283948-71283950 on Build GRCh38
rs752256267 Chr.16: 71283794-71283796 on Build GRCh38
rs766674468 Chr.16: 71283847-71283847 on Build GRCh38
rs745678746 Chr.16: 71283975-71283976 on Build GRCh38
rs769516770 Chr.16: 71283913-71283913 on Build GRCh38
rs368654966 Chr.16: 71283823-71283823 on Build GRCh38
rs757859041 Chr.16: 71283881-71283881 on Build GRCh38
rs149893216 Chr.16: 71283908-71283908 on Build GRCh38
rs779536396 Chr.16: 71283769-71283769 on Build GRCh38
rs777398960 Chr.16: 71283886-71283886 on Build GRCh38
rs773154873 Chr.16: 71283834-71283834 on Build GRCh38
rs746614281 Chr.16: 71283800-71283800 on Build GRCh38
rs946136809 Chr.16: 71283982-71283982 on Build GRCh38
rs767137178 Chr.16: 71283912-71283912 on Build GRCh38
rs751104143 Chr.16: 71283959-71283959 on Build GRCh38
rs201532811 Chr.16: 71283947-71283947 on Build GRCh38
rs200001480 Chr.16: 71283950-71283950 on Build GRCh38
rs763455766 Chr.16: 71283938-71283939 on Build GRCh38
rs776948589 Chr.16: 71283946-71283946 on Build GRCh38
rs1016680975 Chr.16: 71283782-71283782 on Build GRCh38
rs142245314 Chr.16: 71283818-71283818 on Build GRCh38
rs775457941 Chr.16: 71283821-71283821 on Build GRCh38
rs760812171 Chr.16: 71283938-71283938 on Build GRCh38
rs749308887 Chr.16: 71283931-71283931 on Build GRCh38
rs771297237 Chr.16: 71283940-71283940 on Build GRCh38
rs752988391 Chr.16: 71283949-71283949 on Build GRCh38
rs768475701 Chr.16: 71283933-71283933 on Build GRCh38
rs773378729 Chr.16: 71283937-71283937 on Build GRCh38
rs757257549 Chr.16: 71283950-71283951 on Build GRCh38
rs962756005 Chr.16: 71283810-71283811 on Build GRCh38
rs151193694 Chr.16: 71283866-71283866 on Build GRCh38
rs148258352 Chr.16: 71283871-71283871 on Build GRCh38
rs755655082 Chr.16: 71283925-71283927 on Build GRCh38
rs751026064 Chr.16: 71283898-71283898 on Build GRCh38
rs914688615 Chr.16: 71283974-71283974 on Build GRCh38
rs763897670 Chr.16: 71283883-71283883 on Build GRCh38
rs569680257 Chr.16: 71283773-71283773 on Build GRCh38
rs898744882 Chr.16: 71283872-71283872 on Build GRCh38
rs756717787 Chr.16: 71283991-71283991 on Build GRCh38

View Full Product Details