Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 34500671-34500928 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAI1
Entrez Gene ID
27019
Gene Name
dynein axonemal intermediate chain 1
Gene Aliases
CILD1, DIC1, ICS1, PCD
UniGene
Hs.112667
Gene Chromosome Location
Chr.9: 34458752-34520989 on Build GRCh38
Transcripts
NM_001281428.1 NM_012144.3 XM_006716758.3 XM_011517846.1 XM_011517847.2 XM_011517848.1 XM_011517849.1 XM_011517850.2 XM_017014625.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
334936 c.992A>T p.K331M Chr.9: 34500812-34500812 on Build GRCh38
5435751 c.1004T>A p.V335D Chr.9: 34500824-34500824 on Build GRCh38
455868 c.996C>A p.R332R Chr.9: 34500816-34500816 on Build GRCh38
6115474 c.999G>T p.L333L Chr.9: 34500819-34500819 on Build GRCh38
4821135 c.1005C>G p.V335V Chr.9: 34500825-34500825 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754233819 Chr.9: 34500822-34500822 on Build GRCh38
rs992279032 Chr.9: 34500850-34500850 on Build GRCh38
rs914112487 Chr.9: 34500871-34500871 on Build GRCh38
rs764464046 Chr.9: 34500843-34500843 on Build GRCh38
rs749687319 Chr.9: 34500712-34500712 on Build GRCh38
rs182015909 Chr.9: 34500814-34500814 on Build GRCh38
rs766941804 Chr.9: 34500815-34500815 on Build GRCh38
rs745775853 Chr.9: 34500868-34500868 on Build GRCh38
rs776390032 Chr.9: 34500765-34500765 on Build GRCh38
rs778140393 Chr.9: 34500714-34500717 on Build GRCh38
rs755259202 Chr.9: 34500714-34500714 on Build GRCh38
rs267602217 Chr.9: 34500830-34500830 on Build GRCh38
rs10814116 Chr.9: 34500881-34500881 on Build GRCh38
rs747475785 Chr.9: 34500854-34500862 on Build GRCh38
rs777922183 Chr.9: 34500836-34500836 on Build GRCh38
rs747121305 Chr.9: 34500844-34500844 on Build GRCh38
rs527254975 Chr.9: 34500723-34500723 on Build GRCh38
rs201547687 Chr.9: 34500800-34500800 on Build GRCh38
rs769701411 Chr.9: 34500872-34500872 on Build GRCh38
rs45600639 Chr.9: 34500709-34500709 on Build GRCh38
rs11793196 Chr.9: 34500823-34500823 on Build GRCh38
rs377594689 Chr.9: 34500744-34500744 on Build GRCh38
rs984678526 Chr.9: 34500895-34500895 on Build GRCh38
rs761392145 Chr.9: 34500887-34500887 on Build GRCh38
rs373836235 Chr.9: 34500707-34500707 on Build GRCh38
rs757425174 Chr.9: 34500906-34500906 on Build GRCh38
rs570977988 Chr.9: 34500886-34500886 on Build GRCh38
rs200720794 Chr.9: 34500846-34500846 on Build GRCh38
rs746996706 Chr.9: 34500741-34500741 on Build GRCh38
rs775397706 Chr.9: 34500876-34500876 on Build GRCh38
rs780334791 Chr.9: 34500711-34500711 on Build GRCh38
rs762694599 Chr.9: 34500783-34500783 on Build GRCh38
rs763687728 Chr.9: 34500699-34500699 on Build GRCh38
rs878854968 Chr.9: 34500732-34500732 on Build GRCh38
rs775336300 Chr.9: 34500782-34500782 on Build GRCh38
rs762745975 Chr.9: 34500878-34500878 on Build GRCh38
rs201653559 Chr.9: 34500734-34500734 on Build GRCh38
rs757991463 Chr.9: 34500696-34500696 on Build GRCh38
rs773243879 Chr.9: 34500739-34500739 on Build GRCh38
rs764879929 Chr.9: 34500781-34500781 on Build GRCh38
rs772207177 Chr.9: 34500738-34500738 on Build GRCh38
rs935656322 Chr.9: 34500907-34500907 on Build GRCh38
rs758595604 Chr.9: 34500831-34500831 on Build GRCh38
rs982925157 Chr.9: 34500808-34500808 on Build GRCh38
rs535718826 Chr.9: 34500708-34500708 on Build GRCh38
rs759377098 Chr.9: 34500772-34500772 on Build GRCh38
rs16931555 Chr.9: 34500798-34500798 on Build GRCh38
rs781116065 Chr.9: 34500845-34500845 on Build GRCh38
rs375083974 Chr.9: 34500875-34500875 on Build GRCh38
rs959992832 Chr.9: 34500857-34500857 on Build GRCh38

View Full Product Details