Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 161749891-161750152 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUSP12
Entrez Gene ID
11266
Gene Name
dual specificity phosphatase 12
Gene Aliases
DUSP1, YVH1
UniGene
Hs.416216
Gene Chromosome Location
Chr.1: 161749692-161757164 on Build GRCh38
Transcripts
NM_007240.2 XM_005244862.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5968044 c.211G>T p.G71W Chr.1: 161750012-161750012 on Build GRCh38
6650310 c.120G>T p.A40A Chr.1: 161749921-161749921 on Build GRCh38
1335705 c.132G>A p.E44E Chr.1: 161749933-161749933 on Build GRCh38
1319802 c.93G>C p.Q31H Chr.1: 161749894-161749894 on Build GRCh38
6835331 c.194G>A p.S65N Chr.1: 161749995-161749995 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs146495672 Chr.1: 161749946-161749946 on Build GRCh38
rs749380595 Chr.1: 161750068-161750068 on Build GRCh38
rs776052385 Chr.1: 161750114-161750114 on Build GRCh38
rs764136089 Chr.1: 161750038-161750038 on Build GRCh38
rs149144814 Chr.1: 161749985-161749985 on Build GRCh38
rs935771319 Chr.1: 161750111-161750111 on Build GRCh38
rs760308393 Chr.1: 161750105-161750105 on Build GRCh38
rs528195907 Chr.1: 161750025-161750025 on Build GRCh38
rs774257991 Chr.1: 161749924-161749924 on Build GRCh38
rs750902601 Chr.1: 161749951-161749951 on Build GRCh38
rs750506910 Chr.1: 161749968-161749969 on Build GRCh38
rs367765196 Chr.1: 161749937-161749937 on Build GRCh38
rs1018850363 Chr.1: 161749978-161749978 on Build GRCh38
rs773946443 Chr.1: 161750008-161750008 on Build GRCh38
rs972980648 Chr.1: 161750062-161750062 on Build GRCh38
rs35106830 Chr.1: 161749953-161749953 on Build GRCh38
rs764702245 Chr.1: 161750126-161750126 on Build GRCh38
rs940155998 Chr.1: 161750040-161750040 on Build GRCh38
rs1032836652 Chr.1: 161749965-161749965 on Build GRCh38
rs139051375 Chr.1: 161749990-161749990 on Build GRCh38
rs756461045 Chr.1: 161750049-161750049 on Build GRCh38
rs1044063731 Chr.1: 161750088-161750088 on Build GRCh38
rs752113173 Chr.1: 161749944-161749944 on Build GRCh38
rs1006004091 Chr.1: 161750102-161750102 on Build GRCh38
rs745845653 Chr.1: 161750096-161750096 on Build GRCh38
rs1047194626 Chr.1: 161750042-161750042 on Build GRCh38
rs138443680 Chr.1: 161750021-161750021 on Build GRCh38
rs539052931 Chr.1: 161749917-161749917 on Build GRCh38
rs773260246 Chr.1: 161749915-161749915 on Build GRCh38
rs149171825 Chr.1: 161749945-161749945 on Build GRCh38
rs775218704 Chr.1: 161750098-161750098 on Build GRCh38
rs912953847 Chr.1: 161749989-161749989 on Build GRCh38
rs397863929 Chr.1: 161749917-161749917 on Build GRCh38
rs758973283 Chr.1: 161749966-161749966 on Build GRCh38
rs773491849 Chr.1: 161750004-161750004 on Build GRCh38
rs751830896 Chr.1: 161750035-161750035 on Build GRCh38
rs530478292 Chr.1: 161750110-161750110 on Build GRCh38
rs752961173 Chr.1: 161750047-161750047 on Build GRCh38
rs766270697 Chr.1: 161749922-161749922 on Build GRCh38
rs765322308 Chr.1: 161749920-161749961 on Build GRCh38
rs373896135 Chr.1: 161749967-161749967 on Build GRCh38
rs760741271 Chr.1: 161749929-161749929 on Build GRCh38
rs761344690 Chr.1: 161750125-161750125 on Build GRCh38
rs371992106 Chr.1: 161749975-161749975 on Build GRCh38
rs200380226 Chr.1: 161750039-161750039 on Build GRCh38
rs954483708 Chr.1: 161749976-161749976 on Build GRCh38
rs145790817 Chr.1: 161750009-161750009 on Build GRCh38
rs559426253 Chr.1: 161750019-161750019 on Build GRCh38
rs895634989 Chr.1: 161750032-161750032 on Build GRCh38
rs781449881 Chr.1: 161750043-161750043 on Build GRCh38
rs771871393 Chr.1: 161750097-161750097 on Build GRCh38
rs767200899 Chr.1: 161749941-161749941 on Build GRCh38
rs770947784 Chr.1: 161750075-161750075 on Build GRCh38
rs148273673 Chr.1: 161750092-161750092 on Build GRCh38
rs759050136 Chr.1: 161749936-161749936 on Build GRCh38
rs373331530 Chr.1: 161750014-161750014 on Build GRCh38
rs372640173 Chr.1: 161750007-161750007 on Build GRCh38
rs749222428 Chr.1: 161750005-161750005 on Build GRCh38
rs747414191 Chr.1: 161749973-161749973 on Build GRCh38
rs555296043 Chr.1: 161749992-161749992 on Build GRCh38
rs137874739 Chr.1: 161750031-161750031 on Build GRCh38
rs869209328 Chr.1: 161749918-161749918 on Build GRCh38
rs540812291 Chr.1: 161750018-161750018 on Build GRCh38

View Full Product Details