Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 109308923-109309176 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPPA2
Entrez Gene ID
151871
Gene Name
developmental pluripotency associated 2
Gene Aliases
CT100, ECAT15-2, PESCRG1
UniGene
Hs.351113
Gene Chromosome Location
Chr.3: 109293788-109316553 on Build GRCh38
Transcripts
NM_138815.3 XM_011512443.1 XM_011512444.2 XM_011512445.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2917289 c.382C>G p.P128A Chr.3: 109309040-109309040 on Build GRCh38
6162758 c.376G>T p.A126S Chr.3: 109309046-109309046 on Build GRCh38
1632698 c.392G>A p.R131Q Chr.3: 109309030-109309030 on Build GRCh38
7141830 c.348C>A p.I116I Chr.3: 109309074-109309074 on Build GRCh38
5918062 c.343-6C>T p.? Chr.3: 109309085-109309085 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1013835809 Chr.3: 109309059-109309059 on Build GRCh38
rs758443749 Chr.3: 109309052-109309052 on Build GRCh38
rs7611871 Chr.3: 109308973-109308973 on Build GRCh38
rs748603724 Chr.3: 109309096-109309096 on Build GRCh38
rs892151744 Chr.3: 109309101-109309105 on Build GRCh38
rs775903324 Chr.3: 109308976-109308976 on Build GRCh38
rs769190767 Chr.3: 109308961-109308961 on Build GRCh38
rs749652172 Chr.3: 109309074-109309074 on Build GRCh38
rs779676305 Chr.3: 109309058-109309058 on Build GRCh38
rs147277053 Chr.3: 109309150-109309150 on Build GRCh38
rs754783732 Chr.3: 109309040-109309040 on Build GRCh38
rs772729517 Chr.3: 109309003-109309003 on Build GRCh38
rs774728963 Chr.3: 109309095-109309095 on Build GRCh38
rs756330521 Chr.3: 109309066-109309066 on Build GRCh38
rs929270374 Chr.3: 109308992-109308992 on Build GRCh38
rs9809030 Chr.3: 109309030-109309030 on Build GRCh38
rs753659122 Chr.3: 109309031-109309031 on Build GRCh38
rs191956451 Chr.3: 109308959-109308959 on Build GRCh38
rs780208223 Chr.3: 109309073-109309073 on Build GRCh38
rs750157965 Chr.3: 109308951-109308951 on Build GRCh38
rs751364752 Chr.3: 109309089-109309089 on Build GRCh38
rs1002313211 Chr.3: 109309016-109309016 on Build GRCh38
rs200993272 Chr.3: 109309062-109309062 on Build GRCh38
rs887832090 Chr.3: 109309014-109309014 on Build GRCh38
rs762311820 Chr.3: 109308994-109308994 on Build GRCh38
rs985669018 Chr.3: 109308962-109308962 on Build GRCh38
rs769934263 Chr.3: 109308975-109308975 on Build GRCh38
rs768572996 Chr.3: 109309139-109309141 on Build GRCh38
rs760166607 Chr.3: 109309105-109309105 on Build GRCh38
rs910533288 Chr.3: 109309034-109309034 on Build GRCh38
rs72935076 Chr.3: 109309097-109309097 on Build GRCh38
rs201912260 Chr.3: 109309050-109309050 on Build GRCh38
rs759234368 Chr.3: 109309119-109309119 on Build GRCh38
rs746920221 Chr.3: 109309103-109309108 on Build GRCh38
rs912415712 Chr.3: 109308971-109308971 on Build GRCh38
rs868458879 Chr.3: 109309037-109309037 on Build GRCh38
rs376565430 Chr.3: 109309002-109309002 on Build GRCh38
rs765088051 Chr.3: 109309121-109309121 on Build GRCh38
rs766037517 Chr.3: 109309117-109309117 on Build GRCh38
rs765141410 Chr.3: 109309045-109309045 on Build GRCh38
rs752564717 Chr.3: 109309123-109309123 on Build GRCh38
rs372379947 Chr.3: 109308983-109308983 on Build GRCh38
rs924983219 Chr.3: 109308952-109308952 on Build GRCh38
rs370779235 Chr.3: 109309129-109309129 on Build GRCh38
rs760349846 Chr.3: 109309004-109309004 on Build GRCh38
rs781374715 Chr.3: 109309056-109309056 on Build GRCh38
rs867289548 Chr.3: 109309153-109309153 on Build GRCh38
rs376971262 Chr.3: 109308972-109308972 on Build GRCh38
rs764168482 Chr.3: 109309148-109309148 on Build GRCh38
rs772674651 Chr.3: 109309104-109309104 on Build GRCh38
rs377415113 Chr.3: 109309118-109309118 on Build GRCh38

View Full Product Details