Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 8599392-8599617 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPZ
Entrez Gene ID
8532
Gene Name
carboxypeptidase Z
Gene Aliases
-
UniGene
Hs.61995
Gene Chromosome Location
Chr.4: 8592660-8619761 on Build GRCh38
Transcripts
NM_001014447.2 NM_001014448.2 NM_003652.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7276122 c.103C>T p.P35S Chr.4: 8599467-8599467 on Build GRCh38
1670898 c.95G>A p.C32Y Chr.4: 8599459-8599459 on Build GRCh38
4674522 c.102G>A p.R34R Chr.4: 8599466-8599466 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs562883635 Chr.4: 8599547-8599547 on Build GRCh38
rs1006577397 Chr.4: 8599526-8599526 on Build GRCh38
rs771709153 Chr.4: 8599490-8599490 on Build GRCh38
rs959603747 Chr.4: 8599588-8599588 on Build GRCh38
rs763180250 Chr.4: 8599498-8599498 on Build GRCh38
rs376032947 Chr.4: 8599449-8599449 on Build GRCh38
rs771176077 Chr.4: 8599505-8599505 on Build GRCh38
rs759674551 Chr.4: 8599445-8599445 on Build GRCh38
rs763518903 Chr.4: 8599537-8599537 on Build GRCh38
rs776781355 Chr.4: 8599591-8599591 on Build GRCh38
rs371453540 Chr.4: 8599419-8599419 on Build GRCh38
rs369837263 Chr.4: 8599535-8599535 on Build GRCh38
rs776874069 Chr.4: 8599509-8599509 on Build GRCh38
rs377044104 Chr.4: 8599531-8599531 on Build GRCh38
rs754490047 Chr.4: 8599555-8599555 on Build GRCh38
rs779629312 Chr.4: 8599484-8599484 on Build GRCh38
rs1035586987 Chr.4: 8599586-8599586 on Build GRCh38
rs1025230105 Chr.4: 8599453-8599453 on Build GRCh38
rs41289049 Chr.4: 8599551-8599551 on Build GRCh38
rs768079752 Chr.4: 8599503-8599503 on Build GRCh38
rs557791844 Chr.4: 8599414-8599414 on Build GRCh38
rs574181492 Chr.4: 8599485-8599485 on Build GRCh38
rs201983596 Chr.4: 8599418-8599418 on Build GRCh38
rs746800732 Chr.4: 8599574-8599574 on Build GRCh38
rs776691412 Chr.4: 8599443-8599443 on Build GRCh38
rs759929439 Chr.4: 8599440-8599441 on Build GRCh38
rs765319818 Chr.4: 8599527-8599527 on Build GRCh38
rs544789422 Chr.4: 8599542-8599542 on Build GRCh38
rs773067438 Chr.4: 8599496-8599496 on Build GRCh38
rs11723641 Chr.4: 8599571-8599571 on Build GRCh38
rs764574237 Chr.4: 8599539-8599539 on Build GRCh38
rs368153695 Chr.4: 8599450-8599450 on Build GRCh38
rs367578052 Chr.4: 8599415-8599415 on Build GRCh38
rs778580838 Chr.4: 8599560-8599560 on Build GRCh38
rs201923255 Chr.4: 8599476-8599476 on Build GRCh38
rs372399181 Chr.4: 8599471-8599471 on Build GRCh38
rs200031119 Chr.4: 8599522-8599522 on Build GRCh38
rs760967912 Chr.4: 8599436-8599436 on Build GRCh38
rs933940878 Chr.4: 8599482-8599482 on Build GRCh38
rs766306642 Chr.4: 8599441-8599441 on Build GRCh38
rs758354338 Chr.4: 8599570-8599570 on Build GRCh38
rs753577762 Chr.4: 8599553-8599553 on Build GRCh38
rs369712443 Chr.4: 8599536-8599536 on Build GRCh38
rs199962135 Chr.4: 8599461-8599461 on Build GRCh38
rs993761594 Chr.4: 8599550-8599550 on Build GRCh38
rs560135764 Chr.4: 8599583-8599583 on Build GRCh38
rs748120542 Chr.4: 8599489-8599489 on Build GRCh38
rs746850447 Chr.4: 8599501-8599501 on Build GRCh38
rs768769680 Chr.4: 8599500-8599500 on Build GRCh38
rs748034446 Chr.4: 8599565-8599565 on Build GRCh38
rs375261043 Chr.4: 8599495-8599495 on Build GRCh38
rs764530556 Chr.4: 8599463-8599463 on Build GRCh38

View Full Product Details