Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 103375665-103375920 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTHRC1
Entrez Gene ID
115908
Gene Name
collagen triple helix repeat containing 1
Gene Aliases
-
UniGene
Hs.405614
Gene Chromosome Location
Chr.8: 103371515-103383005 on Build GRCh38
Transcripts
NM_001256099.1 NM_138455.3 XM_011516824.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5142921 c.206G>A p.R69Q Chr.8: 103375835-103375835 on Build GRCh38
1158113 c.167T>C p.L56S Chr.8: 103375754-103375754 on Build GRCh38
4936364 c.274T>A p.Y92N Chr.8: 103375903-103375903 on Build GRCh38
7137811 c.295G>C p.E99Q Chr.8: 103375882-103375882 on Build GRCh38
1094470 c.327T>C p.C109C Chr.8: 103375914-103375914 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145881116 Chr.8: 103375834-103375834 on Build GRCh38
rs759365776 Chr.8: 103375792-103375792 on Build GRCh38
rs983881049 Chr.8: 103375897-103375897 on Build GRCh38
rs781609744 Chr.8: 103375773-103375773 on Build GRCh38
rs889397221 Chr.8: 103375762-103375762 on Build GRCh38
rs187299788 Chr.8: 103375785-103375785 on Build GRCh38
rs905171722 Chr.8: 103375704-103375704 on Build GRCh38
rs756369288 Chr.8: 103375875-103375875 on Build GRCh38
rs376472345 Chr.8: 103375866-103375866 on Build GRCh38
rs750617028 Chr.8: 103375890-103375891 on Build GRCh38
rs781710038 Chr.8: 103375846-103375846 on Build GRCh38
rs901866266 Chr.8: 103375718-103375718 on Build GRCh38
rs1033593807 Chr.8: 103375860-103375860 on Build GRCh38
rs113614259 Chr.8: 103375782-103375782 on Build GRCh38
rs369008583 Chr.8: 103375786-103375786 on Build GRCh38
rs748382570 Chr.8: 103375780-103375780 on Build GRCh38
rs145838701 Chr.8: 103375878-103375878 on Build GRCh38
rs1036287767 Chr.8: 103375717-103375717 on Build GRCh38
rs780649365 Chr.8: 103375725-103375725 on Build GRCh38
rs149006532 Chr.8: 103375864-103375864 on Build GRCh38
rs767061518 Chr.8: 103375835-103375835 on Build GRCh38
rs149662990 Chr.8: 103375807-103375807 on Build GRCh38
rs775098206 Chr.8: 103375797-103375797 on Build GRCh38
rs747437502 Chr.8: 103375767-103375767 on Build GRCh38
rs778869312 Chr.8: 103375888-103375888 on Build GRCh38
rs373203731 Chr.8: 103375811-103375811 on Build GRCh38
rs1001036925 Chr.8: 103375775-103375775 on Build GRCh38
rs147175675 Chr.8: 103375781-103375781 on Build GRCh38
rs531041475 Chr.8: 103375724-103375724 on Build GRCh38
rs753249581 Chr.8: 103375709-103375709 on Build GRCh38
rs201468963 Chr.8: 103375746-103375746 on Build GRCh38
rs767275808 Chr.8: 103375796-103375796 on Build GRCh38
rs753401752 Chr.8: 103375812-103375812 on Build GRCh38
rs770966489 Chr.8: 103375881-103375881 on Build GRCh38
rs761868024 Chr.8: 103375704-103375705 on Build GRCh38
rs755392681 Chr.8: 103375770-103375770 on Build GRCh38
rs749448992 Chr.8: 103375880-103375880 on Build GRCh38
rs1050683688 Chr.8: 103375756-103375756 on Build GRCh38
rs755445506 Chr.8: 103375843-103375843 on Build GRCh38
rs767655755 Chr.8: 103375709-103375719 on Build GRCh38
rs763680532 Chr.8: 103375696-103375696 on Build GRCh38
rs752084339 Chr.8: 103375842-103375842 on Build GRCh38

View Full Product Details