Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 126730436-126730699 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHCHD6
Entrez Gene ID
84303
Gene Name
coiled-coil-helix-coiled-coil-helix domain containing 6
Gene Aliases
CHCM1, Mic25, PPP1R23
UniGene
Hs.518119
Gene Chromosome Location
Chr.3: 126704220-126960420 on Build GRCh38
Transcripts
NM_001320610.1 NM_032343.2 XM_011513234.1 XM_011513235.2 XM_017007320.1 XM_017007321.1 XM_017007322.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5695615 c.210C>T p.P70P Chr.3: 126730574-126730574 on Build GRCh38
6054842 c.203C>T p.T68I Chr.3: 126730567-126730567 on Build GRCh38
4961383 c.245G>A p.G82E Chr.3: 126730609-126730609 on Build GRCh38
5918020 c.197-8C>T p.? Chr.3: 126730553-126730553 on Build GRCh38
6544780 c.230G>T p.G77V Chr.3: 126730594-126730594 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs756052525 Chr.3: 126730514-126730514 on Build GRCh38
rs750559434 Chr.3: 126730611-126730611 on Build GRCh38
rs780911382 Chr.3: 126730510-126730510 on Build GRCh38
rs778607363 Chr.3: 126730636-126730636 on Build GRCh38
rs536672538 Chr.3: 126730499-126730499 on Build GRCh38
rs935246111 Chr.3: 126730556-126730556 on Build GRCh38
rs577021869 Chr.3: 126730575-126730575 on Build GRCh38
rs547863510 Chr.3: 126730469-126730469 on Build GRCh38
rs770159472 Chr.3: 126730654-126730654 on Build GRCh38
rs754952754 Chr.3: 126730634-126730634 on Build GRCh38
rs377407417 Chr.3: 126730518-126730518 on Build GRCh38
rs764220748 Chr.3: 126730591-126730591 on Build GRCh38
rs201019715 Chr.3: 126730657-126730657 on Build GRCh38
rs200059535 Chr.3: 126730571-126730571 on Build GRCh38
rs746578837 Chr.3: 126730643-126730643 on Build GRCh38
rs767755060 Chr.3: 126730610-126730610 on Build GRCh38
rs1051531758 Chr.3: 126730458-126730458 on Build GRCh38
rs765482402 Chr.3: 126730572-126730572 on Build GRCh38
rs569663852 Chr.3: 126730477-126730477 on Build GRCh38
rs373049688 Chr.3: 126730511-126730511 on Build GRCh38
rs776911177 Chr.3: 126730559-126730559 on Build GRCh38
rs768286309 Chr.3: 126730531-126730531 on Build GRCh38
rs778888400 Chr.3: 126730673-126730674 on Build GRCh38
rs1042530124 Chr.3: 126730565-126730565 on Build GRCh38
rs763397629 Chr.3: 126730579-126730579 on Build GRCh38
rs774461224 Chr.3: 126730665-126730665 on Build GRCh38
rs149996587 Chr.3: 126730609-126730609 on Build GRCh38
rs747673134 Chr.3: 126730638-126730638 on Build GRCh38
rs750660150 Chr.3: 126730677-126730677 on Build GRCh38
rs760750331 Chr.3: 126730678-126730678 on Build GRCh38
rs771724176 Chr.3: 126730551-126730551 on Build GRCh38
rs199956149 Chr.3: 126730664-126730664 on Build GRCh38
rs761972381 Chr.3: 126730669-126730669 on Build GRCh38
rs751631378 Chr.3: 126730604-126730604 on Build GRCh38
rs747822977 Chr.3: 126730546-126730546 on Build GRCh38
rs771008614 Chr.3: 126730539-126730542 on Build GRCh38
rs773939758 Chr.3: 126730539-126730539 on Build GRCh38
rs777514199 Chr.3: 126730642-126730642 on Build GRCh38
rs771437167 Chr.3: 126730639-126730639 on Build GRCh38
rs775823281 Chr.3: 126730656-126730656 on Build GRCh38
rs201950291 Chr.3: 126730625-126730625 on Build GRCh38
rs749251177 Chr.3: 126730631-126730631 on Build GRCh38
rs878954049 Chr.3: 126730577-126730577 on Build GRCh38
rs749195967 Chr.3: 126730520-126730520 on Build GRCh38
rs762397374 Chr.3: 126730670-126730670 on Build GRCh38
rs755968092 Chr.3: 126730616-126730616 on Build GRCh38

View Full Product Details